Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157325678-157326213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMN
Entrez Gene ID
57471
Gene Name
ermin
Gene Aliases
JN, KIAA1189
UniGene
Hs.443894
Gene Chromosome Location
Chr.2: 157318613-157327713 on Build GRCh38
Transcripts
NM_001009959.2 NM_001304344.1 NM_001304345.1 NM_001304346.1 NM_020711.2

SNP Detail

SNP ID Chromosome Location
rs990964187 Chr.2: 157326062-157326062 on Build GRCh38
rs546232982 Chr.2: 157326142-157326142 on Build GRCh38
rs890370853 Chr.2: 157325782-157325782 on Build GRCh38
rs145602756 Chr.2: 157325855-157325855 on Build GRCh38
rs960731001 Chr.2: 157326050-157326050 on Build GRCh38
rs534403831 Chr.2: 157325745-157325745 on Build GRCh38
rs1010968253 Chr.2: 157325902-157325902 on Build GRCh38
rs796764315 Chr.2: 157325965-157325965 on Build GRCh38
rs781090811 Chr.2: 157326100-157326100 on Build GRCh38
rs755663539 Chr.2: 157326185-157326185 on Build GRCh38
rs562821780 Chr.2: 157325889-157325889 on Build GRCh38
rs763752920 Chr.2: 157325719-157325719 on Build GRCh38
rs920353599 Chr.2: 157326154-157326154 on Build GRCh38
rs1022366528 Chr.2: 157325888-157325888 on Build GRCh38
rs762056061 Chr.2: 157325717-157325718 on Build GRCh38
rs909652850 Chr.2: 157326125-157326125 on Build GRCh38
rs956578034 Chr.2: 157325801-157325801 on Build GRCh38
rs372512094 Chr.2: 157325713-157325713 on Build GRCh38
rs556538167 Chr.2: 157326084-157326084 on Build GRCh38
rs928242297 Chr.2: 157325983-157325983 on Build GRCh38
rs926582965 Chr.2: 157325909-157325909 on Build GRCh38
rs928997405 Chr.2: 157326168-157326168 on Build GRCh38
rs755698964 Chr.2: 157325717-157325717 on Build GRCh38
rs967076028 Chr.2: 157325952-157325952 on Build GRCh38
rs960977427 Chr.2: 157326041-157326041 on Build GRCh38
rs993430118 Chr.2: 157325735-157325735 on Build GRCh38
rs980798422 Chr.2: 157325893-157325893 on Build GRCh38
rs796340617 Chr.2: 157325774-157325774 on Build GRCh38
rs916716917 Chr.2: 157326093-157326093 on Build GRCh38
rs567606898 Chr.2: 157325956-157325956 on Build GRCh38
rs757280167 Chr.2: 157325808-157325808 on Build GRCh38
rs865890820 Chr.2: 157325732-157325732 on Build GRCh38
rs964809366 Chr.2: 157325891-157325891 on Build GRCh38
rs535110137 Chr.2: 157326071-157326071 on Build GRCh38
rs752329649 Chr.2: 157325714-157325714 on Build GRCh38
rs919496411 Chr.2: 157326074-157326074 on Build GRCh38
rs201101603 Chr.2: 157325706-157325706 on Build GRCh38

View Full Product Details