Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168864705-168865205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOSTRIN
Entrez Gene ID
115677
Gene Name
nitric oxide synthase trafficking
Gene Aliases
DaIP2
UniGene
Hs.189780
Gene Chromosome Location
Chr.2: 168786539-168865339 on Build GRCh38
Transcripts
NM_001039724.3 NM_001171631.1 NM_001171632.1 NM_052946.3 XM_005246270.1 XM_011510543.2 XM_017003279.1 XM_017003280.1 XM_011511516.2

Gene Symbol
SPC25
Entrez Gene ID
57405
Gene Name
SPC25, NDC80 kinetochore complex component
Gene Aliases
AD024, SPBC25, hSpc25
UniGene
Hs.421956
Gene Chromosome Location
Chr.2: 168865045-168890443 on Build GRCh38
Transcripts
NM_001039724.3 NM_001171631.1 NM_001171632.1 NM_052946.3 XM_005246270.1 XM_011510543.2 XM_017003279.1 XM_017003280.1 XM_011511516.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
262440 c.1414G>T p.E472* Chr.2: 168864863-168864863 on Build GRCh38
1641692 c.1447_1448insA p.G485fs*>23 Chr.2: 168864896-168864897 on Build GRCh38
1669386 c.1453G>T p.G485C Chr.2: 168864902-168864902 on Build GRCh38
4086536 c.1456C>T p.H486Y Chr.2: 168864905-168864905 on Build GRCh38
5965198 c.1648G>C p.E550Q Chr.2: 168864926-168864926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773751937 Chr.2: 168864913-168864913 on Build GRCh38
rs184878852 Chr.2: 168865153-168865153 on Build GRCh38
rs756061995 Chr.2: 168864796-168864796 on Build GRCh38
rs531626284 Chr.2: 168864866-168864866 on Build GRCh38
rs755255348 Chr.2: 168864875-168864875 on Build GRCh38
rs954316693 Chr.2: 168864830-168864830 on Build GRCh38
rs865861519 Chr.2: 168864894-168864895 on Build GRCh38
rs371895459 Chr.2: 168864834-168864834 on Build GRCh38
rs181853503 Chr.2: 168865113-168865113 on Build GRCh38
rs999880231 Chr.2: 168864770-168864773 on Build GRCh38
rs764115930 Chr.2: 168864865-168864865 on Build GRCh38
rs375031439 Chr.2: 168864986-168864986 on Build GRCh38
rs375288149 Chr.2: 168864983-168864983 on Build GRCh38
rs755688570 Chr.2: 168864821-168864822 on Build GRCh38
rs479661 Chr.2: 168864867-168864867 on Build GRCh38
rs547483781 Chr.2: 168864880-168864880 on Build GRCh38
rs767134914 Chr.2: 168864815-168864816 on Build GRCh38
rs750791844 Chr.2: 168864929-168864930 on Build GRCh38
rs773303748 Chr.2: 168864833-168864833 on Build GRCh38
rs903785433 Chr.2: 168864766-168864766 on Build GRCh38
rs372892123 Chr.2: 168864856-168864856 on Build GRCh38
rs760424889 Chr.2: 168864861-168864861 on Build GRCh38
rs967331463 Chr.2: 168865061-168865061 on Build GRCh38
rs796194578 Chr.2: 168865128-168865129 on Build GRCh38
rs949057940 Chr.2: 168864953-168864953 on Build GRCh38
rs997953056 Chr.2: 168865077-168865077 on Build GRCh38
rs202072618 Chr.2: 168864789-168864789 on Build GRCh38
rs767102256 Chr.2: 168864854-168864854 on Build GRCh38
rs779475008 Chr.2: 168864921-168864922 on Build GRCh38
rs1030656677 Chr.2: 168864777-168864777 on Build GRCh38
rs377563077 Chr.2: 168864917-168864917 on Build GRCh38
rs749923661 Chr.2: 168865056-168865056 on Build GRCh38
rs918844801 Chr.2: 168865081-168865081 on Build GRCh38
rs757176411 Chr.2: 168865003-168865003 on Build GRCh38
rs766364348 Chr.2: 168864870-168864870 on Build GRCh38
rs754642185 Chr.2: 168865159-168865159 on Build GRCh38
rs377429537 Chr.2: 168864980-168864980 on Build GRCh38
rs760255428 Chr.2: 168864865-168864867 on Build GRCh38
rs776698855 Chr.2: 168864935-168864935 on Build GRCh38
rs779681317 Chr.2: 168865104-168865104 on Build GRCh38
rs767898158 Chr.2: 168864967-168864967 on Build GRCh38
rs199524046 Chr.2: 168864975-168864975 on Build GRCh38
rs773541469 Chr.2: 168864847-168864847 on Build GRCh38
rs939386114 Chr.2: 168864864-168864864 on Build GRCh38
rs376851617 Chr.2: 168864809-168864809 on Build GRCh38
rs1030573945 Chr.2: 168865100-168865100 on Build GRCh38
rs775271443 Chr.2: 168864857-168864857 on Build GRCh38
rs763956548 Chr.2: 168864798-168864798 on Build GRCh38
rs760514633 Chr.2: 168864919-168864919 on Build GRCh38
rs189095191 Chr.2: 168864836-168864836 on Build GRCh38
rs527812643 Chr.2: 168865014-168865014 on Build GRCh38
rs909343713 Chr.2: 168865133-168865133 on Build GRCh38
rs577581210 Chr.2: 168865168-168865168 on Build GRCh38
rs758363661 Chr.2: 168864807-168864807 on Build GRCh38
rs117394461 Chr.2: 168865054-168865054 on Build GRCh38
rs777982217 Chr.2: 168864979-168864979 on Build GRCh38
rs761628998 Chr.2: 168865020-168865020 on Build GRCh38
rs963243019 Chr.2: 168865058-168865058 on Build GRCh38
rs752189148 Chr.2: 168864857-168864857 on Build GRCh38
rs759411961 Chr.2: 168864783-168864783 on Build GRCh38
rs745407796 Chr.2: 168864988-168864988 on Build GRCh38
rs758730682 Chr.2: 168864976-168864977 on Build GRCh38
rs999448810 Chr.2: 168864973-168864973 on Build GRCh38
rs963560941 Chr.2: 168865136-168865136 on Build GRCh38
rs903710470 Chr.2: 168864956-168864956 on Build GRCh38
rs10181 Chr.2: 168864937-168864937 on Build GRCh38
rs752932696 Chr.2: 168864974-168864974 on Build GRCh38
rs770528233 Chr.2: 168864904-168864904 on Build GRCh38
rs765124040 Chr.2: 168865013-168865013 on Build GRCh38
rs748543503 Chr.2: 168864884-168864884 on Build GRCh38
rs754372560 Chr.2: 168864888-168864891 on Build GRCh38
rs756522311 Chr.2: 168864885-168864885 on Build GRCh38
rs369879274 Chr.2: 168864826-168864826 on Build GRCh38
rs3931 Chr.2: 168864871-168864871 on Build GRCh38
rs541060772 Chr.2: 168865059-168865059 on Build GRCh38
rs1011588198 Chr.2: 168865050-168865050 on Build GRCh38
rs765154950 Chr.2: 168864947-168864947 on Build GRCh38
rs751718907 Chr.2: 168864948-168864948 on Build GRCh38
rs990196601 Chr.2: 168864776-168864776 on Build GRCh38
rs62176770 Chr.2: 168865130-168865130 on Build GRCh38
rs763454012 Chr.2: 168864868-168864868 on Build GRCh38
rs759638259 Chr.2: 168864962-168864962 on Build GRCh38
rs776294886 Chr.2: 168865015-168865015 on Build GRCh38
rs953170793 Chr.2: 168865034-168865034 on Build GRCh38
rs1043995124 Chr.2: 168864749-168864749 on Build GRCh38
rs371041874 Chr.2: 168864930-168864930 on Build GRCh38
rs960653550 Chr.2: 168865094-168865094 on Build GRCh38
rs745319752 Chr.2: 168864914-168864914 on Build GRCh38
rs771745688 Chr.2: 168864916-168864916 on Build GRCh38
rs778627556 Chr.2: 168865115-168865115 on Build GRCh38
rs778381652 Chr.2: 168864893-168864893 on Build GRCh38
rs767291870 Chr.2: 168864787-168864787 on Build GRCh38
rs367986666 Chr.2: 168865002-168865002 on Build GRCh38
rs748633996 Chr.2: 168864745-168864745 on Build GRCh38
rs757832201 Chr.2: 168864895-168864896 on Build GRCh38
rs771551042 Chr.2: 168864993-168864993 on Build GRCh38
rs564825094 Chr.2: 168864868-168864868 on Build GRCh38
rs373131496 Chr.2: 168864895-168864895 on Build GRCh38
rs949074773 Chr.2: 168865172-168865172 on Build GRCh38
rs781699436 Chr.2: 168864835-168864835 on Build GRCh38
rs770453222 Chr.2: 168864840-168864840 on Build GRCh38
rs780450733 Chr.2: 168864977-168864978 on Build GRCh38
rs373502252 Chr.2: 168864851-168864851 on Build GRCh38
rs750550170 Chr.2: 168864802-168864802 on Build GRCh38
rs747169222 Chr.2: 168864831-168864831 on Build GRCh38
rs370163673 Chr.2: 168864982-168864982 on Build GRCh38
rs368861114 Chr.2: 168864853-168864853 on Build GRCh38
rs184365209 Chr.2: 168864743-168864743 on Build GRCh38
rs925135187 Chr.2: 168865135-168865135 on Build GRCh38
rs748875287 Chr.2: 168864848-168864848 on Build GRCh38

View Full Product Details