Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168922861-168923356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB11
Entrez Gene ID
8647
Gene Name
ATP binding cassette subfamily B member 11
Gene Aliases
ABC16, BRIC2, BSEP, PFIC-2, PFIC2, PGY4, SPGP
UniGene
Hs.658439
Gene Chromosome Location
Chr.2: 168915468-169031396 on Build GRCh38
Transcripts
NM_003742.2 XM_006712817.3 XM_011512077.2 XM_011512078.2 XM_011512081.2 XM_017005165.1 XM_017005166.1 XM_017005167.1

SNP Detail

SNP ID Chromosome Location
rs752531055 Chr.2: 168923259-168923259 on Build GRCh38
rs887581211 Chr.2: 168923297-168923297 on Build GRCh38
rs1038819783 Chr.2: 168923322-168923322 on Build GRCh38
rs886055058 Chr.2: 168923089-168923089 on Build GRCh38
rs773735682 Chr.2: 168922995-168922995 on Build GRCh38
rs932215062 Chr.2: 168923205-168923205 on Build GRCh38
rs1019296334 Chr.2: 168923245-168923245 on Build GRCh38
rs775149398 Chr.2: 168923244-168923244 on Build GRCh38
rs575469714 Chr.2: 168923209-168923209 on Build GRCh38
rs496550 Chr.2: 168923202-168923202 on Build GRCh38
rs975738844 Chr.2: 168923233-168923233 on Build GRCh38
rs553314589 Chr.2: 168923090-168923090 on Build GRCh38
rs980264981 Chr.2: 168923140-168923140 on Build GRCh38
rs562545577 Chr.2: 168923135-168923135 on Build GRCh38
rs902869363 Chr.2: 168923222-168923222 on Build GRCh38
rs924091150 Chr.2: 168922895-168922895 on Build GRCh38
rs748632568 Chr.2: 168922885-168922885 on Build GRCh38
rs560085646 Chr.2: 168923306-168923306 on Build GRCh38
rs375854199 Chr.2: 168922914-168922914 on Build GRCh38
rs539768984 Chr.2: 168923052-168923052 on Build GRCh38
rs868790442 Chr.2: 168922936-168922936 on Build GRCh38
rs77005833 Chr.2: 168923206-168923206 on Build GRCh38
rs577137795 Chr.2: 168923024-168923024 on Build GRCh38
rs73014701 Chr.2: 168923223-168923223 on Build GRCh38
rs79130028 Chr.2: 168923182-168923182 on Build GRCh38
rs574099605 Chr.2: 168922891-168922892 on Build GRCh38
rs994430671 Chr.2: 168922981-168922981 on Build GRCh38
rs145586340 Chr.2: 168922922-168922922 on Build GRCh38
rs985194884 Chr.2: 168922985-168922985 on Build GRCh38
rs533233749 Chr.2: 168923249-168923249 on Build GRCh38
rs922155677 Chr.2: 168922931-168922931 on Build GRCh38
rs1050277879 Chr.2: 168923271-168923271 on Build GRCh38
rs957159673 Chr.2: 168923183-168923183 on Build GRCh38
rs147741834 Chr.2: 168923224-168923224 on Build GRCh38
rs897123303 Chr.2: 168922934-168922934 on Build GRCh38
rs1006035336 Chr.2: 168922919-168922919 on Build GRCh38
rs495714 Chr.2: 168923254-168923254 on Build GRCh38
rs1029157758 Chr.2: 168923328-168923328 on Build GRCh38
rs886055059 Chr.2: 168923113-168923113 on Build GRCh38
rs773303303 Chr.2: 168923026-168923026 on Build GRCh38
rs938460535 Chr.2: 168923068-168923068 on Build GRCh38

View Full Product Details