Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176190118-176190622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD1
Entrez Gene ID
3231
Gene Name
homeobox D1
Gene Aliases
HOX4, HOX4G, Hox-4.7
UniGene
Hs.83465
Gene Chromosome Location
Chr.2: 176188579-176190907 on Build GRCh38
Transcripts
NM_024501.2 XM_005246507.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1401457 c.983C>T p.S328L Chr.2: 176190138-176190138 on Build GRCh38
5036189 c.984G>A p.S328S Chr.2: 176190139-176190139 on Build GRCh38
6089104 c.980C>G p.P327R Chr.2: 176190135-176190135 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756253940 Chr.2: 176190145-176190145 on Build GRCh38
rs548649172 Chr.2: 176190147-176190147 on Build GRCh38
rs888059844 Chr.2: 176190303-176190303 on Build GRCh38
rs993004837 Chr.2: 176190351-176190351 on Build GRCh38
rs936414286 Chr.2: 176190423-176190423 on Build GRCh38
rs977775963 Chr.2: 176190378-176190378 on Build GRCh38
rs748063329 Chr.2: 176190167-176190168 on Build GRCh38
rs1051690330 Chr.2: 176190215-176190215 on Build GRCh38
rs896912153 Chr.2: 176190589-176190589 on Build GRCh38
rs556404052 Chr.2: 176190514-176190514 on Build GRCh38
rs867912242 Chr.2: 176190442-176190442 on Build GRCh38
rs960212621 Chr.2: 176190337-176190337 on Build GRCh38
rs771399938 Chr.2: 176190142-176190162 on Build GRCh38
rs966797006 Chr.2: 176190430-176190430 on Build GRCh38
rs913486087 Chr.2: 176190352-176190352 on Build GRCh38
rs936630532 Chr.2: 176190468-176190468 on Build GRCh38
rs1025483697 Chr.2: 176190405-176190405 on Build GRCh38
rs760253927 Chr.2: 176190180-176190180 on Build GRCh38
rs190962453 Chr.2: 176190431-176190431 on Build GRCh38
rs1004884057 Chr.2: 176190227-176190227 on Build GRCh38
rs1033722456 Chr.2: 176190344-176190344 on Build GRCh38
rs371579717 Chr.2: 176190169-176190169 on Build GRCh38
rs1051637456 Chr.2: 176190573-176190573 on Build GRCh38
rs994089886 Chr.2: 176190217-176190217 on Build GRCh38
rs778044406 Chr.2: 176190146-176190146 on Build GRCh38
rs768277968 Chr.2: 176190552-176190552 on Build GRCh38
rs992138463 Chr.2: 176190385-176190385 on Build GRCh38
rs1040689170 Chr.2: 176190572-176190572 on Build GRCh38
rs746868315 Chr.2: 176190161-176190161 on Build GRCh38
rs772269045 Chr.2: 176190191-176190192 on Build GRCh38
rs745772370 Chr.2: 176190168-176190168 on Build GRCh38
rs921734224 Chr.2: 176190540-176190540 on Build GRCh38
rs182463194 Chr.2: 176190571-176190571 on Build GRCh38
rs538295710 Chr.2: 176190443-176190443 on Build GRCh38
rs896861727 Chr.2: 176190203-176190203 on Build GRCh38
rs772151952 Chr.2: 176190178-176190178 on Build GRCh38
rs1006462065 Chr.2: 176190328-176190328 on Build GRCh38
rs963484188 Chr.2: 176190252-176190252 on Build GRCh38
rs6725515 Chr.2: 176190365-176190365 on Build GRCh38
rs567131327 Chr.2: 176190156-176190156 on Build GRCh38
rs199761075 Chr.2: 176190159-176190160 on Build GRCh38
rs186519463 Chr.2: 176190278-176190278 on Build GRCh38
rs1034459857 Chr.2: 176190308-176190308 on Build GRCh38
rs1016713139 Chr.2: 176190230-176190230 on Build GRCh38

View Full Product Details