Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 191039107-191039603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT4
Entrez Gene ID
6775
Gene Name
signal transducer and activator of transcription 4
Gene Aliases
SLEB11
UniGene
Hs.735572
Gene Chromosome Location
Chr.2: 191029576-191172684 on Build GRCh38
Transcripts
NM_001243835.1 NM_003151.3 XM_006712719.3 XM_011511705.2 XM_017004784.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4730439 c.1343C>A p.S448* Chr.2: 191039290-191039290 on Build GRCh38
1014165 c.1426G>A p.D476N Chr.2: 191039207-191039207 on Build GRCh38
3575613 c.1430C>T p.S477F Chr.2: 191039203-191039203 on Build GRCh38
3391355 c.1421C>T p.T474I Chr.2: 191039212-191039212 on Build GRCh38
3575612 c.1432C>A p.Q478K Chr.2: 191039201-191039201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759785386 Chr.2: 191039207-191039207 on Build GRCh38
rs1056145824 Chr.2: 191039380-191039380 on Build GRCh38
rs371804659 Chr.2: 191039160-191039160 on Build GRCh38
rs896932275 Chr.2: 191039365-191039365 on Build GRCh38
rs745898533 Chr.2: 191039320-191039320 on Build GRCh38
rs956219357 Chr.2: 191039395-191039395 on Build GRCh38
rs762476453 Chr.2: 191039241-191039241 on Build GRCh38
rs929844832 Chr.2: 191039562-191039562 on Build GRCh38
rs777544013 Chr.2: 191039306-191039306 on Build GRCh38
rs531690122 Chr.2: 191039506-191039506 on Build GRCh38
rs758127803 Chr.2: 191039305-191039305 on Build GRCh38
rs147842162 Chr.2: 191039368-191039368 on Build GRCh38
rs938451501 Chr.2: 191039550-191039550 on Build GRCh38
rs914845168 Chr.2: 191039492-191039492 on Build GRCh38
rs544508292 Chr.2: 191039285-191039285 on Build GRCh38
rs200929981 Chr.2: 191039235-191039235 on Build GRCh38
rs758592990 Chr.2: 191039163-191039163 on Build GRCh38
rs370226598 Chr.2: 191039347-191039347 on Build GRCh38
rs1030524333 Chr.2: 191039384-191039384 on Build GRCh38
rs775517900 Chr.2: 191039331-191039331 on Build GRCh38
rs778204330 Chr.2: 191039304-191039305 on Build GRCh38
rs988984058 Chr.2: 191039463-191039463 on Build GRCh38
rs752949877 Chr.2: 191039129-191039129 on Build GRCh38
rs749655316 Chr.2: 191039311-191039311 on Build GRCh38
rs530278225 Chr.2: 191039292-191039292 on Build GRCh38
rs144421302 Chr.2: 191039295-191039295 on Build GRCh38
rs868730606 Chr.2: 191039568-191039568 on Build GRCh38
rs964799096 Chr.2: 191039378-191039378 on Build GRCh38
rs770056176 Chr.2: 191039302-191039302 on Build GRCh38
rs752365784 Chr.2: 191039304-191039304 on Build GRCh38
rs368249753 Chr.2: 191039312-191039312 on Build GRCh38
rs146386562 Chr.2: 191039209-191039209 on Build GRCh38
rs769952683 Chr.2: 191039208-191039208 on Build GRCh38
rs549488539 Chr.2: 191039369-191039369 on Build GRCh38
rs980182110 Chr.2: 191039521-191039521 on Build GRCh38
rs756967396 Chr.2: 191039169-191039169 on Build GRCh38
rs749527470 Chr.2: 191039334-191039334 on Build GRCh38
rs1048222401 Chr.2: 191039571-191039571 on Build GRCh38
rs778632183 Chr.2: 191039265-191039265 on Build GRCh38
rs142491497 Chr.2: 191039220-191039220 on Build GRCh38
rs776636863 Chr.2: 191039191-191039191 on Build GRCh38
rs1023007984 Chr.2: 191039512-191039512 on Build GRCh38
rs148883745 Chr.2: 191039495-191039495 on Build GRCh38
rs770927651 Chr.2: 191039185-191039185 on Build GRCh38
rs1012501547 Chr.2: 191039497-191039497 on Build GRCh38
rs3024881 Chr.2: 191039561-191039561 on Build GRCh38
rs188120696 Chr.2: 191039558-191039558 on Build GRCh38
rs926948891 Chr.2: 191039546-191039546 on Build GRCh38
rs966133299 Chr.2: 191039573-191039573 on Build GRCh38
rs781303864 Chr.2: 191039319-191039319 on Build GRCh38
rs746939995 Chr.2: 191039309-191039309 on Build GRCh38
rs370416058 Chr.2: 191039159-191039159 on Build GRCh38
rs141331848 Chr.2: 191039296-191039296 on Build GRCh38

View Full Product Details