Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197523535-197523973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPE1-MOB4
Entrez Gene ID
100529241
Gene Name
HSPE1-MOB4 readthrough
Gene Aliases
HSPE1-PHOCN
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197499997-197553699 on Build GRCh38
Transcripts
NM_001202485.1 NM_001100819.2 NM_001204094.1 NM_015387.4 NM_199482.3

Gene Symbol
MOB4
Entrez Gene ID
25843
Gene Name
MOB family member 4, phocein
Gene Aliases
2C4D, CGI-95, MOB1, MOB3, MOBKL3, PHOCN, PREI3
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197515571-197553699 on Build GRCh38
Transcripts
NM_001202485.1 NM_001100819.2 NM_001204094.1 NM_015387.4 NM_199482.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7290789 c.61-6T>A p.? Chr.2: 197523618-197523618 on Build GRCh38
1404301 c.77C>A p.P26H Chr.2: 197523640-197523640 on Build GRCh38
1614100 c.76C>T p.P26S Chr.2: 197523639-197523639 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs928283360 Chr.2: 197523896-197523896 on Build GRCh38
rs750822367 Chr.2: 197523763-197523763 on Build GRCh38
rs779871901 Chr.2: 197523864-197523864 on Build GRCh38
rs997207747 Chr.2: 197523932-197523932 on Build GRCh38
rs768922790 Chr.2: 197523653-197523653 on Build GRCh38
rs372104828 Chr.2: 197523732-197523732 on Build GRCh38
rs566826097 Chr.2: 197523860-197523860 on Build GRCh38
rs950531182 Chr.2: 197523817-197523817 on Build GRCh38
rs773682236 Chr.2: 197523612-197523612 on Build GRCh38
rs767505239 Chr.2: 197523609-197523609 on Build GRCh38
rs762880404 Chr.2: 197523644-197523644 on Build GRCh38
rs928748686 Chr.2: 197523838-197523838 on Build GRCh38
rs750951017 Chr.2: 197523573-197523573 on Build GRCh38
rs929994561 Chr.2: 197523810-197523810 on Build GRCh38
rs893335512 Chr.2: 197523881-197523881 on Build GRCh38
rs570875102 Chr.2: 197523888-197523888 on Build GRCh38
rs1016164762 Chr.2: 197523849-197523849 on Build GRCh38
rs187543341 Chr.2: 197523800-197523800 on Build GRCh38
rs564231054 Chr.2: 197523873-197523873 on Build GRCh38
rs746874716 Chr.2: 197523598-197523598 on Build GRCh38
rs35917320 Chr.2: 197523602-197523603 on Build GRCh38
rs1046110083 Chr.2: 197523812-197523812 on Build GRCh38
rs540371188 Chr.2: 197523591-197523591 on Build GRCh38
rs774428873 Chr.2: 197523677-197523677 on Build GRCh38
rs373819349 Chr.2: 197523605-197523605 on Build GRCh38
rs375428652 Chr.2: 197523631-197523631 on Build GRCh38
rs202101608 Chr.2: 197523578-197523578 on Build GRCh38
rs368401341 Chr.2: 197523729-197523729 on Build GRCh38
rs769683553 Chr.2: 197523614-197523614 on Build GRCh38
rs757844625 Chr.2: 197523731-197523731 on Build GRCh38
rs777224040 Chr.2: 197523597-197523597 on Build GRCh38
rs751766766 Chr.2: 197523799-197523799 on Build GRCh38
rs1011741902 Chr.2: 197523884-197523884 on Build GRCh38
rs531093745 Chr.2: 197523869-197523869 on Build GRCh38
rs938456128 Chr.2: 197523897-197523897 on Build GRCh38
rs985947502 Chr.2: 197523748-197523748 on Build GRCh38
rs767969805 Chr.2: 197523711-197523711 on Build GRCh38
rs751005457 Chr.2: 197523712-197523712 on Build GRCh38
rs368168359 Chr.2: 197523730-197523730 on Build GRCh38
rs754582109 Chr.2: 197523575-197523575 on Build GRCh38
rs762323082 Chr.2: 197523705-197523705 on Build GRCh38
rs753783870 Chr.2: 197523933-197523933 on Build GRCh38
rs971990431 Chr.2: 197523779-197523779 on Build GRCh38
rs191221622 Chr.2: 197523832-197523832 on Build GRCh38
rs984686089 Chr.2: 197523846-197523846 on Build GRCh38
rs77654215 Chr.2: 197523944-197523944 on Build GRCh38
rs778369826 Chr.2: 197523735-197523736 on Build GRCh38
rs548880938 Chr.2: 197523828-197523828 on Build GRCh38
rs901296206 Chr.2: 197523791-197523791 on Build GRCh38
rs988925562 Chr.2: 197523931-197523931 on Build GRCh38
rs761295048 Chr.2: 197523717-197523717 on Build GRCh38
rs913418523 Chr.2: 197523934-197523934 on Build GRCh38
rs963168987 Chr.2: 197523773-197523773 on Build GRCh38
rs909073884 Chr.2: 197523754-197523754 on Build GRCh38
rs77102528 Chr.2: 197523606-197523606 on Build GRCh38
rs551207838 Chr.2: 197523874-197523874 on Build GRCh38
rs745777224 Chr.2: 197523607-197523607 on Build GRCh38
rs370712205 Chr.2: 197523586-197523586 on Build GRCh38

View Full Product Details