Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197535414-197535875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPE1-MOB4
Entrez Gene ID
100529241
Gene Name
HSPE1-MOB4 readthrough
Gene Aliases
HSPE1-PHOCN
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197499997-197553699 on Build GRCh38
Transcripts
NM_001202485.1 NM_001100819.2 NM_001204094.1 NM_015387.4 NM_199482.3

Gene Symbol
MOB4
Entrez Gene ID
25843
Gene Name
MOB family member 4, phocein
Gene Aliases
2C4D, CGI-95, MOB1, MOB3, MOBKL3, PHOCN, PREI3
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197515571-197553699 on Build GRCh38
Transcripts
NM_001202485.1 NM_001100819.2 NM_001204094.1 NM_015387.4 NM_199482.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7188116 c.140T>G p.I47R Chr.2: 197535546-197535546 on Build GRCh38
282842 c.215A>G p.E72G Chr.2: 197535621-197535621 on Build GRCh38
1404302 c.159T>C p.N53N Chr.2: 197535565-197535565 on Build GRCh38
6714092 c.148G>A p.D50N Chr.2: 197535554-197535554 on Build GRCh38
1014534 c.183T>C p.P61P Chr.2: 197535589-197535589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928861786 Chr.2: 197535469-197535469 on Build GRCh38
rs892983101 Chr.2: 197535594-197535594 on Build GRCh38
rs761055609 Chr.2: 197535563-197535563 on Build GRCh38
rs369325047 Chr.2: 197535818-197535818 on Build GRCh38
rs745617843 Chr.2: 197535512-197535512 on Build GRCh38
rs780769123 Chr.2: 197535625-197535625 on Build GRCh38
rs781000268 Chr.2: 197535510-197535510 on Build GRCh38
rs751032029 Chr.2: 197535602-197535602 on Build GRCh38
rs759890206 Chr.2: 197535581-197535581 on Build GRCh38
rs532365456 Chr.2: 197535634-197535634 on Build GRCh38
rs745505800 Chr.2: 197535712-197535712 on Build GRCh38
rs771545434 Chr.2: 197535717-197535717 on Build GRCh38
rs970309198 Chr.2: 197535438-197535438 on Build GRCh38
rs779785023 Chr.2: 197535518-197535518 on Build GRCh38
rs374956554 Chr.2: 197535612-197535612 on Build GRCh38
rs369012087 Chr.2: 197535666-197535666 on Build GRCh38
rs756772370 Chr.2: 197535841-197535841 on Build GRCh38
rs962889822 Chr.2: 197535761-197535761 on Build GRCh38
rs774761562 Chr.2: 197535522-197535522 on Build GRCh38
rs1016005451 Chr.2: 197535803-197535803 on Build GRCh38
rs933037445 Chr.2: 197535744-197535744 on Build GRCh38
rs937132419 Chr.2: 197535570-197535570 on Build GRCh38
rs1011338076 Chr.2: 197535600-197535600 on Build GRCh38
rs527993078 Chr.2: 197535770-197535770 on Build GRCh38
rs866825525 Chr.2: 197535440-197535440 on Build GRCh38
rs906062149 Chr.2: 197535807-197535807 on Build GRCh38
rs918735203 Chr.2: 197535776-197535776 on Build GRCh38
rs145016112 Chr.2: 197535788-197535788 on Build GRCh38
rs754289114 Chr.2: 197535573-197535573 on Build GRCh38
rs1043558836 Chr.2: 197535571-197535571 on Build GRCh38
rs779163218 Chr.2: 197535674-197535674 on Build GRCh38
rs1047396432 Chr.2: 197535757-197535757 on Build GRCh38
rs780094338 Chr.2: 197535661-197535661 on Build GRCh38
rs372788864 Chr.2: 197535719-197535719 on Build GRCh38
rs749449145 Chr.2: 197535519-197535519 on Build GRCh38
rs974334558 Chr.2: 197535517-197535518 on Build GRCh38
rs755853637 Chr.2: 197535656-197535656 on Build GRCh38
rs192173929 Chr.2: 197535508-197535508 on Build GRCh38
rs960106271 Chr.2: 197535474-197535474 on Build GRCh38
rs751193699 Chr.2: 197535503-197535503 on Build GRCh38
rs980407389 Chr.2: 197535468-197535468 on Build GRCh38
rs559959704 Chr.2: 197535802-197535802 on Build GRCh38
rs183999666 Chr.2: 197535716-197535716 on Build GRCh38
rs886205566 Chr.2: 197535763-197535763 on Build GRCh38
rs777540882 Chr.2: 197535485-197535485 on Build GRCh38
rs768602283 Chr.2: 197535520-197535520 on Build GRCh38
rs543741522 Chr.2: 197535460-197535460 on Build GRCh38
rs769780307 Chr.2: 197535515-197535515 on Build GRCh38
rs547028578 Chr.2: 197535747-197535747 on Build GRCh38
rs188729437 Chr.2: 197535828-197535828 on Build GRCh38
rs1002102851 Chr.2: 197535821-197535821 on Build GRCh38
rs372102734 Chr.2: 197535556-197535556 on Build GRCh38
rs947436125 Chr.2: 197535561-197535561 on Build GRCh38
rs772379044 Chr.2: 197535545-197535545 on Build GRCh38
rs763878186 Chr.2: 197535675-197535675 on Build GRCh38
rs559039956 Chr.2: 197535691-197535691 on Build GRCh38
rs778548646 Chr.2: 197535591-197535591 on Build GRCh38
rs997830528 Chr.2: 197535689-197535689 on Build GRCh38
rs762094211 Chr.2: 197535538-197535538 on Build GRCh38
rs766976397 Chr.2: 197535572-197535572 on Build GRCh38
rs574647598 Chr.2: 197535446-197535446 on Build GRCh38
rs750219281 Chr.2: 197535649-197535649 on Build GRCh38

View Full Product Details