Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202766127-202766635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM117B
Entrez Gene ID
150864
Gene Name
family with sequence similarity 117 member B
Gene Aliases
ALS2CR13
UniGene
Hs.471130
Gene Chromosome Location
Chr.2: 202635178-202769757 on Build GRCh38
Transcripts
NM_173511.3

SNP Detail

SNP ID Chromosome Location
rs910970623 Chr.2: 202766257-202766258 on Build GRCh38
rs1048774707 Chr.2: 202766351-202766351 on Build GRCh38
rs1014725809 Chr.2: 202766538-202766538 on Build GRCh38
rs558146075 Chr.2: 202766362-202766362 on Build GRCh38
rs371661189 Chr.2: 202766259-202766260 on Build GRCh38
rs928444163 Chr.2: 202766300-202766300 on Build GRCh38
rs371625897 Chr.2: 202766591-202766591 on Build GRCh38
rs1001532291 Chr.2: 202766452-202766452 on Build GRCh38
rs993568382 Chr.2: 202766586-202766586 on Build GRCh38
rs971931194 Chr.2: 202766195-202766195 on Build GRCh38
rs951972715 Chr.2: 202766202-202766202 on Build GRCh38
rs72930798 Chr.2: 202766250-202766250 on Build GRCh38
rs576487985 Chr.2: 202766398-202766398 on Build GRCh38
rs1049503348 Chr.2: 202766254-202766254 on Build GRCh38
rs1034311772 Chr.2: 202766474-202766474 on Build GRCh38
rs946861622 Chr.2: 202766345-202766345 on Build GRCh38
rs1007594599 Chr.2: 202766481-202766481 on Build GRCh38
rs1040774886 Chr.2: 202766580-202766580 on Build GRCh38
rs979400848 Chr.2: 202766197-202766197 on Build GRCh38
rs773749239 Chr.2: 202766216-202766216 on Build GRCh38
rs924974423 Chr.2: 202766233-202766233 on Build GRCh38
rs960234131 Chr.2: 202766173-202766173 on Build GRCh38
rs1043987894 Chr.2: 202766380-202766380 on Build GRCh38
rs943753584 Chr.2: 202766311-202766311 on Build GRCh38
rs966112108 Chr.2: 202766193-202766193 on Build GRCh38
rs967932529 Chr.2: 202766596-202766596 on Build GRCh38
rs138826390 Chr.2: 202766440-202766440 on Build GRCh38
rs915774163 Chr.2: 202766243-202766243 on Build GRCh38
rs146037490 Chr.2: 202766199-202766199 on Build GRCh38
rs994120307 Chr.2: 202766349-202766349 on Build GRCh38
rs981506663 Chr.2: 202766267-202766267 on Build GRCh38
rs914083139 Chr.2: 202766343-202766343 on Build GRCh38
rs934391068 Chr.2: 202766332-202766332 on Build GRCh38
rs960119717 Chr.2: 202766526-202766526 on Build GRCh38
rs75456597 Chr.2: 202766180-202766180 on Build GRCh38
rs1026374792 Chr.2: 202766593-202766593 on Build GRCh38
rs766822128 Chr.2: 202766333-202766333 on Build GRCh38
rs901951960 Chr.2: 202766584-202766584 on Build GRCh38
rs568652098 Chr.2: 202766303-202766303 on Build GRCh38

View Full Product Details