Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219003028-219003533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP65
Entrez Gene ID
255101
Gene Name
cilia and flagella associated protein 65
Gene Aliases
CCDC108
UniGene
Hs.147762
Gene Chromosome Location
Chr.2: 219002846-219043142 on Build GRCh38
Transcripts
NM_194302.3 XM_011510903.1 XM_011510904.2 XM_011510908.1 XM_011510909.1 XM_011510910.1 XM_011510911.1 XM_011510912.1 XM_011510914.1 XM_011510915.1 XM_011510916.1 XM_011510921.1 XM_017003752.1 XM_017003753.1 XM_017003754.1 XM_017003755.1 NR_046086.1

Gene Symbol
LOC100129175
Entrez Gene ID
100129175
Gene Name
uncharacterized LOC100129175
Gene Aliases
-
UniGene
Hs.147762
Gene Chromosome Location
Chr.2: 219002215-219015722 on Build GRCh38
Transcripts
NM_194302.3 XM_011510903.1 XM_011510904.2 XM_011510908.1 XM_011510909.1 XM_011510910.1 XM_011510911.1 XM_011510912.1 XM_011510914.1 XM_011510915.1 XM_011510916.1 XM_011510921.1 XM_017003752.1 XM_017003753.1 XM_017003754.1 XM_017003755.1 NR_046086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639498 c.5565C>A p.A1855A Chr.2: 219003263-219003263 on Build GRCh38
7359689 c.5628C>A p.S1876R Chr.2: 219003200-219003200 on Build GRCh38
4133463 c.5618T>G p.V1873G Chr.2: 219003210-219003210 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs150663004 Chr.2: 219003120-219003120 on Build GRCh38
rs956756509 Chr.2: 219003295-219003295 on Build GRCh38
rs1048165805 Chr.2: 219003365-219003365 on Build GRCh38
rs372088760 Chr.2: 219003065-219003065 on Build GRCh38
rs199732478 Chr.2: 219003068-219003068 on Build GRCh38
rs773812184 Chr.2: 219003213-219003213 on Build GRCh38
rs750322732 Chr.2: 219003124-219003124 on Build GRCh38
rs574146665 Chr.2: 219003453-219003453 on Build GRCh38
rs992295279 Chr.2: 219003237-219003237 on Build GRCh38
rs896745962 Chr.2: 219003460-219003460 on Build GRCh38
rs551755249 Chr.2: 219003097-219003097 on Build GRCh38
rs754719037 Chr.2: 219003497-219003497 on Build GRCh38
rs767302280 Chr.2: 219003349-219003349 on Build GRCh38
rs886805557 Chr.2: 219003155-219003155 on Build GRCh38
rs771704295 Chr.2: 219003204-219003204 on Build GRCh38
rs567327535 Chr.2: 219003189-219003189 on Build GRCh38
rs1003868351 Chr.2: 219003403-219003403 on Build GRCh38
rs912519295 Chr.2: 219003328-219003328 on Build GRCh38
rs776824623 Chr.2: 219003242-219003242 on Build GRCh38
rs555952352 Chr.2: 219003435-219003435 on Build GRCh38
rs775671978 Chr.2: 219003279-219003279 on Build GRCh38
rs778582401 Chr.2: 219003181-219003181 on Build GRCh38
rs890946142 Chr.2: 219003081-219003081 on Build GRCh38
rs369362490 Chr.2: 219003100-219003100 on Build GRCh38
rs765335761 Chr.2: 219003085-219003085 on Build GRCh38
rs544730281 Chr.2: 219003472-219003472 on Build GRCh38
rs1039574392 Chr.2: 219003338-219003338 on Build GRCh38
rs886770624 Chr.2: 219003371-219003371 on Build GRCh38
rs979428434 Chr.2: 219003281-219003281 on Build GRCh38
rs925264408 Chr.2: 219003289-219003289 on Build GRCh38
rs934641819 Chr.2: 219003260-219003260 on Build GRCh38
rs763083974 Chr.2: 219003307-219003307 on Build GRCh38
rs902086253 Chr.2: 219003161-219003161 on Build GRCh38
rs1041352803 Chr.2: 219003302-219003302 on Build GRCh38
rs531743353 Chr.2: 219003288-219003288 on Build GRCh38
rs1023636033 Chr.2: 219003245-219003245 on Build GRCh38
rs1005526536 Chr.2: 219003191-219003191 on Build GRCh38
rs920993065 Chr.2: 219003339-219003339 on Build GRCh38
rs959900230 Chr.2: 219003121-219003121 on Build GRCh38
rs1006401738 Chr.2: 219003348-219003348 on Build GRCh38
rs747719089 Chr.2: 219003203-219003203 on Build GRCh38
rs755976651 Chr.2: 219003125-219003125 on Build GRCh38
rs545869238 Chr.2: 219003153-219003153 on Build GRCh38
rs374766845 Chr.2: 219003221-219003221 on Build GRCh38
rs11893183 Chr.2: 219003255-219003255 on Build GRCh38
rs897491888 Chr.2: 219003369-219003369 on Build GRCh38
rs765382990 Chr.2: 219003265-219003265 on Build GRCh38
rs932826847 Chr.2: 219003323-219003323 on Build GRCh38
rs899583835 Chr.2: 219003103-219003103 on Build GRCh38
rs61739963 Chr.2: 219003147-219003147 on Build GRCh38
rs549456927 Chr.2: 219003209-219003209 on Build GRCh38
rs916020467 Chr.2: 219003208-219003208 on Build GRCh38
rs537992850 Chr.2: 219003194-219003194 on Build GRCh38
rs993117757 Chr.2: 219003407-219003407 on Build GRCh38
rs534210151 Chr.2: 219003389-219003389 on Build GRCh38
rs866500654 Chr.2: 219003362-219003362 on Build GRCh38
rs988056953 Chr.2: 219003312-219003312 on Build GRCh38
rs766624456 Chr.2: 219003059-219003059 on Build GRCh38
rs549196797 Chr.2: 219003158-219003158 on Build GRCh38
rs73993385 Chr.2: 219003350-219003350 on Build GRCh38
rs754612567 Chr.2: 219003175-219003175 on Build GRCh38
rs1015168010 Chr.2: 219003197-219003197 on Build GRCh38
rs114331223 Chr.2: 219003332-219003332 on Build GRCh38
rs757197711 Chr.2: 219003111-219003111 on Build GRCh38
rs771351859 Chr.2: 219003229-219003229 on Build GRCh38

View Full Product Details