Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219003869-219004378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP65
Entrez Gene ID
255101
Gene Name
cilia and flagella associated protein 65
Gene Aliases
CCDC108
UniGene
Hs.147762
Gene Chromosome Location
Chr.2: 219002846-219043142 on Build GRCh38
Transcripts
NM_194302.3 XM_011510903.1 XM_011510904.2 XM_011510908.1 XM_011510909.1 XM_011510910.1 XM_011510911.1 XM_011510912.1 XM_011510914.1 XM_011510915.1 XM_011510916.1 XM_011510921.1 XM_017003752.1 XM_017003753.1 XM_017003754.1 XM_017003755.1 NR_046086.1

Gene Symbol
LOC100129175
Entrez Gene ID
100129175
Gene Name
uncharacterized LOC100129175
Gene Aliases
-
UniGene
Hs.147762
Gene Chromosome Location
Chr.2: 219002215-219015722 on Build GRCh38
Transcripts
NM_194302.3 XM_011510903.1 XM_011510904.2 XM_011510908.1 XM_011510909.1 XM_011510910.1 XM_011510911.1 XM_011510912.1 XM_011510914.1 XM_011510915.1 XM_011510916.1 XM_011510921.1 XM_017003752.1 XM_017003753.1 XM_017003754.1 XM_017003755.1 NR_046086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5689198 c.5524C>T p.Q1842* Chr.2: 219003983-219003983 on Build GRCh38
234502 c.5368T>C p.L1790L Chr.2: 219004139-219004139 on Build GRCh38
1016537 c.5139G>A p.E1713E Chr.2: 219004368-219004368 on Build GRCh38
6576874 c.5327A>G p.E1776G Chr.2: 219004180-219004180 on Build GRCh38
6988597 c.5293G>A p.E1765K Chr.2: 219004214-219004214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941565550 Chr.2: 219004143-219004143 on Build GRCh38
rs564723222 Chr.2: 219004310-219004310 on Build GRCh38
rs756573002 Chr.2: 219004179-219004179 on Build GRCh38
rs772344237 Chr.2: 219004062-219004062 on Build GRCh38
rs892665423 Chr.2: 219004284-219004284 on Build GRCh38
rs781553221 Chr.2: 219004186-219004188 on Build GRCh38
rs956133675 Chr.2: 219004060-219004060 on Build GRCh38
rs762718817 Chr.2: 219004238-219004238 on Build GRCh38
rs766226357 Chr.2: 219003921-219003921 on Build GRCh38
rs766938631 Chr.2: 219004270-219004270 on Build GRCh38
rs777913055 Chr.2: 219003990-219003992 on Build GRCh38
rs1056744252 Chr.2: 219003898-219003898 on Build GRCh38
rs761866557 Chr.2: 219004070-219004070 on Build GRCh38
rs531605013 Chr.2: 219004288-219004288 on Build GRCh38
rs555774149 Chr.2: 219004071-219004071 on Build GRCh38
rs1056787212 Chr.2: 219004226-219004226 on Build GRCh38
rs747585408 Chr.2: 219003969-219003969 on Build GRCh38
rs748380747 Chr.2: 219004205-219004205 on Build GRCh38
rs746340865 Chr.2: 219004045-219004045 on Build GRCh38
rs780316098 Chr.2: 219004140-219004142 on Build GRCh38
rs368952935 Chr.2: 219004265-219004265 on Build GRCh38
rs779288534 Chr.2: 219004286-219004286 on Build GRCh38
rs775604190 Chr.2: 219004319-219004319 on Build GRCh38
rs148044633 Chr.2: 219004038-219004038 on Build GRCh38
rs866590111 Chr.2: 219003968-219003968 on Build GRCh38
rs775551113 Chr.2: 219004236-219004236 on Build GRCh38
rs756774974 Chr.2: 219004037-219004037 on Build GRCh38
rs770134961 Chr.2: 219003987-219003987 on Build GRCh38
rs779045715 Chr.2: 219004190-219004190 on Build GRCh38
rs571029384 Chr.2: 219003942-219003942 on Build GRCh38
rs749978288 Chr.2: 219004155-219004157 on Build GRCh38
rs774253176 Chr.2: 219004007-219004007 on Build GRCh38
rs750424298 Chr.2: 219003914-219003914 on Build GRCh38
rs148521256 Chr.2: 219004107-219004107 on Build GRCh38
rs62193333 Chr.2: 219004073-219004073 on Build GRCh38
rs763916400 Chr.2: 219004246-219004246 on Build GRCh38
rs61738865 Chr.2: 219003972-219003972 on Build GRCh38
rs910043256 Chr.2: 219003907-219003907 on Build GRCh38
rs79858834 Chr.2: 219004269-219004269 on Build GRCh38
rs111416503 Chr.2: 219004097-219004097 on Build GRCh38
rs780401221 Chr.2: 219004183-219004183 on Build GRCh38
rs916896885 Chr.2: 219004215-219004215 on Build GRCh38
rs768801955 Chr.2: 219004006-219004006 on Build GRCh38
rs891610321 Chr.2: 219004299-219004299 on Build GRCh38
rs565707514 Chr.2: 219004352-219004352 on Build GRCh38
rs566349288 Chr.2: 219004332-219004332 on Build GRCh38
rs547083948 Chr.2: 219004348-219004348 on Build GRCh38
rs146427311 Chr.2: 219004094-219004094 on Build GRCh38
rs773746924 Chr.2: 219004350-219004351 on Build GRCh38
rs370228808 Chr.2: 219003935-219003935 on Build GRCh38
rs774247985 Chr.2: 219004015-219004015 on Build GRCh38
rs757149562 Chr.2: 219004278-219004278 on Build GRCh38
rs772848133 Chr.2: 219004351-219004351 on Build GRCh38
rs147887290 Chr.2: 219004030-219004030 on Build GRCh38
rs113091985 Chr.2: 219004139-219004139 on Build GRCh38
rs201649638 Chr.2: 219004064-219004064 on Build GRCh38
rs758593810 Chr.2: 219004125-219004127 on Build GRCh38
rs543228128 Chr.2: 219004308-219004308 on Build GRCh38
rs748528828 Chr.2: 219004194-219004195 on Build GRCh38
rs535436534 Chr.2: 219004022-219004022 on Build GRCh38
rs992566993 Chr.2: 219004213-219004213 on Build GRCh38
rs777103237 Chr.2: 219003967-219003967 on Build GRCh38
rs771127455 Chr.2: 219004318-219004318 on Build GRCh38
rs370515327 Chr.2: 219004218-219004218 on Build GRCh38
rs554256421 Chr.2: 219004184-219004184 on Build GRCh38
rs74775944 Chr.2: 219004321-219004321 on Build GRCh38
rs758087222 Chr.2: 219004027-219004027 on Build GRCh38
rs753521414 Chr.2: 219003926-219003926 on Build GRCh38
rs750949568 Chr.2: 219004145-219004145 on Build GRCh38
rs538080749 Chr.2: 219004089-219004089 on Build GRCh38
rs73993388 Chr.2: 219003901-219003901 on Build GRCh38
rs1045345491 Chr.2: 219003925-219003925 on Build GRCh38
rs1009635435 Chr.2: 219004285-219004285 on Build GRCh38
rs374958890 Chr.2: 219004255-219004255 on Build GRCh38
rs768774161 Chr.2: 219004174-219004176 on Build GRCh38
rs761477114 Chr.2: 219004261-219004261 on Build GRCh38
rs780727838 Chr.2: 219004130-219004130 on Build GRCh38
rs763139603 Chr.2: 219004004-219004004 on Build GRCh38
rs750511867 Chr.2: 219004104-219004106 on Build GRCh38
rs146675518 Chr.2: 219004221-219004221 on Build GRCh38
rs147008412 Chr.2: 219004280-219004280 on Build GRCh38
rs758034561 Chr.2: 219003965-219003965 on Build GRCh38
rs73089095 Chr.2: 219004092-219004092 on Build GRCh38
rs146656596 Chr.2: 219004035-219004035 on Build GRCh38
rs746287779 Chr.2: 219003986-219003986 on Build GRCh38
rs908265916 Chr.2: 219004182-219004182 on Build GRCh38
rs766286598 Chr.2: 219004013-219004013 on Build GRCh38
rs199507646 Chr.2: 219003915-219003915 on Build GRCh38
rs770191945 Chr.2: 219004048-219004048 on Build GRCh38
rs144952439 Chr.2: 219004256-219004256 on Build GRCh38
rs774621115 Chr.2: 219004068-219004068 on Build GRCh38
rs747147500 Chr.2: 219004312-219004312 on Build GRCh38
rs139026486 Chr.2: 219004235-219004235 on Build GRCh38
rs139829270 Chr.2: 219004065-219004065 on Build GRCh38
rs372658951 Chr.2: 219004054-219004054 on Build GRCh38
rs772070772 Chr.2: 219004211-219004211 on Build GRCh38
rs770323664 Chr.2: 219004228-219004228 on Build GRCh38
rs759444384 Chr.2: 219004016-219004016 on Build GRCh38
rs567905966 Chr.2: 219004072-219004072 on Build GRCh38
rs367854251 Chr.2: 219004342-219004342 on Build GRCh38
rs767042867 Chr.2: 219004264-219004264 on Build GRCh38
rs760676584 Chr.2: 219004008-219004008 on Build GRCh38
rs868016521 Chr.2: 219004208-219004208 on Build GRCh38
rs754339706 Chr.2: 219004274-219004274 on Build GRCh38
rs150073241 Chr.2: 219003971-219003971 on Build GRCh38
rs759501252 Chr.2: 219004078-219004078 on Build GRCh38
rs774154389 Chr.2: 219004339-219004339 on Build GRCh38

View Full Product Details