Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219004270-219004762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP65
Entrez Gene ID
255101
Gene Name
cilia and flagella associated protein 65
Gene Aliases
CCDC108
UniGene
Hs.147762
Gene Chromosome Location
Chr.2: 219002846-219043142 on Build GRCh38
Transcripts
NM_194302.3 XM_011510903.1 XM_011510904.2 XM_011510908.1 XM_011510909.1 XM_011510910.1 XM_011510911.1 XM_011510912.1 XM_011510914.1 XM_011510915.1 XM_011510916.1 XM_011510921.1 XM_017003752.1 XM_017003753.1 XM_017003754.1 XM_017003755.1 NR_046086.1

Gene Symbol
LOC100129175
Entrez Gene ID
100129175
Gene Name
uncharacterized LOC100129175
Gene Aliases
-
UniGene
Hs.147762
Gene Chromosome Location
Chr.2: 219002215-219015722 on Build GRCh38
Transcripts
NM_194302.3 XM_011510903.1 XM_011510904.2 XM_011510908.1 XM_011510909.1 XM_011510910.1 XM_011510911.1 XM_011510912.1 XM_011510914.1 XM_011510915.1 XM_011510916.1 XM_011510921.1 XM_017003752.1 XM_017003753.1 XM_017003754.1 XM_017003755.1 NR_046086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7330486 c.5232G>T p.K1744N Chr.2: 219004275-219004275 on Build GRCh38
3972025 c.5237C>T p.P1746L Chr.2: 219004270-219004270 on Build GRCh38
5058677 c.5213G>A p.W1738* Chr.2: 219004294-219004294 on Build GRCh38
1016537 c.5139G>A p.E1713E Chr.2: 219004368-219004368 on Build GRCh38
3044240 c.5106G>T p.E1702D Chr.2: 219004401-219004401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028975122 Chr.2: 219004467-219004467 on Build GRCh38
rs144291969 Chr.2: 219004411-219004411 on Build GRCh38
rs773746924 Chr.2: 219004350-219004351 on Build GRCh38
rs564723222 Chr.2: 219004310-219004310 on Build GRCh38
rs992547345 Chr.2: 219004575-219004575 on Build GRCh38
rs771824935 Chr.2: 219004447-219004447 on Build GRCh38
rs556826450 Chr.2: 219004452-219004452 on Build GRCh38
rs772848133 Chr.2: 219004351-219004351 on Build GRCh38
rs779857781 Chr.2: 219004691-219004692 on Build GRCh38
rs762434075 Chr.2: 219004666-219004666 on Build GRCh38
rs543228128 Chr.2: 219004308-219004308 on Build GRCh38
rs771127455 Chr.2: 219004318-219004318 on Build GRCh38
rs926836678 Chr.2: 219004434-219004434 on Build GRCh38
rs542677205 Chr.2: 219004649-219004649 on Build GRCh38
rs74775944 Chr.2: 219004321-219004321 on Build GRCh38
rs1041115668 Chr.2: 219004559-219004559 on Build GRCh38
rs948358337 Chr.2: 219004393-219004393 on Build GRCh38
rs559443098 Chr.2: 219004450-219004450 on Build GRCh38
rs553984858 Chr.2: 219004552-219004552 on Build GRCh38
rs568591993 Chr.2: 219004720-219004720 on Build GRCh38
rs750457337 Chr.2: 219004507-219004507 on Build GRCh38
rs765855481 Chr.2: 219004360-219004360 on Build GRCh38
rs776142812 Chr.2: 219004460-219004460 on Build GRCh38
rs945560569 Chr.2: 219004551-219004551 on Build GRCh38
rs749213358 Chr.2: 219004410-219004410 on Build GRCh38
rs770461580 Chr.2: 219004454-219004454 on Build GRCh38
rs1028338408 Chr.2: 219004694-219004694 on Build GRCh38
rs538526809 Chr.2: 219004404-219004404 on Build GRCh38
rs138227108 Chr.2: 219004354-219004354 on Build GRCh38
rs757459317 Chr.2: 219004397-219004397 on Build GRCh38
rs749823774 Chr.2: 219004354-219004354 on Build GRCh38
rs775604190 Chr.2: 219004319-219004319 on Build GRCh38
rs146131399 Chr.2: 219004380-219004380 on Build GRCh38
rs916991489 Chr.2: 219004644-219004644 on Build GRCh38
rs777029692 Chr.2: 219004415-219004415 on Build GRCh38
rs781135930 Chr.2: 219004505-219004505 on Build GRCh38
rs150010715 Chr.2: 219004385-219004385 on Build GRCh38
rs759976714 Chr.2: 219004468-219004468 on Build GRCh38
rs567875851 Chr.2: 219004405-219004405 on Build GRCh38
rs77580905 Chr.2: 219004549-219004549 on Build GRCh38
rs751851632 Chr.2: 219004495-219004495 on Build GRCh38
rs199601952 Chr.2: 219004379-219004379 on Build GRCh38
rs765834442 Chr.2: 219004480-219004480 on Build GRCh38
rs757512359 Chr.2: 219004504-219004504 on Build GRCh38
rs139532198 Chr.2: 219004392-219004392 on Build GRCh38
rs778204349 Chr.2: 219004389-219004389 on Build GRCh38
rs529664789 Chr.2: 219004353-219004353 on Build GRCh38
rs748032938 Chr.2: 219004432-219004432 on Build GRCh38
rs764527945 Chr.2: 219004492-219004492 on Build GRCh38
rs760273581 Chr.2: 219004453-219004453 on Build GRCh38
rs753143685 Chr.2: 219004481-219004481 on Build GRCh38
rs753052024 Chr.2: 219004384-219004384 on Build GRCh38
rs146392908 Chr.2: 219004382-219004382 on Build GRCh38
rs747147500 Chr.2: 219004312-219004312 on Build GRCh38
rs901259890 Chr.2: 219004617-219004617 on Build GRCh38
rs778585830 Chr.2: 219004428-219004428 on Build GRCh38
rs367854251 Chr.2: 219004342-219004342 on Build GRCh38
rs199851338 Chr.2: 219004486-219004486 on Build GRCh38
rs774154389 Chr.2: 219004339-219004339 on Build GRCh38
rs1054468141 Chr.2: 219004532-219004532 on Build GRCh38
rs891610321 Chr.2: 219004299-219004299 on Build GRCh38
rs565707514 Chr.2: 219004352-219004352 on Build GRCh38
rs572457400 Chr.2: 219004614-219004614 on Build GRCh38
rs566349288 Chr.2: 219004332-219004332 on Build GRCh38
rs547083948 Chr.2: 219004348-219004348 on Build GRCh38

View Full Product Details