Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232014694-232015204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3L2
Entrez Gene ID
129563
Gene Name
DIS3 like 3'-5' exoribonuclease 2
Gene Aliases
FAM6A, PRLMNS, hDIS3L2
UniGene
Hs.732236
Gene Chromosome Location
Chr.2: 231961583-232343968 on Build GRCh38
Transcripts
NM_001257281.1 NM_001257282.1 NM_152383.4 NR_046476.1 NR_046477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3771815 c.52+1G>A p.? Chr.2: 232014980-232014980 on Build GRCh38
3771814 c.52+1G>A p.? Chr.2: 232014980-232014980 on Build GRCh38
40869 c.52+1G>A p.? Chr.2: 232014980-232014980 on Build GRCh38
3771813 c.52+1G>A p.? Chr.2: 232014980-232014980 on Build GRCh38
1203652 c.32G>A p.R11Q Chr.2: 232014959-232014959 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1052729966 Chr.2: 232015116-232015116 on Build GRCh38
rs574055101 Chr.2: 232014955-232014955 on Build GRCh38
rs79766624 Chr.2: 232015089-232015089 on Build GRCh38
rs146573032 Chr.2: 232014889-232014889 on Build GRCh38
rs749719015 Chr.2: 232014971-232014971 on Build GRCh38
rs922102239 Chr.2: 232014995-232014995 on Build GRCh38
rs564154963 Chr.2: 232015021-232015022 on Build GRCh38
rs1016971486 Chr.2: 232014791-232014791 on Build GRCh38
rs78207230 Chr.2: 232015090-232015090 on Build GRCh38
rs770359206 Chr.2: 232014963-232014963 on Build GRCh38
rs902790701 Chr.2: 232014810-232014810 on Build GRCh38
rs974812898 Chr.2: 232014973-232014973 on Build GRCh38
rs565446201 Chr.2: 232015166-232015166 on Build GRCh38
rs1045972795 Chr.2: 232015079-232015079 on Build GRCh38
rs554832054 Chr.2: 232014964-232014964 on Build GRCh38
rs747590766 Chr.2: 232015113-232015113 on Build GRCh38
rs751777329 Chr.2: 232014958-232014958 on Build GRCh38
rs530022857 Chr.2: 232014775-232014775 on Build GRCh38
rs934304024 Chr.2: 232014844-232014844 on Build GRCh38
rs376955979 Chr.2: 232014923-232014923 on Build GRCh38
rs766169815 Chr.2: 232014922-232014922 on Build GRCh38
rs981695324 Chr.2: 232014800-232014800 on Build GRCh38
rs773905464 Chr.2: 232014982-232014982 on Build GRCh38
rs723044 Chr.2: 232014961-232014961 on Build GRCh38
rs944832487 Chr.2: 232014790-232014790 on Build GRCh38
rs949752211 Chr.2: 232015153-232015153 on Build GRCh38
rs1022761528 Chr.2: 232014861-232014861 on Build GRCh38
rs370302381 Chr.2: 232014935-232014935 on Build GRCh38
rs540447709 Chr.2: 232015080-232015080 on Build GRCh38
rs769111524 Chr.2: 232014974-232014974 on Build GRCh38
rs563786015 Chr.2: 232014792-232014792 on Build GRCh38
rs114491317 Chr.2: 232014850-232014850 on Build GRCh38
rs78129899 Chr.2: 232015026-232015026 on Build GRCh38
rs750961522 Chr.2: 232014876-232014876 on Build GRCh38
rs747785349 Chr.2: 232014999-232014999 on Build GRCh38
rs886055769 Chr.2: 232014870-232014874 on Build GRCh38
rs979655805 Chr.2: 232015074-232015074 on Build GRCh38
rs566709027 Chr.2: 232014802-232014802 on Build GRCh38
rs922103500 Chr.2: 232014799-232014799 on Build GRCh38
rs760551384 Chr.2: 232014908-232014908 on Build GRCh38
rs937323761 Chr.2: 232015099-232015099 on Build GRCh38
rs111359519 Chr.2: 232015138-232015138 on Build GRCh38
rs754758868 Chr.2: 232014928-232014928 on Build GRCh38
rs542604774 Chr.2: 232014942-232014942 on Build GRCh38
rs750351609 Chr.2: 232014898-232014898 on Build GRCh38
rs983439393 Chr.2: 232014755-232014755 on Build GRCh38
rs924182328 Chr.2: 232015097-232015097 on Build GRCh38
rs895564243 Chr.2: 232014855-232014855 on Build GRCh38
rs905834667 Chr.2: 232014728-232014728 on Build GRCh38
rs868547326 Chr.2: 232015072-232015072 on Build GRCh38
rs533740964 Chr.2: 232014803-232014803 on Build GRCh38
rs144258056 Chr.2: 232014860-232014860 on Build GRCh38
rs886055768 Chr.2: 232014861-232014861 on Build GRCh38
rs60514104 Chr.2: 232014877-232014877 on Build GRCh38
rs548460800 Chr.2: 232014801-232014801 on Build GRCh38
rs987364927 Chr.2: 232014858-232014858 on Build GRCh38
rs757314939 Chr.2: 232014960-232014960 on Build GRCh38
rs770033625 Chr.2: 232014851-232014851 on Build GRCh38
rs898595971 Chr.2: 232015126-232015126 on Build GRCh38
rs368127511 Chr.2: 232015004-232015004 on Build GRCh38
rs767015111 Chr.2: 232014896-232014896 on Build GRCh38
rs916925730 Chr.2: 232015133-232015133 on Build GRCh38

View Full Product Details