Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232136330-232136842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3L2
Entrez Gene ID
129563
Gene Name
DIS3 like 3'-5' exoribonuclease 2
Gene Aliases
FAM6A, PRLMNS, hDIS3L2
UniGene
Hs.732236
Gene Chromosome Location
Chr.2: 231961583-232343968 on Build GRCh38
Transcripts
NM_001257281.1 NM_152383.4 NR_046476.1 NR_046477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4963932 c.714C>G p.I238M Chr.2: 232136483-232136483 on Build GRCh38
6644336 c.762-7delT p.? Chr.2: 232136465-232136465 on Build GRCh38
3579050 c.733C>T p.R245* Chr.2: 232136502-232136502 on Build GRCh38
4963934 c.773C>G p.S258C Chr.2: 232136483-232136483 on Build GRCh38
1017782 c.745G>A p.G249S Chr.2: 232136514-232136514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939770688 Chr.2: 232136645-232136645 on Build GRCh38
rs754258699 Chr.2: 232136513-232136513 on Build GRCh38
rs570981537 Chr.2: 232136649-232136649 on Build GRCh38
rs765716069 Chr.2: 232136744-232136744 on Build GRCh38
rs749133430 Chr.2: 232136696-232136696 on Build GRCh38
rs770321098 Chr.2: 232136532-232136532 on Build GRCh38
rs202042951 Chr.2: 232136564-232136564 on Build GRCh38
rs16828636 Chr.2: 232136409-232136409 on Build GRCh38
rs376763206 Chr.2: 232136618-232136618 on Build GRCh38
rs781163337 Chr.2: 232136589-232136589 on Build GRCh38
rs922412610 Chr.2: 232136673-232136673 on Build GRCh38
rs768529345 Chr.2: 232136707-232136707 on Build GRCh38
rs568422819 Chr.2: 232136767-232136767 on Build GRCh38
rs745823386 Chr.2: 232136666-232136666 on Build GRCh38
rs753802899 Chr.2: 232136767-232136768 on Build GRCh38
rs961972899 Chr.2: 232136415-232136415 on Build GRCh38
rs200011409 Chr.2: 232136447-232136447 on Build GRCh38
rs780127954 Chr.2: 232136688-232136688 on Build GRCh38
rs375555351 Chr.2: 232136747-232136747 on Build GRCh38
rs560705101 Chr.2: 232136592-232136592 on Build GRCh38
rs747141045 Chr.2: 232136714-232136714 on Build GRCh38
rs758109640 Chr.2: 232136522-232136522 on Build GRCh38
rs759890520 Chr.2: 232136622-232136622 on Build GRCh38
rs755391038 Chr.2: 232136515-232136515 on Build GRCh38
rs985913473 Chr.2: 232136514-232136514 on Build GRCh38
rs533388211 Chr.2: 232136799-232136799 on Build GRCh38
rs775360586 Chr.2: 232136547-232136547 on Build GRCh38
rs747746718 Chr.2: 232136444-232136444 on Build GRCh38
rs766947723 Chr.2: 232136511-232136511 on Build GRCh38
rs924879950 Chr.2: 232136607-232136607 on Build GRCh38
rs780510878 Chr.2: 232136653-232136653 on Build GRCh38
rs775318567 Chr.2: 232136465-232136465 on Build GRCh38
rs746514811 Chr.2: 232136453-232136453 on Build GRCh38
rs760475750 Chr.2: 232136563-232136564 on Build GRCh38
rs772470899 Chr.2: 232136565-232136565 on Build GRCh38
rs776638867 Chr.2: 232136727-232136727 on Build GRCh38
rs199648534 Chr.2: 232136502-232136502 on Build GRCh38
rs768117442 Chr.2: 232136644-232136644 on Build GRCh38
rs149157216 Chr.2: 232136679-232136679 on Build GRCh38
rs777194934 Chr.2: 232136450-232136450 on Build GRCh38
rs777394342 Chr.2: 232136527-232136527 on Build GRCh38
rs375649949 Chr.2: 232136546-232136546 on Build GRCh38
rs200720002 Chr.2: 232136768-232136768 on Build GRCh38
rs774656238 Chr.2: 232136481-232136481 on Build GRCh38
rs763950696 Chr.2: 232136565-232136565 on Build GRCh38
rs781224944 Chr.2: 232136536-232136536 on Build GRCh38
rs770915716 Chr.2: 232136455-232136455 on Build GRCh38
rs759644979 Chr.2: 232136729-232136729 on Build GRCh38
rs759420051 Chr.2: 232136462-232136462 on Build GRCh38
rs752349220 Chr.2: 232136519-232136519 on Build GRCh38
rs773590340 Chr.2: 232136712-232136712 on Build GRCh38
rs762069645 Chr.2: 232136487-232136487 on Build GRCh38
rs371095624 Chr.2: 232136498-232136498 on Build GRCh38
rs1033352725 Chr.2: 232136555-232136555 on Build GRCh38
rs755267179 Chr.2: 232136424-232136424 on Build GRCh38
rs187563594 Chr.2: 232136647-232136647 on Build GRCh38
rs1010297427 Chr.2: 232136792-232136792 on Build GRCh38
rs753115829 Chr.2: 232136640-232136640 on Build GRCh38
rs145001670 Chr.2: 232136441-232136441 on Build GRCh38
rs182004457 Chr.2: 232136643-232136643 on Build GRCh38
rs367638559 Chr.2: 232136552-232136552 on Build GRCh38
rs769440116 Chr.2: 232136474-232136474 on Build GRCh38
rs888550125 Chr.2: 232136813-232136813 on Build GRCh38
rs563841810 Chr.2: 232136372-232136372 on Build GRCh38
rs766319323 Chr.2: 232136605-232136605 on Build GRCh38
rs765766984 Chr.2: 232136627-232136627 on Build GRCh38
rs749634733 Chr.2: 232136669-232136669 on Build GRCh38
rs771043528 Chr.2: 232136726-232136726 on Build GRCh38
rs779084603 Chr.2: 232136516-232136516 on Build GRCh38
rs771893747 Chr.2: 232136439-232136439 on Build GRCh38
rs1005992864 Chr.2: 232136397-232136397 on Build GRCh38
rs952754100 Chr.2: 232136380-232136380 on Build GRCh38
rs781622806 Chr.2: 232136507-232136507 on Build GRCh38
rs933918889 Chr.2: 232136678-232136678 on Build GRCh38
rs746573965 Chr.2: 232136529-232136529 on Build GRCh38

View Full Product Details