Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237877121-237877616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAMP1
Entrez Gene ID
10267
Gene Name
receptor activity modifying protein 1
Gene Aliases
-
UniGene
Hs.471783
Gene Chromosome Location
Chr.2: 237858714-237912117 on Build GRCh38
Transcripts
NM_001308353.1 NM_005855.3 XM_017003152.1 XM_017003153.1 XM_017003154.1 XM_017003155.1 XM_017003156.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5461619 c.110G>A p.R37Q Chr.2: 237877281-237877281 on Build GRCh38
7181394 c.153C>T p.V51V Chr.2: 237877324-237877324 on Build GRCh38
5862129 c.127C>A p.Q43K Chr.2: 237877298-237877298 on Build GRCh38
7426305 c.128A>T p.Q43L Chr.2: 237877299-237877299 on Build GRCh38
5728141 c.74C>T p.T25I Chr.2: 237877245-237877245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530531134 Chr.2: 237877448-237877448 on Build GRCh38
rs199557016 Chr.2: 237877192-237877192 on Build GRCh38
rs138140919 Chr.2: 237877189-237877189 on Build GRCh38
rs768699419 Chr.2: 237877360-237877360 on Build GRCh38
rs908742471 Chr.2: 237877496-237877496 on Build GRCh38
rs940037704 Chr.2: 237877209-237877209 on Build GRCh38
rs749409676 Chr.2: 237877349-237877349 on Build GRCh38
rs111827579 Chr.2: 237877151-237877151 on Build GRCh38
rs199917306 Chr.2: 237877211-237877211 on Build GRCh38
rs147407024 Chr.2: 237877339-237877339 on Build GRCh38
rs771395188 Chr.2: 237877277-237877277 on Build GRCh38
rs772822050 Chr.2: 237877392-237877392 on Build GRCh38
rs940194840 Chr.2: 237877516-237877516 on Build GRCh38
rs372494526 Chr.2: 237877322-237877322 on Build GRCh38
rs757218127 Chr.2: 237877231-237877231 on Build GRCh38
rs757922636 Chr.2: 237877411-237877411 on Build GRCh38
rs186592424 Chr.2: 237877210-237877210 on Build GRCh38
rs754712934 Chr.2: 237877332-237877332 on Build GRCh38
rs12477382 Chr.2: 237877561-237877561 on Build GRCh38
rs201860745 Chr.2: 237877235-237877235 on Build GRCh38
rs375505993 Chr.2: 237877194-237877194 on Build GRCh38
rs183713343 Chr.2: 237877492-237877492 on Build GRCh38
rs898940801 Chr.2: 237877524-237877524 on Build GRCh38
rs573157338 Chr.2: 237877154-237877154 on Build GRCh38
rs1049834416 Chr.2: 237877533-237877533 on Build GRCh38
rs772148962 Chr.2: 237877500-237877500 on Build GRCh38
rs200451024 Chr.2: 237877397-237877397 on Build GRCh38
rs750953979 Chr.2: 237877489-237877490 on Build GRCh38
rs748950129 Chr.2: 237877266-237877266 on Build GRCh38
rs191395915 Chr.2: 237877419-237877419 on Build GRCh38
rs371519125 Chr.2: 237877404-237877404 on Build GRCh38
rs914336435 Chr.2: 237877475-237877475 on Build GRCh38
rs570291308 Chr.2: 237877173-237877173 on Build GRCh38
rs755710265 Chr.2: 237877251-237877251 on Build GRCh38
rs3214634 Chr.2: 237877497-237877497 on Build GRCh38
rs950228510 Chr.2: 237877401-237877401 on Build GRCh38
rs373098611 Chr.2: 237877190-237877190 on Build GRCh38
rs774612683 Chr.2: 237877370-237877370 on Build GRCh38
rs967051037 Chr.2: 237877145-237877145 on Build GRCh38
rs775366491 Chr.2: 237877283-237877283 on Build GRCh38
rs955896486 Chr.2: 237877470-237877470 on Build GRCh38
rs888987308 Chr.2: 237877229-237877229 on Build GRCh38
rs891196839 Chr.2: 237877537-237877537 on Build GRCh38
rs767305303 Chr.2: 237877438-237877438 on Build GRCh38
rs1008135554 Chr.2: 237877437-237877437 on Build GRCh38
rs781748441 Chr.2: 237877280-237877280 on Build GRCh38
rs1056697895 Chr.2: 237877495-237877495 on Build GRCh38
rs773649251 Chr.2: 237877180-237877180 on Build GRCh38
rs550249200 Chr.2: 237877163-237877163 on Build GRCh38
rs955237778 Chr.2: 237877195-237877195 on Build GRCh38
rs137909772 Chr.2: 237877237-237877237 on Build GRCh38
rs770143286 Chr.2: 237877341-237877341 on Build GRCh38
rs372292082 Chr.2: 237877318-237877318 on Build GRCh38
rs774524755 Chr.2: 237877294-237877294 on Build GRCh38
rs549388071 Chr.2: 237877494-237877494 on Build GRCh38
rs772855534 Chr.2: 237877260-237877261 on Build GRCh38
rs747341610 Chr.2: 237877276-237877276 on Build GRCh38
rs1049785829 Chr.2: 237877566-237877566 on Build GRCh38
rs991278836 Chr.2: 237877376-237877376 on Build GRCh38
rs990341816 Chr.2: 237877474-237877474 on Build GRCh38
rs771902021 Chr.2: 237877390-237877390 on Build GRCh38
rs144179532 Chr.2: 237877267-237877267 on Build GRCh38
rs975364587 Chr.2: 237877187-237877187 on Build GRCh38
rs764828803 Chr.2: 237877399-237877399 on Build GRCh38
rs566952640 Chr.2: 237877550-237877550 on Build GRCh38
rs565977270 Chr.2: 237877499-237877499 on Build GRCh38
rs534877854 Chr.2: 237877549-237877549 on Build GRCh38
rs183312906 Chr.2: 237877153-237877153 on Build GRCh38
rs939581097 Chr.2: 237877328-237877328 on Build GRCh38
rs780204782 Chr.2: 237877345-237877345 on Build GRCh38
rs1031945918 Chr.2: 237877469-237877469 on Build GRCh38
rs757856061 Chr.2: 237877156-237877156 on Build GRCh38
rs530518055 Chr.2: 237877158-237877158 on Build GRCh38
rs377248643 Chr.2: 237877324-237877324 on Build GRCh38
rs369223632 Chr.2: 237877333-237877333 on Build GRCh38
rs542472424 Chr.2: 237877395-237877395 on Build GRCh38
rs753345064 Chr.2: 237877331-237877331 on Build GRCh38
rs1028323509 Chr.2: 237877226-237877226 on Build GRCh38
rs772634722 Chr.2: 237877177-237877177 on Build GRCh38
rs200746457 Chr.2: 237877268-237877268 on Build GRCh38
rs369928837 Chr.2: 237877155-237877155 on Build GRCh38
rs763176383 Chr.2: 237877291-237877291 on Build GRCh38
rs777181015 Chr.2: 237877165-237877165 on Build GRCh38
rs373719548 Chr.2: 237877338-237877338 on Build GRCh38
rs142335491 Chr.2: 237877281-237877281 on Build GRCh38
rs573852299 Chr.2: 237877372-237877372 on Build GRCh38
rs768826156 Chr.2: 237877292-237877292 on Build GRCh38
rs374749100 Chr.2: 237877260-237877260 on Build GRCh38
rs564963802 Chr.2: 237877420-237877420 on Build GRCh38
rs762621127 Chr.2: 237877410-237877411 on Build GRCh38
rs563852512 Chr.2: 237877491-237877491 on Build GRCh38
rs760459192 Chr.2: 237877325-237877325 on Build GRCh38
rs779807147 Chr.2: 237877174-237877174 on Build GRCh38
rs148658989 Chr.2: 237877336-237877336 on Build GRCh38
rs746391827 Chr.2: 237877340-237877340 on Build GRCh38
rs765207546 Chr.2: 237877206-237877206 on Build GRCh38
rs372540157 Chr.2: 237877193-237877193 on Build GRCh38
rs1044004510 Chr.2: 237877313-237877313 on Build GRCh38
rs761644796 Chr.2: 237877321-237877321 on Build GRCh38
rs760680585 Chr.2: 237877394-237877394 on Build GRCh38
rs1210881 Chr.2: 237877490-237877490 on Build GRCh38
rs559204344 Chr.2: 237877139-237877139 on Build GRCh38
rs1040196786 Chr.2: 237877560-237877560 on Build GRCh38

View Full Product Details