Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 71063558-71064106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB2
Entrez Gene ID
9406
Gene Name
zinc finger RANBP2-type containing 2
Gene Aliases
ZIS, ZIS1, ZIS2, ZNF265
UniGene
Hs.194718
Gene Chromosome Location
Chr.1: 71063291-71081289 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

Gene Symbol
ZRANB2-AS1
Entrez Gene ID
100132618
Gene Name
ZRANB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.720513
Gene Chromosome Location
Chr.1: 71046506-71067184 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

SNP Detail

SNP ID Chromosome Location
rs545863253 Chr.1: 71063776-71063776 on Build GRCh38
rs1017367707 Chr.1: 71063752-71063752 on Build GRCh38
rs1052179291 Chr.1: 71063889-71063889 on Build GRCh38
rs751459093 Chr.1: 71063860-71063860 on Build GRCh38
rs1045313970 Chr.1: 71063934-71063934 on Build GRCh38
rs764419425 Chr.1: 71063853-71063854 on Build GRCh38
rs942349360 Chr.1: 71063607-71063607 on Build GRCh38
rs757324276 Chr.1: 71063879-71063879 on Build GRCh38
rs1056009578 Chr.1: 71063644-71063644 on Build GRCh38
rs1013390358 Chr.1: 71063904-71063904 on Build GRCh38
rs531436718 Chr.1: 71063994-71063994 on Build GRCh38
rs1051284614 Chr.1: 71063597-71063597 on Build GRCh38
rs905086297 Chr.1: 71063944-71063944 on Build GRCh38
rs201525423 Chr.1: 71063710-71063711 on Build GRCh38
rs749978864 Chr.1: 71063913-71063913 on Build GRCh38
rs867288353 Chr.1: 71063695-71063695 on Build GRCh38
rs116251378 Chr.1: 71063851-71063851 on Build GRCh38
rs997087350 Chr.1: 71063779-71063779 on Build GRCh38
rs189292118 Chr.1: 71063883-71063883 on Build GRCh38
rs903580893 Chr.1: 71063630-71063630 on Build GRCh38
rs184399198 Chr.1: 71063638-71063638 on Build GRCh38
rs550907236 Chr.1: 71063989-71063989 on Build GRCh38
rs1025649152 Chr.1: 71064069-71064069 on Build GRCh38
rs981959759 Chr.1: 71063625-71063625 on Build GRCh38
rs896238444 Chr.1: 71063902-71063902 on Build GRCh38
rs1013463860 Chr.1: 71063732-71063732 on Build GRCh38
rs890038431 Chr.1: 71063605-71063605 on Build GRCh38
rs559215986 Chr.1: 71063788-71063788 on Build GRCh38
rs928712650 Chr.1: 71063876-71063876 on Build GRCh38
rs561668480 Chr.1: 71063922-71063922 on Build GRCh38
rs886800557 Chr.1: 71063747-71063747 on Build GRCh38
rs764057741 Chr.1: 71063834-71063834 on Build GRCh38
rs899642133 Chr.1: 71063777-71063777 on Build GRCh38
rs1023517518 Chr.1: 71063740-71063740 on Build GRCh38
rs142073360 Chr.1: 71064081-71064081 on Build GRCh38
rs1016215750 Chr.1: 71063968-71063968 on Build GRCh38
rs1044008544 Chr.1: 71063861-71063861 on Build GRCh38
rs1006537749 Chr.1: 71063963-71063963 on Build GRCh38
rs937522852 Chr.1: 71063642-71063642 on Build GRCh38
rs962771055 Chr.1: 71063763-71063763 on Build GRCh38
rs1040761753 Chr.1: 71064068-71064068 on Build GRCh38
rs1001514503 Chr.1: 71063750-71063750 on Build GRCh38
rs931548826 Chr.1: 71063915-71063915 on Build GRCh38
rs942966027 Chr.1: 71063852-71063852 on Build GRCh38
rs7411 Chr.1: 71063718-71063718 on Build GRCh38
rs969717740 Chr.1: 71063742-71063742 on Build GRCh38
rs994129897 Chr.1: 71064059-71064059 on Build GRCh38
rs1142189 Chr.1: 71063778-71063778 on Build GRCh38
rs962683264 Chr.1: 71064027-71064027 on Build GRCh38
rs1047331891 Chr.1: 71063916-71063916 on Build GRCh38
rs918711754 Chr.1: 71063805-71063805 on Build GRCh38
rs1043799096 Chr.1: 71063628-71063628 on Build GRCh38
rs957937383 Chr.1: 71063816-71063816 on Build GRCh38
rs913274999 Chr.1: 71063595-71063596 on Build GRCh38
rs572786515 Chr.1: 71063814-71063814 on Build GRCh38
rs986938510 Chr.1: 71063843-71063843 on Build GRCh38
rs1007919861 Chr.1: 71063760-71063760 on Build GRCh38
rs886746588 Chr.1: 71063930-71063930 on Build GRCh38
rs1046714719 Chr.1: 71063739-71063739 on Build GRCh38
rs530781911 Chr.1: 71063961-71063961 on Build GRCh38

View Full Product Details