Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 71064398-71064903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB2
Entrez Gene ID
9406
Gene Name
zinc finger RANBP2-type containing 2
Gene Aliases
ZIS, ZIS1, ZIS2, ZNF265
UniGene
Hs.194718
Gene Chromosome Location
Chr.1: 71063291-71081289 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

Gene Symbol
ZRANB2-AS1
Entrez Gene ID
100132618
Gene Name
ZRANB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.720513
Gene Chromosome Location
Chr.1: 71046506-71067184 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

SNP Detail

SNP ID Chromosome Location
rs181287023 Chr.1: 71064540-71064540 on Build GRCh38
rs575331901 Chr.1: 71064680-71064680 on Build GRCh38
rs988379292 Chr.1: 71064791-71064791 on Build GRCh38
rs926394487 Chr.1: 71064517-71064517 on Build GRCh38
rs146951263 Chr.1: 71064866-71064866 on Build GRCh38
rs17090769 Chr.1: 71064451-71064451 on Build GRCh38
rs561630418 Chr.1: 71064671-71064671 on Build GRCh38
rs1051658095 Chr.1: 71064569-71064569 on Build GRCh38
rs932351497 Chr.1: 71064503-71064503 on Build GRCh38
rs141263007 Chr.1: 71064624-71064624 on Build GRCh38
rs1015701124 Chr.1: 71064699-71064699 on Build GRCh38
rs888524615 Chr.1: 71064635-71064635 on Build GRCh38
rs1042244323 Chr.1: 71064545-71064545 on Build GRCh38
rs949066191 Chr.1: 71064483-71064483 on Build GRCh38
rs908753401 Chr.1: 71064455-71064455 on Build GRCh38
rs920900596 Chr.1: 71064502-71064502 on Build GRCh38
rs956883625 Chr.1: 71064736-71064736 on Build GRCh38
rs546434261 Chr.1: 71064867-71064867 on Build GRCh38
rs1037552450 Chr.1: 71064534-71064534 on Build GRCh38
rs568130613 Chr.1: 71064459-71064459 on Build GRCh38
rs926335887 Chr.1: 71064855-71064855 on Build GRCh38
rs17090773 Chr.1: 71064631-71064631 on Build GRCh38
rs979629345 Chr.1: 71064441-71064441 on Build GRCh38
rs879467161 Chr.1: 71064804-71064804 on Build GRCh38
rs1032531160 Chr.1: 71064714-71064714 on Build GRCh38
rs1001413989 Chr.1: 71064712-71064712 on Build GRCh38
rs768884958 Chr.1: 71064688-71064688 on Build GRCh38
rs894443768 Chr.1: 71064608-71064609 on Build GRCh38
rs557260772 Chr.1: 71064529-71064529 on Build GRCh38
rs982922241 Chr.1: 71064452-71064452 on Build GRCh38
rs540199365 Chr.1: 71064466-71064466 on Build GRCh38
rs771004174 Chr.1: 71064737-71064737 on Build GRCh38
rs964560103 Chr.1: 71064432-71064432 on Build GRCh38
rs112979382 Chr.1: 71064742-71064742 on Build GRCh38
rs776997728 Chr.1: 71064838-71064838 on Build GRCh38
rs1008269805 Chr.1: 71064617-71064617 on Build GRCh38
rs553028872 Chr.1: 71064600-71064600 on Build GRCh38
rs537321616 Chr.1: 71064510-71064510 on Build GRCh38
rs564267368 Chr.1: 71064837-71064837 on Build GRCh38
rs965994634 Chr.1: 71064482-71064482 on Build GRCh38
rs11802971 Chr.1: 71064558-71064558 on Build GRCh38
rs958765811 Chr.1: 71064424-71064424 on Build GRCh38
rs370181295 Chr.1: 71064821-71064821 on Build GRCh38
rs552298743 Chr.1: 71064656-71064656 on Build GRCh38
rs956956340 Chr.1: 71064454-71064454 on Build GRCh38
rs372855432 Chr.1: 71064818-71064818 on Build GRCh38
rs562573141 Chr.1: 71064810-71064810 on Build GRCh38
rs987782645 Chr.1: 71064457-71064457 on Build GRCh38

View Full Product Details