Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6042025-6042562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRN4
Entrez Gene ID
164312
Gene Name
leucine rich repeat neuronal 4
Gene Aliases
C20orf75, NLRR-4, NLRR4, dJ1056H1.1
UniGene
Hs.149133
Gene Chromosome Location
Chr.20: 6040546-6054080 on Build GRCh38
Transcripts
NM_152611.4 XM_011529183.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5626792 c.1212G>A p.Q404Q Chr.20: 6042033-6042033 on Build GRCh38
6704496 c.1138C>T p.R380C Chr.20: 6042107-6042107 on Build GRCh38
6704512 c.1213_1214delAG p.S405fs*6 Chr.20: 6042031-6042032 on Build GRCh38
7273096 c.1153C>T p.Q385* Chr.20: 6042092-6042092 on Build GRCh38
252591 c.1138C>A p.R380S Chr.20: 6042107-6042107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553659981 Chr.20: 6042385-6042385 on Build GRCh38
rs369401908 Chr.20: 6042050-6042050 on Build GRCh38
rs990179729 Chr.20: 6042103-6042103 on Build GRCh38
rs757104075 Chr.20: 6042253-6042253 on Build GRCh38
rs150930367 Chr.20: 6042205-6042205 on Build GRCh38
rs1054106287 Chr.20: 6042361-6042361 on Build GRCh38
rs986078792 Chr.20: 6042418-6042418 on Build GRCh38
rs867722111 Chr.20: 6042326-6042326 on Build GRCh38
rs777927705 Chr.20: 6042081-6042081 on Build GRCh38
rs148688277 Chr.20: 6042498-6042498 on Build GRCh38
rs759039858 Chr.20: 6042207-6042207 on Build GRCh38
rs749384086 Chr.20: 6042166-6042166 on Build GRCh38
rs570868926 Chr.20: 6042352-6042352 on Build GRCh38
rs771479846 Chr.20: 6042203-6042203 on Build GRCh38
rs956144402 Chr.20: 6042327-6042327 on Build GRCh38
rs180918925 Chr.20: 6042409-6042409 on Build GRCh38
rs750923397 Chr.20: 6042064-6042064 on Build GRCh38
rs142910883 Chr.20: 6042107-6042107 on Build GRCh38
rs780309801 Chr.20: 6042097-6042097 on Build GRCh38
rs539524551 Chr.20: 6042530-6042530 on Build GRCh38
rs753877694 Chr.20: 6042275-6042277 on Build GRCh38
rs764857911 Chr.20: 6042135-6042135 on Build GRCh38
rs557148176 Chr.20: 6042431-6042431 on Build GRCh38
rs537070528 Chr.20: 6042323-6042323 on Build GRCh38
rs748417102 Chr.20: 6042182-6042182 on Build GRCh38
rs202048241 Chr.20: 6042067-6042067 on Build GRCh38
rs919003506 Chr.20: 6042480-6042480 on Build GRCh38
rs760677718 Chr.20: 6042532-6042532 on Build GRCh38
rs986329748 Chr.20: 6042125-6042125 on Build GRCh38
rs754618457 Chr.20: 6042072-6042072 on Build GRCh38
rs757298264 Chr.20: 6042162-6042162 on Build GRCh38
rs1048676497 Chr.20: 6042519-6042519 on Build GRCh38
rs761193242 Chr.20: 6042131-6042131 on Build GRCh38
rs560730339 Chr.20: 6042305-6042305 on Build GRCh38
rs529138479 Chr.20: 6042226-6042226 on Build GRCh38
rs910354691 Chr.20: 6042424-6042424 on Build GRCh38
rs919224528 Chr.20: 6042507-6042507 on Build GRCh38
rs768777949 Chr.20: 6042158-6042158 on Build GRCh38
rs752393963 Chr.20: 6042140-6042140 on Build GRCh38
rs944538861 Chr.20: 6042432-6042432 on Build GRCh38
rs535719373 Chr.20: 6042420-6042420 on Build GRCh38
rs904985819 Chr.20: 6042350-6042350 on Build GRCh38
rs763806187 Chr.20: 6042250-6042250 on Build GRCh38
rs751446361 Chr.20: 6042252-6042252 on Build GRCh38
rs976353772 Chr.20: 6042479-6042479 on Build GRCh38
rs764811357 Chr.20: 6042210-6042210 on Build GRCh38
rs919275429 Chr.20: 6042300-6042300 on Build GRCh38
rs758203108 Chr.20: 6042080-6042080 on Build GRCh38
rs772489757 Chr.20: 6042119-6042119 on Build GRCh38
rs923414392 Chr.20: 6042176-6042176 on Build GRCh38
rs766407477 Chr.20: 6042232-6042233 on Build GRCh38
rs762693769 Chr.20: 6042228-6042228 on Build GRCh38
rs542002484 Chr.20: 6042106-6042106 on Build GRCh38
rs767627724 Chr.20: 6042257-6042257 on Build GRCh38
rs952331446 Chr.20: 6042046-6042046 on Build GRCh38
rs753295986 Chr.20: 6042069-6042069 on Build GRCh38
rs373456456 Chr.20: 6042095-6042095 on Build GRCh38
rs1020554739 Chr.20: 6042386-6042386 on Build GRCh38
rs570714715 Chr.20: 6042052-6042052 on Build GRCh38
rs773599965 Chr.20: 6042198-6042198 on Build GRCh38
rs540567438 Chr.20: 6042200-6042200 on Build GRCh38
rs772997863 Chr.20: 6042174-6042174 on Build GRCh38
rs759121311 Chr.20: 6042477-6042479 on Build GRCh38
rs73896718 Chr.20: 6042294-6042294 on Build GRCh38
rs201145558 Chr.20: 6042075-6042075 on Build GRCh38
rs145141217 Chr.20: 6042382-6042382 on Build GRCh38
rs965027808 Chr.20: 6042405-6042405 on Build GRCh38
rs760454809 Chr.20: 6042187-6042190 on Build GRCh38
rs374733989 Chr.20: 6042077-6042077 on Build GRCh38
rs897606775 Chr.20: 6042206-6042206 on Build GRCh38
rs1032064781 Chr.20: 6042088-6042088 on Build GRCh38
rs751356312 Chr.20: 6042153-6042153 on Build GRCh38
rs6117045 Chr.20: 6042401-6042401 on Build GRCh38
rs750377706 Chr.20: 6042276-6042276 on Build GRCh38
rs762150923 Chr.20: 6042108-6042108 on Build GRCh38
rs147438354 Chr.20: 6042134-6042134 on Build GRCh38
rs778600524 Chr.20: 6042073-6042073 on Build GRCh38
rs528029625 Chr.20: 6042195-6042195 on Build GRCh38
rs777729280 Chr.20: 6042146-6042146 on Build GRCh38
rs201837434 Chr.20: 6042227-6042227 on Build GRCh38
rs769189475 Chr.20: 6042299-6042299 on Build GRCh38
rs369964130 Chr.20: 6042089-6042089 on Build GRCh38
rs373615341 Chr.20: 6042400-6042400 on Build GRCh38
rs117101542 Chr.20: 6042167-6042167 on Build GRCh38
rs188024999 Chr.20: 6042122-6042122 on Build GRCh38
rs1003474714 Chr.20: 6042321-6042321 on Build GRCh38
rs34836928 Chr.20: 6042078-6042079 on Build GRCh38
rs953538799 Chr.20: 6042470-6042470 on Build GRCh38
rs752530533 Chr.20: 6042370-6042370 on Build GRCh38
rs1013498100 Chr.20: 6042063-6042063 on Build GRCh38
rs778790357 Chr.20: 6042169-6042169 on Build GRCh38
rs749353171 Chr.20: 6042100-6042100 on Build GRCh38

View Full Product Details