Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37198965-37199468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPN2
Entrez Gene ID
6185
Gene Name
ribophorin II
Gene Aliases
RIBIIR, RPN-II, RPNII, SWP1
UniGene
Hs.370895
Gene Chromosome Location
Chr.20: 37179046-37241623 on Build GRCh38
Transcripts
NM_001135771.2 NM_001324299.1 NM_001324301.1 NM_001324302.1 NM_001324303.1 NM_001324304.1 NM_001324305.1 NM_001324306.1 NM_002951.4 XM_006723851.2 XM_006723852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6430908 c.446C>A p.T149N Chr.20: 37199192-37199192 on Build GRCh38
3911229 c.412C>T p.P138S Chr.20: 37199158-37199158 on Build GRCh38
1224220 c.479C>T p.A160V Chr.20: 37199225-37199225 on Build GRCh38
226842 c.442C>T p.L148F Chr.20: 37199188-37199188 on Build GRCh38
6093204 c.435C>G p.L145L Chr.20: 37199181-37199181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753444079 Chr.20: 37199043-37199043 on Build GRCh38
rs1004851734 Chr.20: 37199304-37199304 on Build GRCh38
rs745715557 Chr.20: 37199068-37199068 on Build GRCh38
rs746059869 Chr.20: 37199421-37199421 on Build GRCh38
rs553035371 Chr.20: 37199100-37199100 on Build GRCh38
rs951565669 Chr.20: 37199286-37199286 on Build GRCh38
rs982093678 Chr.20: 37199351-37199351 on Build GRCh38
rs575407563 Chr.20: 37199192-37199192 on Build GRCh38
rs769144386 Chr.20: 37199268-37199268 on Build GRCh38
rs546650562 Chr.20: 37199245-37199245 on Build GRCh38
rs562016524 Chr.20: 37199343-37199343 on Build GRCh38
rs1056949197 Chr.20: 37199191-37199191 on Build GRCh38
rs754710451 Chr.20: 37199251-37199251 on Build GRCh38
rs991637958 Chr.20: 37199331-37199331 on Build GRCh38
rs750057859 Chr.20: 37199139-37199139 on Build GRCh38
rs772692443 Chr.20: 37199220-37199220 on Build GRCh38
rs753454532 Chr.20: 37199132-37199132 on Build GRCh38
rs764791936 Chr.20: 37199240-37199240 on Build GRCh38
rs749073065 Chr.20: 37199285-37199285 on Build GRCh38
rs748313134 Chr.20: 37199186-37199186 on Build GRCh38
rs755012885 Chr.20: 37199165-37199165 on Build GRCh38
rs746917244 Chr.20: 37199085-37199085 on Build GRCh38
rs1039309975 Chr.20: 37199438-37199438 on Build GRCh38
rs370382737 Chr.20: 37199126-37199126 on Build GRCh38
rs916179119 Chr.20: 37199118-37199118 on Build GRCh38
rs150520116 Chr.20: 37199052-37199052 on Build GRCh38
rs755010089 Chr.20: 37199330-37199330 on Build GRCh38
rs756902264 Chr.20: 37199047-37199047 on Build GRCh38
rs916026225 Chr.20: 37199335-37199335 on Build GRCh38
rs759814470 Chr.20: 37199211-37199211 on Build GRCh38
rs934461848 Chr.20: 37199161-37199161 on Build GRCh38
rs780172990 Chr.20: 37199084-37199084 on Build GRCh38
rs951496866 Chr.20: 37198989-37198989 on Build GRCh38
rs778993349 Chr.20: 37199342-37199342 on Build GRCh38
rs748829395 Chr.20: 37199190-37199191 on Build GRCh38
rs776720366 Chr.20: 37199409-37199409 on Build GRCh38
rs770063411 Chr.20: 37199105-37199105 on Build GRCh38
rs1000328675 Chr.20: 37198991-37198991 on Build GRCh38
rs116240215 Chr.20: 37199387-37199387 on Build GRCh38
rs144151309 Chr.20: 37199140-37199140 on Build GRCh38
rs904201171 Chr.20: 37198990-37198990 on Build GRCh38
rs951596815 Chr.20: 37199123-37199123 on Build GRCh38
rs963023318 Chr.20: 37199341-37199341 on Build GRCh38
rs147803642 Chr.20: 37199197-37199197 on Build GRCh38
rs375178656 Chr.20: 37199148-37199148 on Build GRCh38
rs774709106 Chr.20: 37199198-37199198 on Build GRCh38
rs937157330 Chr.20: 37199177-37199177 on Build GRCh38
rs552994576 Chr.20: 37199445-37199445 on Build GRCh38
rs750026130 Chr.20: 37199059-37199059 on Build GRCh38
rs200831578 Chr.20: 37199076-37199076 on Build GRCh38
rs746973238 Chr.20: 37199178-37199178 on Build GRCh38
rs763344652 Chr.20: 37199111-37199111 on Build GRCh38
rs771074467 Chr.20: 37199347-37199347 on Build GRCh38
rs529026231 Chr.20: 37199295-37199295 on Build GRCh38
rs4812882 Chr.20: 37199439-37199439 on Build GRCh38
rs531083756 Chr.20: 37199374-37199374 on Build GRCh38
rs763515303 Chr.20: 37199127-37199127 on Build GRCh38
rs765022625 Chr.20: 37199138-37199138 on Build GRCh38
rs2076398 Chr.20: 37199279-37199279 on Build GRCh38
rs376893781 Chr.20: 37199254-37199254 on Build GRCh38
rs751458474 Chr.20: 37199153-37199153 on Build GRCh38
rs769892718 Chr.20: 37199263-37199263 on Build GRCh38
rs926731087 Chr.20: 37199169-37199169 on Build GRCh38
rs769830164 Chr.20: 37199187-37199187 on Build GRCh38
rs570844390 Chr.20: 37199432-37199432 on Build GRCh38
rs201261133 Chr.20: 37199241-37199241 on Build GRCh38
rs765946560 Chr.20: 37199242-37199242 on Build GRCh38
rs749460659 Chr.20: 37199195-37199195 on Build GRCh38
rs761393245 Chr.20: 37199137-37199137 on Build GRCh38
rs962763966 Chr.20: 37199147-37199147 on Build GRCh38
rs999894436 Chr.20: 37199253-37199253 on Build GRCh38
rs372064976 Chr.20: 37199225-37199225 on Build GRCh38
rs773397382 Chr.20: 37199106-37199106 on Build GRCh38
rs202197936 Chr.20: 37199214-37199214 on Build GRCh38
rs377232352 Chr.20: 37199267-37199267 on Build GRCh38
rs376045139 Chr.20: 37199032-37199032 on Build GRCh38
rs200634422 Chr.20: 37199095-37199095 on Build GRCh38
rs144329207 Chr.20: 37199103-37199103 on Build GRCh38
rs934295945 Chr.20: 37199369-37199369 on Build GRCh38
rs950326392 Chr.20: 37199119-37199119 on Build GRCh38
rs992258545 Chr.20: 37199363-37199363 on Build GRCh38
rs1038887694 Chr.20: 37199427-37199427 on Build GRCh38
rs117835643 Chr.20: 37199221-37199221 on Build GRCh38
rs924340468 Chr.20: 37199368-37199368 on Build GRCh38
rs774740409 Chr.20: 37199122-37199122 on Build GRCh38
rs374512118 Chr.20: 37199001-37199001 on Build GRCh38
rs563539009 Chr.20: 37199185-37199185 on Build GRCh38
rs11906195 Chr.20: 37199310-37199310 on Build GRCh38
rs763608737 Chr.20: 37199038-37199038 on Build GRCh38
rs771242581 Chr.20: 37199114-37199114 on Build GRCh38
rs78121372 Chr.20: 37199017-37199017 on Build GRCh38

View Full Product Details