Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38628685-38629195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP40
Entrez Gene ID
343578
Gene Name
Rho GTPase activating protein 40
Gene Aliases
C20orf95, dJ1100H13.4
UniGene
Hs.451997
Gene Chromosome Location
Chr.20: 38601934-38650652 on Build GRCh38
Transcripts
NM_001164431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1318836 c.409T>C p.S137P Chr.20: 38628936-38628936 on Build GRCh38
3963552 c.400-6G>T p.? Chr.20: 38628921-38628921 on Build GRCh38
2761534 c.408G>A p.S136S Chr.20: 38628935-38628935 on Build GRCh38
5818448 c.400-2A>T p.? Chr.20: 38628925-38628925 on Build GRCh38
1592610 c.400-1G>T p.? Chr.20: 38628926-38628926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780775073 Chr.20: 38628947-38628947 on Build GRCh38
rs1040063460 Chr.20: 38628980-38628981 on Build GRCh38
rs76530900 Chr.20: 38628714-38628714 on Build GRCh38
rs1000808693 Chr.20: 38629016-38629016 on Build GRCh38
rs6070809 Chr.20: 38628947-38628947 on Build GRCh38
rs944419632 Chr.20: 38628975-38628975 on Build GRCh38
rs755808297 Chr.20: 38628954-38628954 on Build GRCh38
rs113856214 Chr.20: 38628921-38628921 on Build GRCh38
rs888647249 Chr.20: 38629068-38629068 on Build GRCh38
rs73296981 Chr.20: 38628893-38628893 on Build GRCh38
rs901339608 Chr.20: 38628765-38628765 on Build GRCh38
rs180834546 Chr.20: 38628833-38628833 on Build GRCh38
rs1007824211 Chr.20: 38629095-38629095 on Build GRCh38
rs930118110 Chr.20: 38629056-38629056 on Build GRCh38
rs910894649 Chr.20: 38628863-38628863 on Build GRCh38
rs967966794 Chr.20: 38628772-38628772 on Build GRCh38
rs35399405 Chr.20: 38628741-38628741 on Build GRCh38
rs946610702 Chr.20: 38628948-38628948 on Build GRCh38
rs1000432662 Chr.20: 38628793-38628793 on Build GRCh38
rs956487770 Chr.20: 38628934-38628934 on Build GRCh38
rs889334102 Chr.20: 38629078-38629078 on Build GRCh38
rs531518594 Chr.20: 38629007-38629007 on Build GRCh38
rs1014761636 Chr.20: 38628759-38628759 on Build GRCh38
rs546002152 Chr.20: 38628935-38628935 on Build GRCh38
rs773816327 Chr.20: 38628869-38628869 on Build GRCh38
rs1017925919 Chr.20: 38629116-38629116 on Build GRCh38
rs76359829 Chr.20: 38629071-38629071 on Build GRCh38
rs368771030 Chr.20: 38628901-38628901 on Build GRCh38
rs1005831952 Chr.20: 38628888-38628888 on Build GRCh38
rs753732923 Chr.20: 38628981-38628981 on Build GRCh38
rs1023924634 Chr.20: 38629075-38629075 on Build GRCh38
rs768220386 Chr.20: 38628766-38628766 on Build GRCh38
rs768702378 Chr.20: 38628890-38628890 on Build GRCh38
rs8118958 Chr.20: 38628711-38628711 on Build GRCh38
rs560900459 Chr.20: 38629090-38629090 on Build GRCh38
rs545515793 Chr.20: 38628941-38628941 on Build GRCh38
rs752084657 Chr.20: 38628949-38628949 on Build GRCh38
rs527727194 Chr.20: 38629048-38629048 on Build GRCh38
rs976921691 Chr.20: 38628894-38628894 on Build GRCh38
rs370495268 Chr.20: 38629009-38629009 on Build GRCh38
rs905158105 Chr.20: 38629156-38629157 on Build GRCh38
rs1047207949 Chr.20: 38629064-38629064 on Build GRCh38
rs1003548452 Chr.20: 38629170-38629170 on Build GRCh38
rs372741030 Chr.20: 38628802-38628802 on Build GRCh38
rs888434983 Chr.20: 38628708-38628708 on Build GRCh38
rs560382894 Chr.20: 38628967-38628967 on Build GRCh38
rs909841821 Chr.20: 38628946-38628946 on Build GRCh38
rs531521663 Chr.20: 38629162-38629162 on Build GRCh38
rs957686869 Chr.20: 38628809-38628809 on Build GRCh38
rs780204657 Chr.20: 38629158-38629158 on Build GRCh38
rs982185969 Chr.20: 38628906-38628906 on Build GRCh38
rs965124313 Chr.20: 38628892-38628892 on Build GRCh38
rs563090999 Chr.20: 38628933-38628933 on Build GRCh38

View Full Product Details