Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63659147-63659643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTEL1
Entrez Gene ID
51750
Gene Name
regulator of telomere elongation helicase 1
Gene Aliases
C20orf41, DKCA4, DKCB5, NHL, PFBMFT3, RTEL
UniGene
Hs.745057
Gene Chromosome Location
Chr.20: 63657810-63696253 on Build GRCh38
Transcripts
NM_001283009.1 NM_001283010.1 NM_016434.3 NM_032957.4 NR_037882.1

Gene Symbol
RTEL1-TNFRSF6B
Entrez Gene ID
100533107
Gene Name
RTEL1-TNFRSF6B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.745057
Gene Chromosome Location
Chr.20: 63657810-63698698 on Build GRCh38
Transcripts
NM_001283009.1 NM_001283010.1 NM_016434.3 NM_032957.4 NR_037882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2932383 c.5C>T p.P2L Chr.20: 63659407-63659407 on Build GRCh38
2932384 c.5C>T p.P2L Chr.20: 63659407-63659407 on Build GRCh38
5592947 c.4C>T p.P2S Chr.20: 63659406-63659406 on Build GRCh38
1224350 c.34G>T p.D12Y Chr.20: 63659436-63659436 on Build GRCh38
5592946 c.4C>T p.P2S Chr.20: 63659406-63659406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002550569 Chr.20: 63659348-63659348 on Build GRCh38
rs115422766 Chr.20: 63659595-63659595 on Build GRCh38
rs758025445 Chr.20: 63659413-63659413 on Build GRCh38
rs778302679 Chr.20: 63659391-63659392 on Build GRCh38
rs2297432 Chr.20: 63659310-63659310 on Build GRCh38
rs6062479 Chr.20: 63659369-63659369 on Build GRCh38
rs555745067 Chr.20: 63659343-63659343 on Build GRCh38
rs779512674 Chr.20: 63659552-63659552 on Build GRCh38
rs748145576 Chr.20: 63659449-63659449 on Build GRCh38
rs747335734 Chr.20: 63659471-63659472 on Build GRCh38
rs1051860914 Chr.20: 63659206-63659206 on Build GRCh38
rs765143327 Chr.20: 63659374-63659374 on Build GRCh38
rs1013785003 Chr.20: 63659253-63659253 on Build GRCh38
rs916224821 Chr.20: 63659471-63659471 on Build GRCh38
rs920618786 Chr.20: 63659607-63659607 on Build GRCh38
rs1025165074 Chr.20: 63659263-63659263 on Build GRCh38
rs76322185 Chr.20: 63659358-63659358 on Build GRCh38
rs879413437 Chr.20: 63659232-63659232 on Build GRCh38
rs373760953 Chr.20: 63659389-63659389 on Build GRCh38
rs756345215 Chr.20: 63659290-63659290 on Build GRCh38
rs760616462 Chr.20: 63659513-63659513 on Build GRCh38
rs865824903 Chr.20: 63659616-63659616 on Build GRCh38
rs1036922280 Chr.20: 63659193-63659193 on Build GRCh38
rs201569343 Chr.20: 63659514-63659514 on Build GRCh38
rs757731065 Chr.20: 63659517-63659517 on Build GRCh38
rs1038327156 Chr.20: 63659234-63659234 on Build GRCh38
rs757708000 Chr.20: 63659520-63659521 on Build GRCh38
rs567430622 Chr.20: 63659563-63659563 on Build GRCh38
rs746696107 Chr.20: 63659555-63659555 on Build GRCh38
rs746437645 Chr.20: 63659432-63659432 on Build GRCh38
rs369972043 Chr.20: 63659534-63659534 on Build GRCh38
rs544693377 Chr.20: 63659602-63659602 on Build GRCh38
rs1035447147 Chr.20: 63659363-63659363 on Build GRCh38
rs767851345 Chr.20: 63659498-63659498 on Build GRCh38
rs184263470 Chr.20: 63659202-63659202 on Build GRCh38
rs117750724 Chr.20: 63659201-63659201 on Build GRCh38
rs765782315 Chr.20: 63659520-63659520 on Build GRCh38
rs954545020 Chr.20: 63659559-63659559 on Build GRCh38
rs746271642 Chr.20: 63659411-63659411 on Build GRCh38
rs537744603 Chr.20: 63659359-63659359 on Build GRCh38
rs563905352 Chr.20: 63659301-63659301 on Build GRCh38
rs931001119 Chr.20: 63659401-63659401 on Build GRCh38
rs372129544 Chr.20: 63659367-63659367 on Build GRCh38
rs752228931 Chr.20: 63659381-63659381 on Build GRCh38
rs763778752 Chr.20: 63659422-63659422 on Build GRCh38
rs13043797 Chr.20: 63659344-63659344 on Build GRCh38
rs907962257 Chr.20: 63659597-63659597 on Build GRCh38
rs543148818 Chr.20: 63659560-63659560 on Build GRCh38
rs902654004 Chr.20: 63659282-63659282 on Build GRCh38
rs754384977 Chr.20: 63659399-63659399 on Build GRCh38
rs751341410 Chr.20: 63659283-63659283 on Build GRCh38
rs779282812 Chr.20: 63659410-63659410 on Build GRCh38
rs973426844 Chr.20: 63659604-63659604 on Build GRCh38
rs894047302 Chr.20: 63659580-63659580 on Build GRCh38
rs759400305 Chr.20: 63659317-63659317 on Build GRCh38
rs757516930 Chr.20: 63659407-63659407 on Build GRCh38
rs759796755 Chr.20: 63659377-63659377 on Build GRCh38
rs375849124 Chr.20: 63659496-63659496 on Build GRCh38
rs770615551 Chr.20: 63659273-63659273 on Build GRCh38
rs532800228 Chr.20: 63659309-63659309 on Build GRCh38
rs974684662 Chr.20: 63659456-63659456 on Build GRCh38
rs574193419 Chr.20: 63659550-63659550 on Build GRCh38
rs776379930 Chr.20: 63659437-63659437 on Build GRCh38
rs754260726 Chr.20: 63659516-63659516 on Build GRCh38
rs768473584 Chr.20: 63659366-63659366 on Build GRCh38
rs548912361 Chr.20: 63659352-63659352 on Build GRCh38
rs996182809 Chr.20: 63659341-63659341 on Build GRCh38
rs534207784 Chr.20: 63659391-63659391 on Build GRCh38
rs754403563 Chr.20: 63659556-63659556 on Build GRCh38
rs773115541 Chr.20: 63659479-63659479 on Build GRCh38
rs140905272 Chr.20: 63659342-63659342 on Build GRCh38
rs878926884 Chr.20: 63659303-63659303 on Build GRCh38
rs769888088 Chr.20: 63659453-63659453 on Build GRCh38
rs958315814 Chr.20: 63659327-63659327 on Build GRCh38
rs1028969781 Chr.20: 63659603-63659603 on Build GRCh38
rs775788760 Chr.20: 63659510-63659510 on Build GRCh38
rs764460604 Chr.20: 63659205-63659208 on Build GRCh38
rs907841975 Chr.20: 63659211-63659212 on Build GRCh38
rs779748543 Chr.20: 63659414-63659414 on Build GRCh38
rs774673265 Chr.20: 63659375-63659375 on Build GRCh38
rs751593587 Chr.20: 63659247-63659247 on Build GRCh38
rs181776401 Chr.20: 63659564-63659564 on Build GRCh38
rs751168157 Chr.20: 63659431-63659431 on Build GRCh38
rs768223828 Chr.20: 63659433-63659433 on Build GRCh38
rs973064119 Chr.20: 63659387-63659387 on Build GRCh38
rs189025756 Chr.20: 63659383-63659383 on Build GRCh38
rs138992534 Chr.20: 63659592-63659592 on Build GRCh38

View Full Product Details