Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32733760-32734269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1
Entrez Gene ID
94104
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
UniGene
Hs.644004
Gene Chromosome Location
Chr.21: 32733899-32771858 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
UniGene
Hs.657123
Gene Chromosome Location
Chr.21: 32728114-32743122 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

SNP Detail

SNP ID Chromosome Location
rs371980047 Chr.21: 32734120-32734120 on Build GRCh38
rs140885839 Chr.21: 32734227-32734227 on Build GRCh38
rs899824608 Chr.21: 32733861-32733861 on Build GRCh38
rs960005967 Chr.21: 32734145-32734145 on Build GRCh38
rs988867038 Chr.21: 32733844-32733847 on Build GRCh38
rs923056021 Chr.21: 32733945-32733945 on Build GRCh38
rs553400564 Chr.21: 32733877-32733877 on Build GRCh38
rs914084623 Chr.21: 32733793-32733793 on Build GRCh38
rs79527546 Chr.21: 32733870-32733870 on Build GRCh38
rs1022745087 Chr.21: 32734049-32734049 on Build GRCh38
rs1015424459 Chr.21: 32733959-32733959 on Build GRCh38
rs960098447 Chr.21: 32733821-32733821 on Build GRCh38
rs970216523 Chr.21: 32734062-32734062 on Build GRCh38
rs888343170 Chr.21: 32733946-32733946 on Build GRCh38
rs944112249 Chr.21: 32733815-32733815 on Build GRCh38
rs959883394 Chr.21: 32733961-32733961 on Build GRCh38
rs13047447 Chr.21: 32734137-32734137 on Build GRCh38
rs925640071 Chr.21: 32734115-32734115 on Build GRCh38
rs770344529 Chr.21: 32733986-32733987 on Build GRCh38
rs1038397668 Chr.21: 32734092-32734092 on Build GRCh38
rs772358521 Chr.21: 32734114-32734114 on Build GRCh38
rs761961033 Chr.21: 32733970-32733970 on Build GRCh38
rs913743356 Chr.21: 32733844-32733844 on Build GRCh38
rs966981057 Chr.21: 32734201-32734201 on Build GRCh38
rs558736231 Chr.21: 32733850-32733850 on Build GRCh38
rs1041617852 Chr.21: 32734174-32734174 on Build GRCh38
rs980095982 Chr.21: 32733792-32733792 on Build GRCh38
rs944245026 Chr.21: 32734155-32734155 on Build GRCh38
rs773656891 Chr.21: 32733995-32733998 on Build GRCh38
rs570666066 Chr.21: 32733859-32733859 on Build GRCh38
rs1020335240 Chr.21: 32734195-32734195 on Build GRCh38
rs545736546 Chr.21: 32734188-32734188 on Build GRCh38
rs538047277 Chr.21: 32733802-32733802 on Build GRCh38
rs886613333 Chr.21: 32733976-32733976 on Build GRCh38
rs979288040 Chr.21: 32734203-32734203 on Build GRCh38
rs1041175761 Chr.21: 32733846-32733846 on Build GRCh38
rs932749361 Chr.21: 32734217-32734217 on Build GRCh38
rs1006380036 Chr.21: 32734038-32734038 on Build GRCh38
rs979767251 Chr.21: 32733847-32733848 on Build GRCh38
rs773634618 Chr.21: 32734128-32734128 on Build GRCh38
rs1014416926 Chr.21: 32733983-32733983 on Build GRCh38
rs560329444 Chr.21: 32733840-32733841 on Build GRCh38
rs537340108 Chr.21: 32733812-32733812 on Build GRCh38
rs914070195 Chr.21: 32734149-32734149 on Build GRCh38
rs370211158 Chr.21: 32733958-32733958 on Build GRCh38
rs888478975 Chr.21: 32734235-32734235 on Build GRCh38

View Full Product Details