Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36752596-36753122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
UniGene
Hs.371350
Gene Chromosome Location
Chr.21: 36750888-36990254 on Build GRCh38
Transcripts
NM_000411.6 NM_001242784.1 NM_001242785.1 XM_005260953.3 XM_005260955.3 XM_005260956.3 XM_006723994.2 XM_006723995.1 XM_011529538.1 XM_011529539.2 XM_011529541.2 XM_017028330.1

SNP Detail

SNP ID Chromosome Location
rs937744975 Chr.21: 36752909-36752909 on Build GRCh38
rs896194556 Chr.21: 36752948-36752948 on Build GRCh38
rs34507014 Chr.21: 36752986-36752987 on Build GRCh38
rs926377249 Chr.21: 36752867-36752867 on Build GRCh38
rs1035402957 Chr.21: 36752675-36752675 on Build GRCh38
rs3191054 Chr.21: 36753058-36753058 on Build GRCh38
rs975833877 Chr.21: 36752696-36752696 on Build GRCh38
rs1009642522 Chr.21: 36752956-36752957 on Build GRCh38
rs764870505 Chr.21: 36752918-36752918 on Build GRCh38
rs906661477 Chr.21: 36753060-36753060 on Build GRCh38
rs368517497 Chr.21: 36752996-36752996 on Build GRCh38
rs886057069 Chr.21: 36752665-36752665 on Build GRCh38
rs940693100 Chr.21: 36752631-36752631 on Build GRCh38
rs964074704 Chr.21: 36752666-36752666 on Build GRCh38
rs757830763 Chr.21: 36753019-36753019 on Build GRCh38
rs147913873 Chr.21: 36752776-36752776 on Build GRCh38
rs948364030 Chr.21: 36753043-36753043 on Build GRCh38
rs78926199 Chr.21: 36752762-36752762 on Build GRCh38
rs908785866 Chr.21: 36752780-36752780 on Build GRCh38
rs941536736 Chr.21: 36752800-36752800 on Build GRCh38
rs886057068 Chr.21: 36752631-36752632 on Build GRCh38
rs1009448619 Chr.21: 36752635-36752635 on Build GRCh38
rs191827594 Chr.21: 36753028-36753028 on Build GRCh38
rs904928361 Chr.21: 36752621-36752621 on Build GRCh38
rs530326784 Chr.21: 36753096-36753096 on Build GRCh38
rs922593281 Chr.21: 36752737-36752737 on Build GRCh38
rs751998662 Chr.21: 36752920-36752920 on Build GRCh38
rs955682487 Chr.21: 36752710-36752710 on Build GRCh38
rs765400304 Chr.21: 36753098-36753098 on Build GRCh38
rs750848969 Chr.21: 36753082-36753082 on Build GRCh38
rs1020826468 Chr.21: 36752641-36752641 on Build GRCh38
rs955435132 Chr.21: 36753085-36753085 on Build GRCh38
rs73902720 Chr.21: 36753040-36753040 on Build GRCh38
rs572273747 Chr.21: 36752985-36752985 on Build GRCh38
rs903763142 Chr.21: 36752971-36752971 on Build GRCh38
rs565500474 Chr.21: 36752973-36752973 on Build GRCh38
rs769979101 Chr.21: 36752853-36752862 on Build GRCh38
rs1045732079 Chr.21: 36753056-36753056 on Build GRCh38
rs1003685143 Chr.21: 36753081-36753081 on Build GRCh38
rs776064678 Chr.21: 36752652-36752652 on Build GRCh38
rs1050706 Chr.21: 36752642-36752642 on Build GRCh38
rs921754749 Chr.21: 36752840-36752840 on Build GRCh38
rs1042317201 Chr.21: 36752970-36752970 on Build GRCh38
rs536690378 Chr.21: 36752659-36752659 on Build GRCh38
rs141644229 Chr.21: 36752922-36752922 on Build GRCh38
rs766046373 Chr.21: 36752632-36752632 on Build GRCh38
rs968686301 Chr.21: 36752809-36752809 on Build GRCh38
rs1050707 Chr.21: 36752640-36752640 on Build GRCh38
rs1056108761 Chr.21: 36752632-36752632 on Build GRCh38
rs974978132 Chr.21: 36752824-36752824 on Build GRCh38
rs1017363502 Chr.21: 36752986-36752986 on Build GRCh38
rs573958407 Chr.21: 36752656-36752656 on Build GRCh38
rs201997019 Chr.21: 36752646-36752649 on Build GRCh38
rs896405323 Chr.21: 36752632-36752633 on Build GRCh38
rs1038526586 Chr.21: 36752837-36752837 on Build GRCh38
rs1012811889 Chr.21: 36752649-36752649 on Build GRCh38

View Full Product Details