Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45986432-45986936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A1
Entrez Gene ID
1291
Gene Name
collagen type VI alpha 1
Gene Aliases
BTHLM1, OPLL, UCHMD1
UniGene
Hs.474053
Gene Chromosome Location
Chr.21: 45981749-46005049 on Build GRCh38
Transcripts
NM_001848.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318676 c.573C>A p.I191I Chr.21: 45986670-45986670 on Build GRCh38
5668832 c.537C>A p.A179A Chr.21: 45986634-45986634 on Build GRCh38
6196018 c.543C>G p.H181Q Chr.21: 45986640-45986640 on Build GRCh38
3912299 c.481C>T p.P161S Chr.21: 45986578-45986578 on Build GRCh38
6443895 c.472G>A p.D158N Chr.21: 45986569-45986569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767229013 Chr.21: 45986707-45986707 on Build GRCh38
rs904636925 Chr.21: 45986461-45986461 on Build GRCh38
rs749929221 Chr.21: 45986629-45986629 on Build GRCh38
rs772877614 Chr.21: 45986712-45986712 on Build GRCh38
rs868272036 Chr.21: 45986533-45986533 on Build GRCh38
rs566110491 Chr.21: 45986775-45986775 on Build GRCh38
rs398123636 Chr.21: 45986646-45986646 on Build GRCh38
rs747098777 Chr.21: 45986902-45986902 on Build GRCh38
rs758644548 Chr.21: 45986896-45986896 on Build GRCh38
rs983581635 Chr.21: 45986762-45986762 on Build GRCh38
rs748270048 Chr.21: 45986566-45986566 on Build GRCh38
rs943889175 Chr.21: 45986850-45986850 on Build GRCh38
rs1035822148 Chr.21: 45986754-45986754 on Build GRCh38
rs557525385 Chr.21: 45986808-45986808 on Build GRCh38
rs778931297 Chr.21: 45986563-45986563 on Build GRCh38
rs1047279931 Chr.21: 45986839-45986839 on Build GRCh38
rs536288320 Chr.21: 45986734-45986734 on Build GRCh38
rs754507 Chr.21: 45986698-45986698 on Build GRCh38
rs149391064 Chr.21: 45986891-45986891 on Build GRCh38
rs1001716499 Chr.21: 45986749-45986749 on Build GRCh38
rs776935888 Chr.21: 45986694-45986694 on Build GRCh38
rs1002083012 Chr.21: 45986467-45986467 on Build GRCh38
rs1053341298 Chr.21: 45986854-45986854 on Build GRCh38
rs890926408 Chr.21: 45986695-45986695 on Build GRCh38
rs941806554 Chr.21: 45986535-45986535 on Build GRCh38
rs773280332 Chr.21: 45986571-45986571 on Build GRCh38
rs756761439 Chr.21: 45986494-45986494 on Build GRCh38
rs764985107 Chr.21: 45986736-45986736 on Build GRCh38
rs781164042 Chr.21: 45986531-45986531 on Build GRCh38
rs741956 Chr.21: 45986507-45986507 on Build GRCh38
rs547985792 Chr.21: 45986771-45986771 on Build GRCh38
rs766126332 Chr.21: 45986697-45986697 on Build GRCh38
rs536649872 Chr.21: 45986856-45986856 on Build GRCh38
rs190029634 Chr.21: 45986496-45986496 on Build GRCh38
rs974511722 Chr.21: 45986727-45986727 on Build GRCh38
rs761552446 Chr.21: 45986622-45986622 on Build GRCh38
rs761605481 Chr.21: 45986705-45986705 on Build GRCh38
rs559158458 Chr.21: 45986661-45986661 on Build GRCh38
rs376650683 Chr.21: 45986568-45986568 on Build GRCh38
rs775893001 Chr.21: 45986704-45986704 on Build GRCh38
rs766184155 Chr.21: 45986645-45986645 on Build GRCh38
rs146071423 Chr.21: 45986616-45986616 on Build GRCh38
rs980266782 Chr.21: 45986594-45986594 on Build GRCh38
rs985342849 Chr.21: 45986817-45986817 on Build GRCh38
rs777077720 Chr.21: 45986580-45986580 on Build GRCh38
rs913125478 Chr.21: 45986746-45986746 on Build GRCh38
rs987297259 Chr.21: 45986743-45986744 on Build GRCh38
rs767176038 Chr.21: 45986628-45986628 on Build GRCh38
rs777932859 Chr.21: 45986897-45986897 on Build GRCh38
rs754391596 Chr.21: 45986900-45986901 on Build GRCh38
rs757227732 Chr.21: 45986908-45986908 on Build GRCh38
rs398123637 Chr.21: 45986697-45986698 on Build GRCh38
rs891230395 Chr.21: 45986456-45986456 on Build GRCh38
rs973780191 Chr.21: 45986787-45986787 on Build GRCh38
rs138916267 Chr.21: 45986887-45986887 on Build GRCh38
rs540819892 Chr.21: 45986647-45986647 on Build GRCh38
rs750452623 Chr.21: 45986499-45986499 on Build GRCh38
rs777883035 Chr.21: 45986677-45986677 on Build GRCh38
rs756086657 Chr.21: 45986501-45986501 on Build GRCh38
rs886042409 Chr.21: 45986653-45986653 on Build GRCh38
rs111710378 Chr.21: 45986696-45986697 on Build GRCh38
rs373970248 Chr.21: 45986607-45986607 on Build GRCh38
rs111887380 Chr.21: 45986893-45986893 on Build GRCh38
rs993069719 Chr.21: 45986497-45986497 on Build GRCh38
rs568760186 Chr.21: 45986748-45986748 on Build GRCh38
rs1055863032 Chr.21: 45986798-45986798 on Build GRCh38
rs767370298 Chr.21: 45986582-45986582 on Build GRCh38
rs145049545 Chr.21: 45986761-45986761 on Build GRCh38
rs1015367935 Chr.21: 45986776-45986776 on Build GRCh38
rs185362521 Chr.21: 45986488-45986488 on Build GRCh38
rs371721318 Chr.21: 45986733-45986733 on Build GRCh38
rs554986351 Chr.21: 45986858-45986858 on Build GRCh38
rs1049577216 Chr.21: 45986455-45986455 on Build GRCh38
rs747557222 Chr.21: 45986574-45986574 on Build GRCh38
rs1028686739 Chr.21: 45986904-45986904 on Build GRCh38
rs770664917 Chr.21: 45986747-45986747 on Build GRCh38
rs540036205 Chr.21: 45986799-45986799 on Build GRCh38
rs552872000 Chr.21: 45986892-45986892 on Build GRCh38
rs398123638 Chr.21: 45986693-45986693 on Build GRCh38
rs117470181 Chr.21: 45986722-45986722 on Build GRCh38
rs754667407 Chr.21: 45986651-45986651 on Build GRCh38
rs370889024 Chr.21: 45986605-45986605 on Build GRCh38
rs921797028 Chr.21: 45986600-45986600 on Build GRCh38
rs756141940 Chr.21: 45986636-45986636 on Build GRCh38
rs779947824 Chr.21: 45986506-45986506 on Build GRCh38
rs768951384 Chr.21: 45986518-45986518 on Build GRCh38
rs200132623 Chr.21: 45986598-45986598 on Build GRCh38
rs531559651 Chr.21: 45986715-45986715 on Build GRCh38
rs961406133 Chr.21: 45986782-45986782 on Build GRCh38
rs867218507 Chr.21: 45986554-45986554 on Build GRCh38
rs746258711 Chr.21: 45986697-45986697 on Build GRCh38
rs111850281 Chr.21: 45986687-45986687 on Build GRCh38
rs996707205 Chr.21: 45986453-45986453 on Build GRCh38
rs983091594 Chr.21: 45986699-45986699 on Build GRCh38
rs61751027 Chr.21: 45986676-45986676 on Build GRCh38
rs771408273 Chr.21: 45986578-45986578 on Build GRCh38
rs747043727 Chr.21: 45986685-45986685 on Build GRCh38
rs569738710 Chr.21: 45986849-45986849 on Build GRCh38
rs375177807 Chr.21: 45986515-45986515 on Build GRCh38

View Full Product Details