Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29328344-29328848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1B1
Entrez Gene ID
162
Gene Name
adaptor related protein complex 1 beta 1 subunit
Gene Aliases
ADTB1, AP105A, BAM22, CLAPB2
UniGene
Hs.368794
Gene Chromosome Location
Chr.22: 29327680-29388583 on Build GRCh38
Transcripts
NM_001127.3 NM_001166019.1 NM_145730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4834051 c.2802G>A p.E934E Chr.22: 29328839-29328839 on Build GRCh38
6347757 c.2825C>A p.A942D Chr.22: 29328846-29328846 on Build GRCh38
4834050 c.2832G>A p.E944E Chr.22: 29328839-29328839 on Build GRCh38
1327282 c.2833A>G p.T945A Chr.22: 29328838-29328838 on Build GRCh38
6347758 c.2795C>A p.A932D Chr.22: 29328846-29328846 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs533677242 Chr.22: 29328762-29328762 on Build GRCh38
rs979151948 Chr.22: 29328633-29328633 on Build GRCh38
rs577558660 Chr.22: 29328419-29328419 on Build GRCh38
rs555938386 Chr.22: 29328391-29328391 on Build GRCh38
rs564405243 Chr.22: 29328498-29328498 on Build GRCh38
rs781249234 Chr.22: 29328800-29328800 on Build GRCh38
rs948688764 Chr.22: 29328367-29328367 on Build GRCh38
rs143241832 Chr.22: 29328566-29328566 on Build GRCh38
rs772121462 Chr.22: 29328802-29328803 on Build GRCh38
rs1034842595 Chr.22: 29328586-29328586 on Build GRCh38
rs749250270 Chr.22: 29328482-29328482 on Build GRCh38
rs937313068 Chr.22: 29328729-29328729 on Build GRCh38
rs6006095 Chr.22: 29328803-29328803 on Build GRCh38
rs960494799 Chr.22: 29328592-29328592 on Build GRCh38
rs778791073 Chr.22: 29328783-29328783 on Build GRCh38
rs902911111 Chr.22: 29328502-29328502 on Build GRCh38
rs374011821 Chr.22: 29328631-29328631 on Build GRCh38
rs1049978267 Chr.22: 29328766-29328766 on Build GRCh38
rs917138306 Chr.22: 29328742-29328742 on Build GRCh38
rs998403455 Chr.22: 29328454-29328454 on Build GRCh38
rs771803540 Chr.22: 29328817-29328817 on Build GRCh38
rs377604350 Chr.22: 29328807-29328807 on Build GRCh38
rs369090801 Chr.22: 29328787-29328787 on Build GRCh38
rs901354226 Chr.22: 29328727-29328727 on Build GRCh38
rs527713502 Chr.22: 29328804-29328804 on Build GRCh38
rs919354503 Chr.22: 29328648-29328648 on Build GRCh38
rs953685407 Chr.22: 29328564-29328564 on Build GRCh38
rs749740550 Chr.22: 29328808-29328808 on Build GRCh38
rs1041506646 Chr.22: 29328768-29328768 on Build GRCh38
rs202058803 Chr.22: 29328589-29328589 on Build GRCh38
rs553358364 Chr.22: 29328481-29328481 on Build GRCh38
rs74339735 Chr.22: 29328675-29328675 on Build GRCh38
rs200734849 Chr.22: 29328812-29328812 on Build GRCh38
rs544527259 Chr.22: 29328620-29328620 on Build GRCh38
rs916473006 Chr.22: 29328606-29328606 on Build GRCh38
rs1028493693 Chr.22: 29328476-29328476 on Build GRCh38
rs901468950 Chr.22: 29328431-29328431 on Build GRCh38
rs766482157 Chr.22: 29328778-29328778 on Build GRCh38
rs1053101712 Chr.22: 29328691-29328691 on Build GRCh38
rs926098509 Chr.22: 29328662-29328662 on Build GRCh38
rs1032618849 Chr.22: 29328549-29328550 on Build GRCh38
rs552068696 Chr.22: 29328779-29328779 on Build GRCh38
rs370329968 Chr.22: 29328815-29328815 on Build GRCh38
rs147078620 Chr.22: 29328371-29328371 on Build GRCh38
rs1056119438 Chr.22: 29328741-29328741 on Build GRCh38
rs759793610 Chr.22: 29328723-29328723 on Build GRCh38
rs745389476 Chr.22: 29328370-29328370 on Build GRCh38
rs1055666255 Chr.22: 29328453-29328453 on Build GRCh38
rs149575781 Chr.22: 29328495-29328495 on Build GRCh38
rs544853886 Chr.22: 29328465-29328465 on Build GRCh38
rs60770384 Chr.22: 29328570-29328570 on Build GRCh38
rs761633895 Chr.22: 29328605-29328605 on Build GRCh38
rs999835139 Chr.22: 29328522-29328522 on Build GRCh38
rs966615357 Chr.22: 29328621-29328621 on Build GRCh38
rs773226351 Chr.22: 29328803-29328804 on Build GRCh38
rs138230280 Chr.22: 29328494-29328494 on Build GRCh38
rs999056656 Chr.22: 29328392-29328392 on Build GRCh38
rs375793856 Chr.22: 29328786-29328786 on Build GRCh38
rs909819405 Chr.22: 29328414-29328414 on Build GRCh38
rs972193114 Chr.22: 29328647-29328647 on Build GRCh38
rs199898148 Chr.22: 29328814-29328814 on Build GRCh38
rs776924079 Chr.22: 29328715-29328715 on Build GRCh38

View Full Product Details