Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41868523-41869028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SREBF2
Entrez Gene ID
6721
Gene Name
sterol regulatory element binding transcription factor 2
Gene Aliases
SREBP-2, SREBP2, bHLHd2
UniGene
Hs.443258
Gene Chromosome Location
Chr.22: 41833079-41907308 on Build GRCh38
Transcripts
NM_004599.3 NR_103834.1 XM_006724310.2 XM_011530347.1 XM_017028921.1 XM_017028922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6790158 c.591C>G p.V197V Chr.22: 41868663-41868663 on Build GRCh38
6570734 c.666C>T p.T222T Chr.22: 41868738-41868738 on Build GRCh38
6243106 c.626A>C p.Q209P Chr.22: 41868698-41868698 on Build GRCh38
1416567 c.695C>A p.A232D Chr.22: 41868767-41868767 on Build GRCh38
3939664 c.698C>T p.A233V Chr.22: 41868770-41868770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761722062 Chr.22: 41868830-41868830 on Build GRCh38
rs770444190 Chr.22: 41868782-41868782 on Build GRCh38
rs772417795 Chr.22: 41868659-41868659 on Build GRCh38
rs369833717 Chr.22: 41868862-41868862 on Build GRCh38
rs904779271 Chr.22: 41868604-41868604 on Build GRCh38
rs904726775 Chr.22: 41868925-41868925 on Build GRCh38
rs183061752 Chr.22: 41868938-41868938 on Build GRCh38
rs1047046944 Chr.22: 41868630-41868630 on Build GRCh38
rs1003080729 Chr.22: 41868988-41868988 on Build GRCh38
rs746502764 Chr.22: 41868940-41868940 on Build GRCh38
rs1014264644 Chr.22: 41868941-41868941 on Build GRCh38
rs765847348 Chr.22: 41868698-41868698 on Build GRCh38
rs757396090 Chr.22: 41868637-41868637 on Build GRCh38
rs943949546 Chr.22: 41868890-41868890 on Build GRCh38
rs873638 Chr.22: 41868771-41868771 on Build GRCh38
rs776085622 Chr.22: 41868660-41868660 on Build GRCh38
rs772838144 Chr.22: 41868691-41868691 on Build GRCh38
rs201154921 Chr.22: 41868748-41868748 on Build GRCh38
rs536440370 Chr.22: 41869002-41869002 on Build GRCh38
rs1001721330 Chr.22: 41868930-41868930 on Build GRCh38
rs201991869 Chr.22: 41868728-41868728 on Build GRCh38
rs376619164 Chr.22: 41868716-41868716 on Build GRCh38
rs781479095 Chr.22: 41868753-41868776 on Build GRCh38
rs1021800599 Chr.22: 41868601-41868601 on Build GRCh38
rs755990008 Chr.22: 41868598-41868598 on Build GRCh38
rs377730162 Chr.22: 41868752-41868752 on Build GRCh38
rs1020420775 Chr.22: 41868971-41868971 on Build GRCh38
rs780961761 Chr.22: 41868951-41868951 on Build GRCh38
rs1035869290 Chr.22: 41868993-41868993 on Build GRCh38
rs773697152 Chr.22: 41868700-41868700 on Build GRCh38
rs760740664 Chr.22: 41868805-41868805 on Build GRCh38
rs994956639 Chr.22: 41868990-41868990 on Build GRCh38
rs556273310 Chr.22: 41868914-41868914 on Build GRCh38
rs746352873 Chr.22: 41868652-41868652 on Build GRCh38
rs903335001 Chr.22: 41868987-41868987 on Build GRCh38
rs771256668 Chr.22: 41868727-41868750 on Build GRCh38
rs887021576 Chr.22: 41868632-41868632 on Build GRCh38
rs780420863 Chr.22: 41868755-41868755 on Build GRCh38
rs1052587024 Chr.22: 41868827-41868827 on Build GRCh38
rs199832308 Chr.22: 41868722-41868722 on Build GRCh38
rs144099865 Chr.22: 41868785-41868785 on Build GRCh38
rs757592583 Chr.22: 41868729-41868729 on Build GRCh38
rs747276003 Chr.22: 41868564-41868565 on Build GRCh38
rs537912682 Chr.22: 41868891-41868891 on Build GRCh38
rs762644128 Chr.22: 41868693-41868693 on Build GRCh38
rs778069842 Chr.22: 41868600-41868600 on Build GRCh38
rs761908810 Chr.22: 41868717-41868717 on Build GRCh38
rs753644830 Chr.22: 41868816-41868816 on Build GRCh38
rs200709248 Chr.22: 41868747-41868747 on Build GRCh38
rs868058007 Chr.22: 41868718-41868718 on Build GRCh38
rs931126189 Chr.22: 41868619-41868619 on Build GRCh38
rs1034299504 Chr.22: 41868707-41868707 on Build GRCh38
rs144598295 Chr.22: 41868760-41868760 on Build GRCh38
rs374600446 Chr.22: 41868796-41868796 on Build GRCh38
rs748546477 Chr.22: 41868781-41868781 on Build GRCh38
rs745955887 Chr.22: 41868814-41868815 on Build GRCh38
rs944408091 Chr.22: 41868820-41868820 on Build GRCh38
rs982593284 Chr.22: 41868808-41868808 on Build GRCh38
rs1001773552 Chr.22: 41868605-41868605 on Build GRCh38
rs369398560 Chr.22: 41868587-41868587 on Build GRCh38
rs763947707 Chr.22: 41868814-41868814 on Build GRCh38
rs200294282 Chr.22: 41868604-41868604 on Build GRCh38
rs758908531 Chr.22: 41868789-41868789 on Build GRCh38
rs368708710 Chr.22: 41868835-41868835 on Build GRCh38
rs768987716 Chr.22: 41868770-41868770 on Build GRCh38
rs376909961 Chr.22: 41868725-41868725 on Build GRCh38
rs149336069 Chr.22: 41868648-41868648 on Build GRCh38
rs754914641 Chr.22: 41868584-41868584 on Build GRCh38
rs146478432 Chr.22: 41868792-41868792 on Build GRCh38
rs749584967 Chr.22: 41868615-41868615 on Build GRCh38
rs5758497 Chr.22: 41868904-41868904 on Build GRCh38
rs928320169 Chr.22: 41868753-41868753 on Build GRCh38
rs747764160 Chr.22: 41868960-41868960 on Build GRCh38
rs747315727 Chr.22: 41868665-41868665 on Build GRCh38
rs768481501 Chr.22: 41868835-41868835 on Build GRCh38
rs372943021 Chr.22: 41868588-41868588 on Build GRCh38
rs1040386293 Chr.22: 41868667-41868667 on Build GRCh38

View Full Product Details