Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 99850409-99850915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGL
Entrez Gene ID
178
Gene Name
amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase
Gene Aliases
GDE
UniGene
Hs.904
Gene Chromosome Location
Chr.1: 99850077-99924023 on Build GRCh38
Transcripts
NM_000028.2 NM_000642.2 NM_000643.2 NM_000644.2 XM_005270557.2

SNP Detail

SNP ID Chromosome Location
rs1049763653 Chr.1: 99850788-99850788 on Build GRCh38
rs1037419108 Chr.1: 99850438-99850438 on Build GRCh38
rs998615584 Chr.1: 99850445-99850445 on Build GRCh38
rs909738818 Chr.1: 99850705-99850705 on Build GRCh38
rs764383829 Chr.1: 99850445-99850445 on Build GRCh38
rs568255289 Chr.1: 99850805-99850805 on Build GRCh38
rs983881209 Chr.1: 99850696-99850696 on Build GRCh38
rs1054599524 Chr.1: 99850885-99850885 on Build GRCh38
rs1046476870 Chr.1: 99850501-99850501 on Build GRCh38
rs1021151265 Chr.1: 99850563-99850563 on Build GRCh38
rs901189755 Chr.1: 99850442-99850442 on Build GRCh38
rs970401887 Chr.1: 99850598-99850598 on Build GRCh38
rs1000329504 Chr.1: 99850520-99850520 on Build GRCh38
rs982224973 Chr.1: 99850609-99850609 on Build GRCh38
rs1056104580 Chr.1: 99850464-99850464 on Build GRCh38
rs987920790 Chr.1: 99850562-99850562 on Build GRCh38
rs540388561 Chr.1: 99850448-99850449 on Build GRCh38
rs530809457 Chr.1: 99850526-99850526 on Build GRCh38
rs552648572 Chr.1: 99850433-99850433 on Build GRCh38
rs568150943 Chr.1: 99850468-99850468 on Build GRCh38
rs570733643 Chr.1: 99850626-99850626 on Build GRCh38
rs142331734 Chr.1: 99850856-99850857 on Build GRCh38
rs528149531 Chr.1: 99850808-99850810 on Build GRCh38
rs948045826 Chr.1: 99850469-99850469 on Build GRCh38
rs1037108652 Chr.1: 99850758-99850758 on Build GRCh38
rs888594727 Chr.1: 99850678-99850678 on Build GRCh38
rs557247196 Chr.1: 99850840-99850840 on Build GRCh38
rs940072388 Chr.1: 99850737-99850737 on Build GRCh38
rs933996683 Chr.1: 99850785-99850785 on Build GRCh38
rs371657154 Chr.1: 99850596-99850596 on Build GRCh38
rs998227893 Chr.1: 99850786-99850786 on Build GRCh38
rs374515839 Chr.1: 99850780-99850780 on Build GRCh38
rs890001672 Chr.1: 99850803-99850803 on Build GRCh38
rs535096205 Chr.1: 99850472-99850472 on Build GRCh38
rs777278669 Chr.1: 99850434-99850434 on Build GRCh38
rs758360729 Chr.1: 99850730-99850731 on Build GRCh38
rs550641188 Chr.1: 99850542-99850542 on Build GRCh38
rs937938988 Chr.1: 99850644-99850644 on Build GRCh38
rs377176234 Chr.1: 99850756-99850756 on Build GRCh38
rs548120281 Chr.1: 99850706-99850706 on Build GRCh38
rs759341394 Chr.1: 99850670-99850670 on Build GRCh38
rs957750361 Chr.1: 99850540-99850540 on Build GRCh38
rs566013647 Chr.1: 99850448-99850448 on Build GRCh38
rs967078774 Chr.1: 99850853-99850853 on Build GRCh38
rs537393462 Chr.1: 99850806-99850806 on Build GRCh38
rs926534466 Chr.1: 99850633-99850633 on Build GRCh38
rs906575280 Chr.1: 99850522-99850522 on Build GRCh38
rs994349735 Chr.1: 99850527-99850527 on Build GRCh38
rs935945280 Chr.1: 99850458-99850458 on Build GRCh38

View Full Product Details