Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 35640880-35641412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
UniGene
Hs.475902
Gene Chromosome Location
Chr.3: 35638981-35798933 on Build GRCh38
Transcripts
NM_001267616.1 NM_001267617.1 XM_011533299.2 XM_011533303.2 XM_017005576.1 XM_017005578.1 XM_017005579.1 XM_017005580.1 XM_017005582.1 XM_017005584.1 XM_017005585.1 XM_017005588.1 XM_017005591.1 XM_017005592.1 XM_017005595.1 XM_017005596.1 XM_017005597.1 XM_017005598.1 XM_017005599.1 XM_017005602.1 XM_017005605.1 XM_017005607.1 XM_017005608.1 XM_017005609.1 XM_017005610.1 XM_017005611.1 XM_017005612.1

SNP Detail

SNP ID Chromosome Location
rs917583909 Chr.3: 35641375-35641375 on Build GRCh38
rs1041865381 Chr.3: 35641030-35641030 on Build GRCh38
rs959043850 Chr.3: 35641027-35641027 on Build GRCh38
rs7625602 Chr.3: 35641182-35641182 on Build GRCh38
rs9821328 Chr.3: 35640918-35640918 on Build GRCh38
rs1012414122 Chr.3: 35641142-35641142 on Build GRCh38
rs1024682398 Chr.3: 35641380-35641380 on Build GRCh38
rs902123881 Chr.3: 35640914-35640914 on Build GRCh38
rs181166900 Chr.3: 35641330-35641330 on Build GRCh38
rs151319662 Chr.3: 35641202-35641206 on Build GRCh38
rs575141077 Chr.3: 35640992-35640992 on Build GRCh38
rs887517641 Chr.3: 35641173-35641173 on Build GRCh38
rs67051929 Chr.3: 35641256-35641258 on Build GRCh38
rs914700811 Chr.3: 35641269-35641269 on Build GRCh38
rs181561655 Chr.3: 35640908-35640908 on Build GRCh38
rs7637374 Chr.3: 35641267-35641267 on Build GRCh38
rs1019630352 Chr.3: 35640906-35640906 on Build GRCh38
rs919768906 Chr.3: 35641124-35641124 on Build GRCh38
rs573186828 Chr.3: 35641045-35641045 on Build GRCh38
rs542402022 Chr.3: 35641047-35641047 on Build GRCh38
rs758215604 Chr.3: 35641376-35641376 on Build GRCh38
rs373809595 Chr.3: 35641179-35641179 on Build GRCh38
rs561879316 Chr.3: 35641098-35641098 on Build GRCh38
rs1033441215 Chr.3: 35641307-35641307 on Build GRCh38
rs973054202 Chr.3: 35641111-35641111 on Build GRCh38
rs191863085 Chr.3: 35641105-35641105 on Build GRCh38
rs1009853373 Chr.3: 35641221-35641222 on Build GRCh38
rs961985271 Chr.3: 35641207-35641207 on Build GRCh38
rs904023731 Chr.3: 35641257-35641257 on Build GRCh38
rs778865061 Chr.3: 35641335-35641335 on Build GRCh38
rs765038873 Chr.3: 35641366-35641366 on Build GRCh38
rs923253874 Chr.3: 35640913-35640913 on Build GRCh38
rs552098877 Chr.3: 35641264-35641264 on Build GRCh38
rs763228187 Chr.3: 35641028-35641029 on Build GRCh38
rs538882597 Chr.3: 35640920-35640920 on Build GRCh38
rs1006094016 Chr.3: 35641180-35641180 on Build GRCh38
rs750524399 Chr.3: 35641075-35641075 on Build GRCh38
rs914732424 Chr.3: 35640991-35640991 on Build GRCh38
rs370981731 Chr.3: 35641156-35641156 on Build GRCh38
rs947612953 Chr.3: 35641001-35641001 on Build GRCh38
rs986193090 Chr.3: 35640953-35640953 on Build GRCh38
rs938922745 Chr.3: 35641056-35641056 on Build GRCh38
rs563963996 Chr.3: 35641276-35641276 on Build GRCh38
rs185933140 Chr.3: 35640950-35640950 on Build GRCh38
rs1030339380 Chr.3: 35641220-35641220 on Build GRCh38
rs752469999 Chr.3: 35641374-35641374 on Build GRCh38
rs938953848 Chr.3: 35641370-35641370 on Build GRCh38
rs917560717 Chr.3: 35641059-35641059 on Build GRCh38
rs977586567 Chr.3: 35641343-35641343 on Build GRCh38
rs986529173 Chr.3: 35641265-35641265 on Build GRCh38
rs534632887 Chr.3: 35640905-35640905 on Build GRCh38
rs533461040 Chr.3: 35641216-35641216 on Build GRCh38
rs541088917 Chr.3: 35641139-35641139 on Build GRCh38
rs953371562 Chr.3: 35641262-35641262 on Build GRCh38
rs968199058 Chr.3: 35640959-35640959 on Build GRCh38
rs1033388789 Chr.3: 35640998-35640998 on Build GRCh38
rs924751770 Chr.3: 35641367-35641367 on Build GRCh38

View Full Product Details