Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47235943-47236451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF9
Entrez Gene ID
64147
Gene Name
kinesin family member 9
Gene Aliases
-
UniGene
Hs.373947
Gene Chromosome Location
Chr.3: 47228026-47282847 on Build GRCh38
Transcripts
NM_001134878.1 NM_022342.4 NM_182902.3 XM_005265388.4 XM_005265389.4 XM_006713291.3 XM_011534003.2 XM_011534004.2 XM_011534005.2 XM_011534006.2 XM_011534007.2 XM_017007029.1 XM_017007030.1 XM_017007031.1 NR_033373.1

Gene Symbol
KIF9-AS1
Entrez Gene ID
285352
Gene Name
KIF9 antisense RNA 1
Gene Aliases
-
UniGene
Hs.570631
Gene Chromosome Location
Chr.3: 47164370-47244116 on Build GRCh38
Transcripts
NM_001134878.1 NM_022342.4 NM_182902.3 XM_005265388.4 XM_005265389.4 XM_006713291.3 XM_011534003.2 XM_011534004.2 XM_011534005.2 XM_011534006.2 XM_011534007.2 XM_017007029.1 XM_017007030.1 XM_017007031.1 NR_033373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5864648 c.2100G>A p.M700I Chr.3: 47236444-47236444 on Build GRCh38
4118026 c.2149C>T p.Q717* Chr.3: 47236102-47236102 on Build GRCh38
6337208 c.2148G>T p.M716I Chr.3: 47236103-47236103 on Build GRCh38
4754438 c.2198T>C p.V733A Chr.3: 47236053-47236053 on Build GRCh38
4534998 c.2143G>A p.D715N Chr.3: 47236108-47236108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775606557 Chr.3: 47236157-47236161 on Build GRCh38
rs538488140 Chr.3: 47236229-47236229 on Build GRCh38
rs769281394 Chr.3: 47236087-47236087 on Build GRCh38
rs560394239 Chr.3: 47236071-47236071 on Build GRCh38
rs747973917 Chr.3: 47236063-47236063 on Build GRCh38
rs770148203 Chr.3: 47236418-47236418 on Build GRCh38
rs779121891 Chr.3: 47236072-47236072 on Build GRCh38
rs769301658 Chr.3: 47236404-47236404 on Build GRCh38
rs754889739 Chr.3: 47236058-47236058 on Build GRCh38
rs780348525 Chr.3: 47236388-47236388 on Build GRCh38
rs777104045 Chr.3: 47236026-47236026 on Build GRCh38
rs1046063487 Chr.3: 47236165-47236165 on Build GRCh38
rs758661231 Chr.3: 47236378-47236378 on Build GRCh38
rs776096807 Chr.3: 47236016-47236016 on Build GRCh38
rs777991827 Chr.3: 47236376-47236376 on Build GRCh38
rs563961502 Chr.3: 47235983-47235983 on Build GRCh38
rs2291722 Chr.3: 47236028-47236028 on Build GRCh38
rs748494124 Chr.3: 47236417-47236417 on Build GRCh38
rs141894213 Chr.3: 47236185-47236185 on Build GRCh38
rs766336874 Chr.3: 47236049-47236049 on Build GRCh38
rs752434428 Chr.3: 47236194-47236194 on Build GRCh38
rs761141751 Chr.3: 47236018-47236018 on Build GRCh38
rs777193969 Chr.3: 47236105-47236105 on Build GRCh38
rs781008594 Chr.3: 47236062-47236062 on Build GRCh38
rs567961277 Chr.3: 47236215-47236215 on Build GRCh38
rs779023088 Chr.3: 47236199-47236199 on Build GRCh38
rs755895487 Chr.3: 47236196-47236196 on Build GRCh38
rs377004871 Chr.3: 47236042-47236043 on Build GRCh38
rs557076310 Chr.3: 47236238-47236238 on Build GRCh38
rs35043243 Chr.3: 47236090-47236091 on Build GRCh38
rs369159848 Chr.3: 47235990-47235990 on Build GRCh38
rs767381797 Chr.3: 47236193-47236193 on Build GRCh38
rs961583003 Chr.3: 47236302-47236302 on Build GRCh38
rs562470963 Chr.3: 47236055-47236055 on Build GRCh38
rs745898998 Chr.3: 47236002-47236002 on Build GRCh38
rs760741632 Chr.3: 47236386-47236386 on Build GRCh38
rs1028404154 Chr.3: 47236218-47236218 on Build GRCh38
rs781675406 Chr.3: 47236416-47236416 on Build GRCh38
rs750705193 Chr.3: 47236201-47236201 on Build GRCh38
rs1054236550 Chr.3: 47235973-47235973 on Build GRCh38
rs1019835078 Chr.3: 47236269-47236269 on Build GRCh38
rs997399941 Chr.3: 47236064-47236064 on Build GRCh38
rs532910408 Chr.3: 47236081-47236081 on Build GRCh38
rs865837586 Chr.3: 47236118-47236118 on Build GRCh38
rs1031486302 Chr.3: 47236213-47236214 on Build GRCh38
rs768018243 Chr.3: 47236355-47236355 on Build GRCh38
rs141091505 Chr.3: 47235986-47235986 on Build GRCh38
rs764804389 Chr.3: 47236158-47236158 on Build GRCh38
rs549743218 Chr.3: 47236186-47236186 on Build GRCh38
rs914368874 Chr.3: 47236327-47236327 on Build GRCh38
rs762180414 Chr.3: 47236140-47236140 on Build GRCh38
rs201278437 Chr.3: 47236027-47236027 on Build GRCh38
rs1008192812 Chr.3: 47236267-47236267 on Build GRCh38
rs1874264 Chr.3: 47235968-47235968 on Build GRCh38
rs945792926 Chr.3: 47236346-47236346 on Build GRCh38
rs926959595 Chr.3: 47235967-47235967 on Build GRCh38
rs116326157 Chr.3: 47236038-47236038 on Build GRCh38
rs13094438 Chr.3: 47236395-47236395 on Build GRCh38
rs765925634 Chr.3: 47236166-47236166 on Build GRCh38
rs199770331 Chr.3: 47236023-47236023 on Build GRCh38
rs375567605 Chr.3: 47236160-47236160 on Build GRCh38
rs115270319 Chr.3: 47236144-47236144 on Build GRCh38
rs773429405 Chr.3: 47236419-47236419 on Build GRCh38
rs958429162 Chr.3: 47236230-47236230 on Build GRCh38
rs540169750 Chr.3: 47236007-47236007 on Build GRCh38
rs745962914 Chr.3: 47236076-47236076 on Build GRCh38
rs114811664 Chr.3: 47236082-47236082 on Build GRCh38
rs990299976 Chr.3: 47236280-47236280 on Build GRCh38
rs963199722 Chr.3: 47236095-47236095 on Build GRCh38

View Full Product Details