Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50617332-50617832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPKAPK3
Entrez Gene ID
7867
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
Gene Aliases
3PK, MAPKAP-K3, MAPKAP3, MAPKAPK-3, MK-3
UniGene
Hs.234521
Gene Chromosome Location
Chr.3: 50611862-50649297 on Build GRCh38
Transcripts
NM_001243925.1 NM_001243926.1 NM_004635.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6544445 c.67G>T p.G23C Chr.3: 50617632-50617632 on Build GRCh38
2851523 c.148C>A p.L50M Chr.3: 50617713-50617713 on Build GRCh38
6387858 c.202C>A p.Q68K Chr.3: 50617767-50617767 on Build GRCh38
7141569 c.162G>T p.V54V Chr.3: 50617727-50617727 on Build GRCh38
6446943 c.219+2T>G p.? Chr.3: 50617786-50617786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147044454 Chr.3: 50617596-50617596 on Build GRCh38
rs761898324 Chr.3: 50617757-50617757 on Build GRCh38
rs200053491 Chr.3: 50617542-50617542 on Build GRCh38
rs895182858 Chr.3: 50617619-50617619 on Build GRCh38
rs777789251 Chr.3: 50617597-50617597 on Build GRCh38
rs78528472 Chr.3: 50617689-50617689 on Build GRCh38
rs764298483 Chr.3: 50617743-50617743 on Build GRCh38
rs752802256 Chr.3: 50617595-50617595 on Build GRCh38
rs757254886 Chr.3: 50617655-50617655 on Build GRCh38
rs758219473 Chr.3: 50617679-50617679 on Build GRCh38
rs754733216 Chr.3: 50617812-50617812 on Build GRCh38
rs778722098 Chr.3: 50617612-50617612 on Build GRCh38
rs1010981482 Chr.3: 50617355-50617355 on Build GRCh38
rs759919624 Chr.3: 50617723-50617723 on Build GRCh38
rs554058062 Chr.3: 50617411-50617411 on Build GRCh38
rs993161647 Chr.3: 50617501-50617501 on Build GRCh38
rs751967432 Chr.3: 50617648-50617649 on Build GRCh38
rs202232905 Chr.3: 50617549-50617549 on Build GRCh38
rs1027772520 Chr.3: 50617421-50617421 on Build GRCh38
rs766895982 Chr.3: 50617575-50617575 on Build GRCh38
rs770742107 Chr.3: 50617540-50617540 on Build GRCh38
rs768388741 Chr.3: 50617685-50617685 on Build GRCh38
rs893676261 Chr.3: 50617350-50617350 on Build GRCh38
rs759957245 Chr.3: 50617648-50617648 on Build GRCh38
rs755218440 Chr.3: 50617586-50617586 on Build GRCh38
rs777699096 Chr.3: 50617516-50617516 on Build GRCh38
rs745672353 Chr.3: 50617618-50617618 on Build GRCh38
rs763591183 Chr.3: 50617637-50617637 on Build GRCh38
rs368714447 Chr.3: 50617669-50617669 on Build GRCh38
rs955808780 Chr.3: 50617591-50617591 on Build GRCh38
rs760997594 Chr.3: 50617740-50617740 on Build GRCh38
rs749156678 Chr.3: 50617517-50617517 on Build GRCh38
rs368742045 Chr.3: 50617572-50617572 on Build GRCh38
rs751303152 Chr.3: 50617794-50617794 on Build GRCh38
rs376663700 Chr.3: 50617546-50617546 on Build GRCh38
rs779935731 Chr.3: 50617792-50617792 on Build GRCh38
rs774924055 Chr.3: 50617719-50617719 on Build GRCh38
rs983818889 Chr.3: 50617481-50617481 on Build GRCh38
rs1027886642 Chr.3: 50617654-50617654 on Build GRCh38
rs1003925928 Chr.3: 50617374-50617374 on Build GRCh38
rs753977197 Chr.3: 50617746-50617746 on Build GRCh38
rs959954847 Chr.3: 50617430-50617430 on Build GRCh38
rs1012492737 Chr.3: 50617407-50617407 on Build GRCh38
rs372994175 Chr.3: 50617590-50617590 on Build GRCh38
rs779772290 Chr.3: 50617620-50617620 on Build GRCh38
rs149349769 Chr.3: 50617594-50617594 on Build GRCh38
rs760222732 Chr.3: 50617552-50617552 on Build GRCh38
rs763457916 Chr.3: 50617562-50617562 on Build GRCh38
rs758222981 Chr.3: 50617771-50617771 on Build GRCh38
rs375412266 Chr.3: 50617647-50617647 on Build GRCh38
rs373642329 Chr.3: 50617650-50617650 on Build GRCh38
rs1041696500 Chr.3: 50617671-50617671 on Build GRCh38
rs902614577 Chr.3: 50617370-50617370 on Build GRCh38
rs779678577 Chr.3: 50617681-50617681 on Build GRCh38
rs746700451 Chr.3: 50617684-50617684 on Build GRCh38
rs751937158 Chr.3: 50617579-50617579 on Build GRCh38
rs980812822 Chr.3: 50617554-50617554 on Build GRCh38
rs372238062 Chr.3: 50617686-50617686 on Build GRCh38
rs753884487 Chr.3: 50617652-50617652 on Build GRCh38
rs376982083 Chr.3: 50617622-50617622 on Build GRCh38
rs776254646 Chr.3: 50617627-50617627 on Build GRCh38
rs1042518952 Chr.3: 50617356-50617356 on Build GRCh38
rs757165001 Chr.3: 50617598-50617598 on Build GRCh38
rs1042257886 Chr.3: 50617609-50617609 on Build GRCh38
rs35362731 Chr.3: 50617759-50617759 on Build GRCh38
rs771726192 Chr.3: 50617548-50617548 on Build GRCh38
rs763518219 Chr.3: 50617573-50617573 on Build GRCh38
rs768248070 Chr.3: 50617626-50617626 on Build GRCh38
rs764208748 Chr.3: 50617651-50617651 on Build GRCh38
rs545277836 Chr.3: 50617401-50617401 on Build GRCh38
rs577923066 Chr.3: 50617398-50617398 on Build GRCh38
rs200457380 Chr.3: 50617727-50617727 on Build GRCh38
rs1015738152 Chr.3: 50617500-50617500 on Build GRCh38
rs750410127 Chr.3: 50617761-50617761 on Build GRCh38
rs201487546 Chr.3: 50617640-50617640 on Build GRCh38
rs976782996 Chr.3: 50617533-50617533 on Build GRCh38
rs1004071976 Chr.3: 50617397-50617397 on Build GRCh38

View Full Product Details