Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108929427-108929935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCC1
Entrez Gene ID
23155
Gene Name
chloride channel CLIC like 1
Gene Aliases
MCLC
UniGene
Hs.658489
Gene Chromosome Location
Chr.1: 108929508-108963499 on Build GRCh38
Transcripts
NM_001048210.2 NM_001278202.1 NM_001278203.1 NM_015127.4 NM_001321038.1 NM_001321039.1 NM_013296.4 XM_006710589.2 XM_011541301.2 XM_011541302.2 XM_017001097.1 XM_017001098.1

Gene Symbol
GPSM2
Entrez Gene ID
29899
Gene Name
G-protein signaling modulator 2
Gene Aliases
CMCS, DFNB82, LGN, PINS
UniGene
Hs.584901
Gene Chromosome Location
Chr.1: 108875350-108934335 on Build GRCh38
Transcripts
NM_001048210.2 NM_001278202.1 NM_001278203.1 NM_015127.4 NM_001321038.1 NM_001321039.1 NM_013296.4 XM_006710589.2 XM_011541301.2 XM_011541302.2 XM_017001097.1 XM_017001098.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7089535 c.2032G>T p.G678W Chr.1: 108929917-108929917 on Build GRCh38
7132647 c.1925G>A p.R642K Chr.1: 108929810-108929810 on Build GRCh38
7132648 c.1955G>A p.R652K Chr.1: 108929840-108929840 on Build GRCh38
5137355 c.1909C>T p.R637W Chr.1: 108929794-108929794 on Build GRCh38
6298924 c.2013G>T p.L671F Chr.1: 108929898-108929898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144830055 Chr.1: 108929783-108929783 on Build GRCh38
rs899425466 Chr.1: 108929572-108929572 on Build GRCh38
rs761682246 Chr.1: 108929559-108929559 on Build GRCh38
rs140026774 Chr.1: 108929851-108929851 on Build GRCh38
rs909839111 Chr.1: 108929543-108929543 on Build GRCh38
rs765367400 Chr.1: 108929795-108929795 on Build GRCh38
rs757251927 Chr.1: 108929681-108929682 on Build GRCh38
rs777636994 Chr.1: 108929655-108929656 on Build GRCh38
rs1037062017 Chr.1: 108929614-108929614 on Build GRCh38
rs1031318340 Chr.1: 108929767-108929767 on Build GRCh38
rs199555600 Chr.1: 108929673-108929673 on Build GRCh38
rs776223239 Chr.1: 108929473-108929473 on Build GRCh38
rs574565748 Chr.1: 108929895-108929895 on Build GRCh38
rs939433123 Chr.1: 108929629-108929629 on Build GRCh38
rs752155776 Chr.1: 108929664-108929664 on Build GRCh38
rs774671620 Chr.1: 108929870-108929870 on Build GRCh38
rs768924499 Chr.1: 108929868-108929868 on Build GRCh38
rs993741191 Chr.1: 108929574-108929574 on Build GRCh38
rs748946684 Chr.1: 108929687-108929687 on Build GRCh38
rs35452234 Chr.1: 108929797-108929797 on Build GRCh38
rs201803860 Chr.1: 108929704-108929704 on Build GRCh38
rs745995915 Chr.1: 108929698-108929699 on Build GRCh38
rs35322815 Chr.1: 108929705-108929705 on Build GRCh38
rs750820277 Chr.1: 108929672-108929672 on Build GRCh38
rs1014388324 Chr.1: 108929631-108929631 on Build GRCh38
rs985783540 Chr.1: 108929520-108929520 on Build GRCh38
rs762004200 Chr.1: 108929889-108929889 on Build GRCh38
rs763208536 Chr.1: 108929755-108929755 on Build GRCh38
rs1026921995 Chr.1: 108929613-108929613 on Build GRCh38
rs189033496 Chr.1: 108929794-108929794 on Build GRCh38
rs910307815 Chr.1: 108929824-108929824 on Build GRCh38
rs753527959 Chr.1: 108929675-108929675 on Build GRCh38
rs757866175 Chr.1: 108929817-108929817 on Build GRCh38
rs987223472 Chr.1: 108929799-108929799 on Build GRCh38
rs538348012 Chr.1: 108929606-108929606 on Build GRCh38
rs754645910 Chr.1: 108929804-108929804 on Build GRCh38
rs779594326 Chr.1: 108929685-108929685 on Build GRCh38
rs772011195 Chr.1: 108929715-108929715 on Build GRCh38
rs746839772 Chr.1: 108929735-108929735 on Build GRCh38
rs776485420 Chr.1: 108929865-108929865 on Build GRCh38
rs114558565 Chr.1: 108929756-108929756 on Build GRCh38
rs761238838 Chr.1: 108929906-108929906 on Build GRCh38
rs897224783 Chr.1: 108929620-108929620 on Build GRCh38
rs778687676 Chr.1: 108929695-108929698 on Build GRCh38
rs138132875 Chr.1: 108929693-108929693 on Build GRCh38
rs772268313 Chr.1: 108929878-108929878 on Build GRCh38
rs146291181 Chr.1: 108929670-108929670 on Build GRCh38
rs745587821 Chr.1: 108929867-108929867 on Build GRCh38
rs772254021 Chr.1: 108929891-108929891 on Build GRCh38
rs1037292949 Chr.1: 108929570-108929570 on Build GRCh38
rs766711999 Chr.1: 108929912-108929912 on Build GRCh38
rs727503087 Chr.1: 108929761-108929761 on Build GRCh38
rs775626164 Chr.1: 108929744-108929744 on Build GRCh38
rs774583431 Chr.1: 108929759-108929759 on Build GRCh38
rs548463870 Chr.1: 108929479-108929479 on Build GRCh38
rs757821868 Chr.1: 108929668-108929668 on Build GRCh38
rs770712392 Chr.1: 108929743-108929743 on Build GRCh38
rs778480184 Chr.1: 108929695-108929695 on Build GRCh38
rs777713306 Chr.1: 108929841-108929841 on Build GRCh38
rs200841293 Chr.1: 108929805-108929805 on Build GRCh38
rs1012458908 Chr.1: 108929701-108929701 on Build GRCh38
rs554890802 Chr.1: 108929815-108929815 on Build GRCh38
rs985344827 Chr.1: 108929903-108929903 on Build GRCh38
rs773069843 Chr.1: 108929726-108929726 on Build GRCh38
rs764215938 Chr.1: 108929659-108929659 on Build GRCh38
rs370742816 Chr.1: 108929857-108929857 on Build GRCh38

View Full Product Details