Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109582311-109582826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAI3
Entrez Gene ID
2773
Gene Name
G protein subunit alpha i3
Gene Aliases
87U6, ARCND1
UniGene
Hs.73799
Gene Chromosome Location
Chr.1: 109548564-109595843 on Build GRCh38
Transcripts
NM_006496.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687392 c.547G>T p.G183C Chr.1: 109582522-109582522 on Build GRCh38
893537 c.491A>C p.Q164P Chr.1: 109582466-109582466 on Build GRCh38
1332470 c.492G>A p.Q164Q Chr.1: 109582467-109582467 on Build GRCh38
237454 c.564T>C p.H188H Chr.1: 109582539-109582539 on Build GRCh38
225438 c.488C>T p.S163F Chr.1: 109582463-109582463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577957303 Chr.1: 109582710-109582710 on Build GRCh38
rs747711646 Chr.1: 109582599-109582599 on Build GRCh38
rs747792423 Chr.1: 109582455-109582455 on Build GRCh38
rs755968072 Chr.1: 109582576-109582576 on Build GRCh38
rs923305475 Chr.1: 109582336-109582336 on Build GRCh38
rs767803865 Chr.1: 109582552-109582552 on Build GRCh38
rs757055872 Chr.1: 109582399-109582399 on Build GRCh38
rs993932612 Chr.1: 109582492-109582492 on Build GRCh38
rs72987036 Chr.1: 109582763-109582763 on Build GRCh38
rs971607557 Chr.1: 109582644-109582644 on Build GRCh38
rs769686023 Chr.1: 109582734-109582734 on Build GRCh38
rs188039134 Chr.1: 109582749-109582749 on Build GRCh38
rs933352665 Chr.1: 109582343-109582343 on Build GRCh38
rs776209584 Chr.1: 109582503-109582503 on Build GRCh38
rs758560176 Chr.1: 109582387-109582387 on Build GRCh38
rs778242381 Chr.1: 109582586-109582586 on Build GRCh38
rs745708992 Chr.1: 109582409-109582409 on Build GRCh38
rs540704858 Chr.1: 109582735-109582736 on Build GRCh38
rs11558723 Chr.1: 109582476-109582476 on Build GRCh38
rs942302678 Chr.1: 109582775-109582775 on Build GRCh38
rs1034548471 Chr.1: 109582713-109582713 on Build GRCh38
rs200850070 Chr.1: 109582615-109582615 on Build GRCh38
rs957756865 Chr.1: 109582721-109582721 on Build GRCh38
rs796330658 Chr.1: 109582778-109582778 on Build GRCh38
rs757382503 Chr.1: 109582545-109582545 on Build GRCh38
rs1037845211 Chr.1: 109582463-109582463 on Build GRCh38
rs773969643 Chr.1: 109582454-109582454 on Build GRCh38
rs771785264 Chr.1: 109582460-109582460 on Build GRCh38
rs112551683 Chr.1: 109582422-109582422 on Build GRCh38
rs759955097 Chr.1: 109582497-109582497 on Build GRCh38
rs762506201 Chr.1: 109582566-109582566 on Build GRCh38
rs763113801 Chr.1: 109582505-109582505 on Build GRCh38
rs545188268 Chr.1: 109582581-109582581 on Build GRCh38
rs573260668 Chr.1: 109582579-109582579 on Build GRCh38
rs765776409 Chr.1: 109582502-109582502 on Build GRCh38
rs199684276 Chr.1: 109582563-109582563 on Build GRCh38
rs780890897 Chr.1: 109582406-109582406 on Build GRCh38
rs536687404 Chr.1: 109582544-109582544 on Build GRCh38
rs564233578 Chr.1: 109582698-109582698 on Build GRCh38
rs61754626 Chr.1: 109582484-109582484 on Build GRCh38
rs755956640 Chr.1: 109582421-109582421 on Build GRCh38
rs751789822 Chr.1: 109582396-109582396 on Build GRCh38
rs987986085 Chr.1: 109582716-109582716 on Build GRCh38
rs114926114 Chr.1: 109582744-109582744 on Build GRCh38
rs1005271218 Chr.1: 109582501-109582501 on Build GRCh38
rs183382334 Chr.1: 109582393-109582393 on Build GRCh38
rs1032181350 Chr.1: 109582674-109582674 on Build GRCh38
rs1000760959 Chr.1: 109582673-109582673 on Build GRCh38
rs954800850 Chr.1: 109582747-109582747 on Build GRCh38
rs919439144 Chr.1: 109582790-109582790 on Build GRCh38
rs768212572 Chr.1: 109582430-109582430 on Build GRCh38
rs140210988 Chr.1: 109582361-109582361 on Build GRCh38
rs369731340 Chr.1: 109582565-109582565 on Build GRCh38
rs754453509 Chr.1: 109582584-109582584 on Build GRCh38
rs573944457 Chr.1: 109582506-109582506 on Build GRCh38
rs1037876402 Chr.1: 109582777-109582777 on Build GRCh38
rs942659457 Chr.1: 109582772-109582772 on Build GRCh38
rs12721541 Chr.1: 109582424-109582424 on Build GRCh38
rs773134740 Chr.1: 109582752-109582752 on Build GRCh38

View Full Product Details