Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109593496-109594006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAI3
Entrez Gene ID
2773
Gene Name
G protein subunit alpha i3
Gene Aliases
87U6, ARCND1
UniGene
Hs.73799
Gene Chromosome Location
Chr.1: 109548564-109595843 on Build GRCh38
Transcripts
NM_006496.3

SNP Detail

SNP ID Chromosome Location
rs897331948 Chr.1: 109593953-109593954 on Build GRCh38
rs977674998 Chr.1: 109593775-109593775 on Build GRCh38
rs944193349 Chr.1: 109593594-109593594 on Build GRCh38
rs965597490 Chr.1: 109593821-109593821 on Build GRCh38
rs3204436 Chr.1: 109593803-109593803 on Build GRCh38
rs940331411 Chr.1: 109593857-109593857 on Build GRCh38
rs113638529 Chr.1: 109593899-109593899 on Build GRCh38
rs532876891 Chr.1: 109593686-109593686 on Build GRCh38
rs1040161114 Chr.1: 109593601-109593601 on Build GRCh38
rs1004765195 Chr.1: 109593651-109593651 on Build GRCh38
rs867154150 Chr.1: 109593948-109593948 on Build GRCh38
rs912801854 Chr.1: 109593788-109593788 on Build GRCh38
rs546468184 Chr.1: 109593947-109593947 on Build GRCh38
rs190395214 Chr.1: 109593646-109593646 on Build GRCh38
rs3833901 Chr.1: 109593701-109593702 on Build GRCh38
rs529874646 Chr.1: 109593885-109593885 on Build GRCh38
rs975973142 Chr.1: 109593825-109593825 on Build GRCh38
rs907069497 Chr.1: 109593673-109593673 on Build GRCh38
rs1056623909 Chr.1: 109593976-109593979 on Build GRCh38
rs2301230 Chr.1: 109593836-109593836 on Build GRCh38
rs140944776 Chr.1: 109593811-109593811 on Build GRCh38
rs1048728157 Chr.1: 109593627-109593627 on Build GRCh38
rs931695202 Chr.1: 109593609-109593609 on Build GRCh38
rs373132580 Chr.1: 109593679-109593679 on Build GRCh38
rs908846155 Chr.1: 109593849-109593849 on Build GRCh38
rs921714962 Chr.1: 109593831-109593831 on Build GRCh38
rs186301843 Chr.1: 109593607-109593607 on Build GRCh38
rs575123495 Chr.1: 109593566-109593566 on Build GRCh38
rs561280016 Chr.1: 109593557-109593557 on Build GRCh38
rs181671309 Chr.1: 109593553-109593553 on Build GRCh38
rs144806197 Chr.1: 109593847-109593847 on Build GRCh38
rs989304722 Chr.1: 109593829-109593829 on Build GRCh38
rs1014775498 Chr.1: 109593671-109593671 on Build GRCh38
rs941620461 Chr.1: 109593889-109593889 on Build GRCh38
rs935155649 Chr.1: 109593801-109593801 on Build GRCh38
rs967609815 Chr.1: 109593742-109593742 on Build GRCh38
rs946275787 Chr.1: 109593549-109593549 on Build GRCh38
rs566203921 Chr.1: 109593707-109593707 on Build GRCh38
rs750829750 Chr.1: 109593633-109593633 on Build GRCh38
rs189181690 Chr.1: 109593529-109593529 on Build GRCh38
rs994304976 Chr.1: 109593965-109593966 on Build GRCh38
rs1021559351 Chr.1: 109593723-109593723 on Build GRCh38
rs1003117207 Chr.1: 109593684-109593684 on Build GRCh38
rs775106621 Chr.1: 109593984-109593984 on Build GRCh38
rs34068100 Chr.1: 109593954-109593954 on Build GRCh38
rs977560324 Chr.1: 109593560-109593560 on Build GRCh38
rs912695048 Chr.1: 109593577-109593577 on Build GRCh38
rs907098556 Chr.1: 109593970-109593970 on Build GRCh38
rs960654495 Chr.1: 109593598-109593601 on Build GRCh38
rs41280246 Chr.1: 109593694-109593694 on Build GRCh38

View Full Product Details