Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 68752623-68753136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A4
Entrez Gene ID
151647
Gene Name
family with sequence similarity 19 member A4, C-C motif chemokine like
Gene Aliases
TAFA-4, TAFA4
UniGene
Hs.187873
Gene Chromosome Location
Chr.3: 68731764-68932610 on Build GRCh38
Transcripts
NM_001005527.2 NM_182522.4 XM_011533371.1 XM_011533372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6029405 c.286+5G>T p.? Chr.3: 68752858-68752858 on Build GRCh38
3597222 c.145A>G p.K49E Chr.3: 68753004-68753004 on Build GRCh38
6098092 c.222C>G p.V74V Chr.3: 68752927-68752927 on Build GRCh38
6098091 c.204A>T p.E68D Chr.3: 68752945-68752945 on Build GRCh38
3597221 c.249G>A p.V83V Chr.3: 68752900-68752900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002526972 Chr.3: 68752707-68752707 on Build GRCh38
rs762511311 Chr.3: 68752923-68752923 on Build GRCh38
rs201545497 Chr.3: 68752827-68752827 on Build GRCh38
rs920991208 Chr.3: 68752751-68752751 on Build GRCh38
rs963267841 Chr.3: 68753019-68753019 on Build GRCh38
rs115258186 Chr.3: 68752648-68752648 on Build GRCh38
rs376072070 Chr.3: 68752799-68752799 on Build GRCh38
rs1021812096 Chr.3: 68752960-68752960 on Build GRCh38
rs774084488 Chr.3: 68752930-68752930 on Build GRCh38
rs919807607 Chr.3: 68753049-68753049 on Build GRCh38
rs766004073 Chr.3: 68752734-68752734 on Build GRCh38
rs372737877 Chr.3: 68753022-68753022 on Build GRCh38
rs780233318 Chr.3: 68752887-68752887 on Build GRCh38
rs776417855 Chr.3: 68752741-68752741 on Build GRCh38
rs764658790 Chr.3: 68753038-68753038 on Build GRCh38
rs769594198 Chr.3: 68752921-68752921 on Build GRCh38
rs146024975 Chr.3: 68752910-68752910 on Build GRCh38
rs35815841 Chr.3: 68752977-68752977 on Build GRCh38
rs775169718 Chr.3: 68753021-68753021 on Build GRCh38
rs867966396 Chr.3: 68752947-68752947 on Build GRCh38
rs752695856 Chr.3: 68753032-68753032 on Build GRCh38
rs536646432 Chr.3: 68752933-68752933 on Build GRCh38
rs368834384 Chr.3: 68752952-68752952 on Build GRCh38
rs773606490 Chr.3: 68752980-68752980 on Build GRCh38
rs35338153 Chr.3: 68752941-68752941 on Build GRCh38
rs138231744 Chr.3: 68752742-68752742 on Build GRCh38
rs765342148 Chr.3: 68753027-68753027 on Build GRCh38
rs9835691 Chr.3: 68752770-68752770 on Build GRCh38
rs199571082 Chr.3: 68753054-68753054 on Build GRCh38
rs912595562 Chr.3: 68753068-68753068 on Build GRCh38
rs974011361 Chr.3: 68753047-68753047 on Build GRCh38
rs760856756 Chr.3: 68752857-68752857 on Build GRCh38
rs768423607 Chr.3: 68752929-68752929 on Build GRCh38
rs773660779 Chr.3: 68753006-68753006 on Build GRCh38
rs775162025 Chr.3: 68752922-68752922 on Build GRCh38
rs760885189 Chr.3: 68753014-68753014 on Build GRCh38
rs571268666 Chr.3: 68752897-68752897 on Build GRCh38
rs187111065 Chr.3: 68753078-68753078 on Build GRCh38
rs149113907 Chr.3: 68752875-68752875 on Build GRCh38
rs779041263 Chr.3: 68752904-68752904 on Build GRCh38
rs148555861 Chr.3: 68752996-68752996 on Build GRCh38
rs768241072 Chr.3: 68752692-68752692 on Build GRCh38
rs1035324822 Chr.3: 68752710-68752710 on Build GRCh38
rs956230482 Chr.3: 68753083-68753083 on Build GRCh38
rs201735936 Chr.3: 68752931-68752931 on Build GRCh38
rs767988662 Chr.3: 68752932-68752932 on Build GRCh38
rs563233058 Chr.3: 68752721-68752721 on Build GRCh38
rs963537607 Chr.3: 68752787-68752787 on Build GRCh38
rs954627813 Chr.3: 68753037-68753037 on Build GRCh38
rs762198656 Chr.3: 68752873-68752873 on Build GRCh38
rs761271480 Chr.3: 68752940-68752940 on Build GRCh38
rs202244953 Chr.3: 68752867-68752867 on Build GRCh38
rs746821600 Chr.3: 68752906-68752906 on Build GRCh38
rs750992903 Chr.3: 68752878-68752878 on Build GRCh38
rs939655357 Chr.3: 68753033-68753033 on Build GRCh38
rs201536721 Chr.3: 68752991-68752991 on Build GRCh38
rs758517467 Chr.3: 68752973-68752973 on Build GRCh38
rs542438958 Chr.3: 68752992-68752992 on Build GRCh38
rs550448426 Chr.3: 68752693-68752693 on Build GRCh38
rs752000474 Chr.3: 68752858-68752858 on Build GRCh38
rs964927848 Chr.3: 68752966-68752966 on Build GRCh38
rs181032828 Chr.3: 68752746-68752746 on Build GRCh38
rs893258225 Chr.3: 68752662-68752662 on Build GRCh38
rs1031892607 Chr.3: 68752650-68752650 on Build GRCh38
rs143159144 Chr.3: 68753074-68753074 on Build GRCh38
rs377366529 Chr.3: 68753051-68753051 on Build GRCh38
rs368282756 Chr.3: 68752820-68752820 on Build GRCh38
rs762741649 Chr.3: 68752853-68752853 on Build GRCh38
rs954076269 Chr.3: 68752843-68752843 on Build GRCh38
rs267599929 Chr.3: 68752953-68752953 on Build GRCh38
rs1011863703 Chr.3: 68752670-68752670 on Build GRCh38
rs755227804 Chr.3: 68752898-68752898 on Build GRCh38
rs770818329 Chr.3: 68752909-68752909 on Build GRCh38
rs763133092 Chr.3: 68752919-68752919 on Build GRCh38
rs573581692 Chr.3: 68752986-68752986 on Build GRCh38
rs377215380 Chr.3: 68753034-68753034 on Build GRCh38
rs1011810873 Chr.3: 68752959-68752959 on Build GRCh38
rs752985210 Chr.3: 68752969-68752969 on Build GRCh38
rs988933563 Chr.3: 68753109-68753109 on Build GRCh38
rs549619294 Chr.3: 68752753-68752753 on Build GRCh38
rs377213840 Chr.3: 68752849-68752849 on Build GRCh38
rs148065010 Chr.3: 68752886-68752886 on Build GRCh38
rs779978104 Chr.3: 68753060-68753060 on Build GRCh38
rs907033432 Chr.3: 68753092-68753092 on Build GRCh38
rs766885476 Chr.3: 68753021-68753021 on Build GRCh38
rs201978822 Chr.3: 68752981-68752981 on Build GRCh38
rs763789648 Chr.3: 68752854-68752854 on Build GRCh38
rs771578231 Chr.3: 68752999-68752999 on Build GRCh38
rs905550428 Chr.3: 68752706-68752706 on Build GRCh38
rs374609099 Chr.3: 68753035-68753035 on Build GRCh38

View Full Product Details