Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 99848216-99848718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMSS1
Entrez Gene ID
84319
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Gene Aliases
C3orf26
UniGene
Hs.581320
Gene Chromosome Location
Chr.3: 99817834-100178632 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Gene Symbol
FILIP1L
Entrez Gene ID
11259
Gene Name
filamin A interacting protein 1 like
Gene Aliases
DOC-1, DOC1, GIP130, GIP90
UniGene
Hs.104672
Gene Chromosome Location
Chr.3: 99833144-100114513 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4453723 c.3393A>T p.I1131I Chr.3: 99848283-99848283 on Build GRCh38
5682523 c.3365G>T p.R1122L Chr.3: 99848311-99848311 on Build GRCh38
7162818 c.3364C>G p.R1122G Chr.3: 99848312-99848312 on Build GRCh38
4839402 c.3272G>A p.R1091Q Chr.3: 99848404-99848404 on Build GRCh38
219252 c.3096C>G p.F1032L Chr.3: 99848580-99848580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761594087 Chr.3: 99848446-99848446 on Build GRCh38
rs374311994 Chr.3: 99848539-99848539 on Build GRCh38
rs781236985 Chr.3: 99848611-99848611 on Build GRCh38
rs760359050 Chr.3: 99848655-99848655 on Build GRCh38
rs371644128 Chr.3: 99848552-99848552 on Build GRCh38
rs755559496 Chr.3: 99848487-99848487 on Build GRCh38
rs776719704 Chr.3: 99848548-99848548 on Build GRCh38
rs756852166 Chr.3: 99848380-99848380 on Build GRCh38
rs201016244 Chr.3: 99848674-99848674 on Build GRCh38
rs200133385 Chr.3: 99848564-99848564 on Build GRCh38
rs981847130 Chr.3: 99848530-99848530 on Build GRCh38
rs770984258 Chr.3: 99848428-99848428 on Build GRCh38
rs761097817 Chr.3: 99848442-99848443 on Build GRCh38
rs191265397 Chr.3: 99848385-99848385 on Build GRCh38
rs748588338 Chr.3: 99848501-99848501 on Build GRCh38
rs751748900 Chr.3: 99848605-99848605 on Build GRCh38
rs537619245 Chr.3: 99848333-99848333 on Build GRCh38
rs370091852 Chr.3: 99848311-99848311 on Build GRCh38
rs531864166 Chr.3: 99848584-99848584 on Build GRCh38
rs374214004 Chr.3: 99848270-99848270 on Build GRCh38
rs746025167 Chr.3: 99848623-99848623 on Build GRCh38
rs961757882 Chr.3: 99848542-99848542 on Build GRCh38
rs529055199 Chr.3: 99848364-99848364 on Build GRCh38
rs371989728 Chr.3: 99848662-99848662 on Build GRCh38
rs775484361 Chr.3: 99848337-99848337 on Build GRCh38
rs757544873 Chr.3: 99848609-99848609 on Build GRCh38
rs767218439 Chr.3: 99848449-99848449 on Build GRCh38
rs758820845 Chr.3: 99848509-99848509 on Build GRCh38
rs769809880 Chr.3: 99848624-99848624 on Build GRCh38
rs762088031 Chr.3: 99848296-99848296 on Build GRCh38
rs201557511 Chr.3: 99848474-99848474 on Build GRCh38
rs202015243 Chr.3: 99848414-99848414 on Build GRCh38
rs747046056 Chr.3: 99848456-99848456 on Build GRCh38
rs372920695 Chr.3: 99848445-99848445 on Build GRCh38
rs765926299 Chr.3: 99848581-99848581 on Build GRCh38
rs36118713 Chr.3: 99848428-99848428 on Build GRCh38
rs140926387 Chr.3: 99848328-99848329 on Build GRCh38
rs751240509 Chr.3: 99848376-99848376 on Build GRCh38
rs866526364 Chr.3: 99848364-99848364 on Build GRCh38
rs749740482 Chr.3: 99848395-99848395 on Build GRCh38
rs758837225 Chr.3: 99848588-99848588 on Build GRCh38
rs755330466 Chr.3: 99848399-99848399 on Build GRCh38
rs79681183 Chr.3: 99848381-99848381 on Build GRCh38
rs773561739 Chr.3: 99848420-99848420 on Build GRCh38
rs200374375 Chr.3: 99848373-99848373 on Build GRCh38
rs778153475 Chr.3: 99848513-99848513 on Build GRCh38
rs368522732 Chr.3: 99848540-99848540 on Build GRCh38
rs140594225 Chr.3: 99848431-99848431 on Build GRCh38
rs747114419 Chr.3: 99848422-99848422 on Build GRCh38
rs748039430 Chr.3: 99848638-99848638 on Build GRCh38
rs776132796 Chr.3: 99848676-99848676 on Build GRCh38
rs376027916 Chr.3: 99848498-99848498 on Build GRCh38
rs182963235 Chr.3: 99848563-99848563 on Build GRCh38
rs772775239 Chr.3: 99848567-99848567 on Build GRCh38
rs771880511 Chr.3: 99848640-99848640 on Build GRCh38
rs759128263 Chr.3: 99848680-99848680 on Build GRCh38
rs1038463323 Chr.3: 99848532-99848532 on Build GRCh38
rs754267001 Chr.3: 99848569-99848569 on Build GRCh38
rs367809134 Chr.3: 99848652-99848652 on Build GRCh38
rs1009779464 Chr.3: 99848572-99848572 on Build GRCh38
rs778830328 Chr.3: 99848633-99848633 on Build GRCh38
rs377189691 Chr.3: 99848272-99848272 on Build GRCh38
rs750004088 Chr.3: 99848383-99848383 on Build GRCh38
rs749676489 Chr.3: 99848271-99848271 on Build GRCh38
rs772465470 Chr.3: 99848417-99848417 on Build GRCh38
rs764422883 Chr.3: 99848466-99848466 on Build GRCh38
rs750024164 Chr.3: 99848452-99848452 on Build GRCh38
rs377075355 Chr.3: 99848404-99848404 on Build GRCh38
rs762904498 Chr.3: 99848436-99848436 on Build GRCh38
rs556347627 Chr.3: 99848355-99848355 on Build GRCh38
rs372595024 Chr.3: 99848312-99848312 on Build GRCh38
rs759303833 Chr.3: 99848520-99848521 on Build GRCh38
rs755588488 Chr.3: 99848459-99848459 on Build GRCh38
rs774720280 Chr.3: 99848273-99848273 on Build GRCh38
rs759114259 Chr.3: 99848586-99848586 on Build GRCh38
rs760804100 Chr.3: 99848330-99848330 on Build GRCh38
rs549292963 Chr.3: 99848580-99848580 on Build GRCh38
rs747188276 Chr.3: 99848526-99848526 on Build GRCh38
rs746394253 Chr.3: 99848256-99848256 on Build GRCh38
rs778243256 Chr.3: 99848590-99848590 on Build GRCh38
rs781184828 Chr.3: 99848269-99848269 on Build GRCh38
rs780442992 Chr.3: 99848393-99848393 on Build GRCh38
rs763879422 Chr.3: 99848360-99848360 on Build GRCh38
rs763895478 Chr.3: 99848444-99848444 on Build GRCh38
rs550375139 Chr.3: 99848589-99848589 on Build GRCh38
rs775903903 Chr.3: 99848387-99848392 on Build GRCh38
rs765900822 Chr.3: 99848587-99848587 on Build GRCh38

View Full Product Details