Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113903927-113904440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1C
Entrez Gene ID
54762
Gene Name
GRAM domain containing 1C
Gene Aliases
-
UniGene
Hs.24583
Gene Chromosome Location
Chr.3: 113828182-113947174 on Build GRCh38
Transcripts
NM_001172105.1 NM_017577.4 XM_005247546.2 XM_005247547.1 XM_011512930.1 XM_011512931.1 XM_017006646.1 XM_017006647.1 XM_017006648.1 XM_017006649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3119181 c.787G>A p.G263R Chr.3: 113904270-113904270 on Build GRCh38
727150 c.711C>A p.S237S Chr.3: 113904194-113904194 on Build GRCh38
7276347 c.741T>G p.I247M Chr.3: 113904224-113904224 on Build GRCh38
1253619 c.775A>G p.N259D Chr.3: 113904258-113904258 on Build GRCh38
5784976 c.789A>T p.G263G Chr.3: 113904272-113904272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543693993 Chr.3: 113904117-113904117 on Build GRCh38
rs952395235 Chr.3: 113904152-113904152 on Build GRCh38
rs529767349 Chr.3: 113904154-113904154 on Build GRCh38
rs747557960 Chr.3: 113904291-113904291 on Build GRCh38
rs766164685 Chr.3: 113904192-113904192 on Build GRCh38
rs1042746129 Chr.3: 113903976-113903976 on Build GRCh38
rs377301886 Chr.3: 113904244-113904244 on Build GRCh38
rs889642988 Chr.3: 113904080-113904080 on Build GRCh38
rs145722654 Chr.3: 113903969-113903969 on Build GRCh38
rs777703882 Chr.3: 113904091-113904091 on Build GRCh38
rs776471004 Chr.3: 113904194-113904194 on Build GRCh38
rs201881349 Chr.3: 113904276-113904276 on Build GRCh38
rs936946466 Chr.3: 113904166-113904166 on Build GRCh38
rs750971322 Chr.3: 113904251-113904251 on Build GRCh38
rs371245272 Chr.3: 113904155-113904155 on Build GRCh38
rs148862856 Chr.3: 113904016-113904016 on Build GRCh38
rs531928756 Chr.3: 113904044-113904044 on Build GRCh38
rs778657971 Chr.3: 113904289-113904289 on Build GRCh38
rs925297124 Chr.3: 113904393-113904393 on Build GRCh38
rs1027108516 Chr.3: 113904128-113904128 on Build GRCh38
rs987196891 Chr.3: 113904081-113904081 on Build GRCh38
rs754552487 Chr.3: 113904242-113904243 on Build GRCh38
rs754554885 Chr.3: 113904284-113904284 on Build GRCh38
rs762857848 Chr.3: 113904005-113904005 on Build GRCh38
rs751556575 Chr.3: 113904214-113904214 on Build GRCh38
rs774123805 Chr.3: 113904188-113904188 on Build GRCh38
rs967229574 Chr.3: 113904350-113904350 on Build GRCh38
rs57338076 Chr.3: 113904173-113904173 on Build GRCh38
rs886528922 Chr.3: 113904260-113904260 on Build GRCh38
rs763142006 Chr.3: 113904197-113904197 on Build GRCh38
rs576890490 Chr.3: 113904231-113904231 on Build GRCh38
rs945399308 Chr.3: 113903970-113903970 on Build GRCh38
rs773711093 Chr.3: 113904157-113904157 on Build GRCh38
rs145513627 Chr.3: 113904000-113904000 on Build GRCh38
rs770297653 Chr.3: 113904138-113904138 on Build GRCh38
rs374197239 Chr.3: 113904229-113904229 on Build GRCh38
rs1040816051 Chr.3: 113904111-113904111 on Build GRCh38
rs757380657 Chr.3: 113904226-113904226 on Build GRCh38
rs773151721 Chr.3: 113904187-113904187 on Build GRCh38
rs757677321 Chr.3: 113904113-113904113 on Build GRCh38
rs371194067 Chr.3: 113904252-113904252 on Build GRCh38
rs527540910 Chr.3: 113904355-113904355 on Build GRCh38
rs779788028 Chr.3: 113904142-113904142 on Build GRCh38
rs936630652 Chr.3: 113904053-113904053 on Build GRCh38
rs751713626 Chr.3: 113904108-113904108 on Build GRCh38
rs749718635 Chr.3: 113904278-113904278 on Build GRCh38
rs552450618 Chr.3: 113904375-113904375 on Build GRCh38
rs182122033 Chr.3: 113904125-113904125 on Build GRCh38
rs543490044 Chr.3: 113904014-113904014 on Build GRCh38
rs1049611191 Chr.3: 113904061-113904061 on Build GRCh38
rs764420275 Chr.3: 113904201-113904201 on Build GRCh38
rs376774312 Chr.3: 113904126-113904126 on Build GRCh38
rs1007985401 Chr.3: 113904109-113904109 on Build GRCh38
rs564474189 Chr.3: 113904031-113904031 on Build GRCh38
rs747733918 Chr.3: 113904163-113904163 on Build GRCh38
rs771621710 Chr.3: 113904312-113904312 on Build GRCh38
rs57654265 Chr.3: 113904357-113904357 on Build GRCh38
rs1016713014 Chr.3: 113904407-113904407 on Build GRCh38

View Full Product Details