Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 115804543-115805072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSAMP
Entrez Gene ID
4045
Gene Name
limbic system-associated membrane protein
Gene Aliases
IGLON3, LAMP
UniGene
Hs.26409
Gene Chromosome Location
Chr.3: 115802363-116445542 on Build GRCh38
Transcripts
NM_001318915.1 NM_002338.4

SNP Detail

SNP ID Chromosome Location
rs982228800 Chr.3: 115804712-115804712 on Build GRCh38
rs998540558 Chr.3: 115804739-115804739 on Build GRCh38
rs950319118 Chr.3: 115804580-115804581 on Build GRCh38
rs929133264 Chr.3: 115804805-115804805 on Build GRCh38
rs139369136 Chr.3: 115804666-115804666 on Build GRCh38
rs959735360 Chr.3: 115805011-115805011 on Build GRCh38
rs542568367 Chr.3: 115804959-115804959 on Build GRCh38
rs183356695 Chr.3: 115804711-115804711 on Build GRCh38
rs1006720445 Chr.3: 115804770-115804770 on Build GRCh38
rs188417444 Chr.3: 115804762-115804762 on Build GRCh38
rs1051559540 Chr.3: 115804845-115804845 on Build GRCh38
rs927539690 Chr.3: 115804881-115804881 on Build GRCh38
rs1045947858 Chr.3: 115804581-115804584 on Build GRCh38
rs1003717309 Chr.3: 115804987-115804987 on Build GRCh38
rs1007634011 Chr.3: 115804589-115804589 on Build GRCh38
rs1041792889 Chr.3: 115804941-115804941 on Build GRCh38
rs755789244 Chr.3: 115804934-115804934 on Build GRCh38
rs1037564605 Chr.3: 115804787-115804787 on Build GRCh38
rs143265450 Chr.3: 115804957-115804957 on Build GRCh38
rs1016696185 Chr.3: 115804792-115804792 on Build GRCh38
rs111700486 Chr.3: 115804872-115804872 on Build GRCh38
rs937521579 Chr.3: 115804890-115804890 on Build GRCh38
rs927487860 Chr.3: 115804589-115804590 on Build GRCh38
rs541199069 Chr.3: 115804735-115804735 on Build GRCh38
rs779457288 Chr.3: 115804788-115804788 on Build GRCh38
rs1021149279 Chr.3: 115804674-115804674 on Build GRCh38
rs958537514 Chr.3: 115804619-115804619 on Build GRCh38
rs541175985 Chr.3: 115804649-115804649 on Build GRCh38
rs893179466 Chr.3: 115804632-115804632 on Build GRCh38
rs972438384 Chr.3: 115804819-115804819 on Build GRCh38
rs914682699 Chr.3: 115804926-115804926 on Build GRCh38
rs564028461 Chr.3: 115804849-115804849 on Build GRCh38
rs778617977 Chr.3: 115804993-115804993 on Build GRCh38
rs753773505 Chr.3: 115804823-115804823 on Build GRCh38
rs115379975 Chr.3: 115804935-115804935 on Build GRCh38
rs890203737 Chr.3: 115804847-115804847 on Build GRCh38
rs1007274282 Chr.3: 115804896-115804896 on Build GRCh38
rs919119663 Chr.3: 115804789-115804789 on Build GRCh38
rs937505522 Chr.3: 115804590-115804590 on Build GRCh38
rs750129195 Chr.3: 115804866-115804866 on Build GRCh38
rs556065801 Chr.3: 115805031-115805031 on Build GRCh38
rs901865817 Chr.3: 115804955-115804955 on Build GRCh38
rs1042088745 Chr.3: 115804734-115804734 on Build GRCh38
rs111827272 Chr.3: 115804917-115804917 on Build GRCh38
rs1021211293 Chr.3: 115804969-115804969 on Build GRCh38
rs1006746505 Chr.3: 115805045-115805045 on Build GRCh38
rs971068128 Chr.3: 115804846-115804846 on Build GRCh38
rs981696604 Chr.3: 115804863-115804863 on Build GRCh38
rs966910233 Chr.3: 115804970-115804970 on Build GRCh38
rs1017229441 Chr.3: 115804612-115804612 on Build GRCh38
rs889668265 Chr.3: 115805008-115805008 on Build GRCh38
rs928037499 Chr.3: 115804722-115804722 on Build GRCh38
rs560999169 Chr.3: 115804994-115804994 on Build GRCh38
rs941970554 Chr.3: 115804763-115804763 on Build GRCh38
rs991236885 Chr.3: 115804743-115804743 on Build GRCh38
rs1054501364 Chr.3: 115804601-115804601 on Build GRCh38

View Full Product Details