Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132447810-132448313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC13
Entrez Gene ID
23317
Gene Name
DnaJ heat shock protein family (Hsp40) member C13
Gene Aliases
PARK21, RME8
UniGene
Hs.12707
Gene Chromosome Location
Chr.3: 132417527-132539032 on Build GRCh38
Transcripts
NM_015268.3 XM_005247245.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6647199 c.321delA p.I109fs*20 Chr.3: 132447924-132447924 on Build GRCh38
5978720 c.332G>A p.G111E Chr.3: 132447935-132447935 on Build GRCh38
6647246 c.320_321insA p.I109fs*10 Chr.3: 132447923-132447924 on Build GRCh38
5699267 c.316G>A p.E106K Chr.3: 132447919-132447919 on Build GRCh38
1484627 c.304A>G p.T102A Chr.3: 132447907-132447907 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs978152927 Chr.3: 132448188-132448188 on Build GRCh38
rs906197566 Chr.3: 132448143-132448143 on Build GRCh38
rs200037269 Chr.3: 132447961-132447961 on Build GRCh38
rs891937633 Chr.3: 132447977-132447977 on Build GRCh38
rs1037094873 Chr.3: 132447838-132447838 on Build GRCh38
rs79484675 Chr.3: 132447944-132447944 on Build GRCh38
rs553468764 Chr.3: 132448020-132448020 on Build GRCh38
rs975920265 Chr.3: 132448113-132448113 on Build GRCh38
rs759793587 Chr.3: 132447910-132447910 on Build GRCh38
rs767329689 Chr.3: 132447986-132447986 on Build GRCh38
rs199967561 Chr.3: 132447855-132447855 on Build GRCh38
rs772855237 Chr.3: 132447953-132447953 on Build GRCh38
rs184768221 Chr.3: 132448265-132448265 on Build GRCh38
rs1055095325 Chr.3: 132448166-132448166 on Build GRCh38
rs756777370 Chr.3: 132448130-132448130 on Build GRCh38
rs1023971167 Chr.3: 132447994-132447994 on Build GRCh38
rs747409180 Chr.3: 132447879-132447879 on Build GRCh38
rs533581215 Chr.3: 132448215-132448215 on Build GRCh38
rs1003853784 Chr.3: 132448109-132448109 on Build GRCh38
rs978056613 Chr.3: 132447995-132447995 on Build GRCh38
rs772683718 Chr.3: 132447873-132447873 on Build GRCh38
rs201115847 Chr.3: 132447956-132447956 on Build GRCh38
rs748705018 Chr.3: 132447858-132447858 on Build GRCh38
rs570371231 Chr.3: 132448085-132448085 on Build GRCh38
rs1010327533 Chr.3: 132448175-132448175 on Build GRCh38
rs906960630 Chr.3: 132448239-132448239 on Build GRCh38
rs1011041247 Chr.3: 132448170-132448170 on Build GRCh38
rs866692930 Chr.3: 132448010-132448010 on Build GRCh38
rs755637002 Chr.3: 132447852-132447852 on Build GRCh38
rs755398108 Chr.3: 132448267-132448267 on Build GRCh38
rs1049731558 Chr.3: 132448153-132448153 on Build GRCh38
rs35275519 Chr.3: 132448062-132448063 on Build GRCh38
rs921150387 Chr.3: 132448281-132448281 on Build GRCh38
rs959810108 Chr.3: 132448160-132448160 on Build GRCh38
rs139413962 Chr.3: 132448153-132448153 on Build GRCh38
rs181809057 Chr.3: 132447991-132447991 on Build GRCh38
rs188289098 Chr.3: 132447875-132447875 on Build GRCh38
rs866385737 Chr.3: 132448114-132448114 on Build GRCh38
rs932496636 Chr.3: 132448084-132448084 on Build GRCh38
rs571681734 Chr.3: 132448049-132448049 on Build GRCh38
rs1010381404 Chr.3: 132447992-132447992 on Build GRCh38
rs538310102 Chr.3: 132448016-132448016 on Build GRCh38
rs753357931 Chr.3: 132448100-132448100 on Build GRCh38
rs771262156 Chr.3: 132447889-132447889 on Build GRCh38
rs12639443 Chr.3: 132448171-132448171 on Build GRCh38
rs368990477 Chr.3: 132447955-132447955 on Build GRCh38
rs776734868 Chr.3: 132447907-132447907 on Build GRCh38
rs976702383 Chr.3: 132448247-132448247 on Build GRCh38
rs560761477 Chr.3: 132448070-132448070 on Build GRCh38
rs934101272 Chr.3: 132448147-132448147 on Build GRCh38
rs751428770 Chr.3: 132447962-132447962 on Build GRCh38
rs1001716518 Chr.3: 132448270-132448270 on Build GRCh38
rs544578517 Chr.3: 132448155-132448155 on Build GRCh38
rs921128643 Chr.3: 132448064-132448064 on Build GRCh38
rs966024450 Chr.3: 132448235-132448235 on Build GRCh38
rs1050221443 Chr.3: 132447918-132447918 on Build GRCh38
rs762967623 Chr.3: 132447960-132447960 on Build GRCh38
rs765305595 Chr.3: 132447928-132447928 on Build GRCh38
rs77604326 Chr.3: 132448050-132448050 on Build GRCh38
rs371535590 Chr.3: 132448152-132448153 on Build GRCh38
rs751513409 Chr.3: 132448086-132448086 on Build GRCh38
rs757055216 Chr.3: 132447983-132447983 on Build GRCh38
rs985140763 Chr.3: 132448063-132448063 on Build GRCh38
rs1045913297 Chr.3: 132448197-132448197 on Build GRCh38
rs915291247 Chr.3: 132447869-132447869 on Build GRCh38
rs911924118 Chr.3: 132448206-132448206 on Build GRCh38
rs966794028 Chr.3: 132448179-132448179 on Build GRCh38

View Full Product Details