Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 136945341-136945840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCK1
Entrez Gene ID
4690
Gene Name
NCK adaptor protein 1
Gene Aliases
NCK, NCKalpha, nck-1
UniGene
Hs.126889
Gene Chromosome Location
Chr.3: 136862208-136951610 on Build GRCh38
Transcripts
NM_001190796.2 NM_001291999.1 NM_006153.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
373489 c.358G>A p.E120K Chr.3: 136945714-136945714 on Build GRCh38
5671761 c.256G>T p.V86L Chr.3: 136945612-136945612 on Build GRCh38
5480265 c.227-8T>A p.? Chr.3: 136945575-136945575 on Build GRCh38
1316244 c.409G>T p.E137* Chr.3: 136945765-136945765 on Build GRCh38
6544867 c.263A>T p.D88V Chr.3: 136945619-136945619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541989658 Chr.3: 136945370-136945370 on Build GRCh38
rs780640959 Chr.3: 136945705-136945705 on Build GRCh38
rs749532686 Chr.3: 136945579-136945579 on Build GRCh38
rs779358036 Chr.3: 136945588-136945588 on Build GRCh38
rs185485335 Chr.3: 136945634-136945634 on Build GRCh38
rs764250587 Chr.3: 136945668-136945668 on Build GRCh38
rs924408333 Chr.3: 136945480-136945480 on Build GRCh38
rs201351324 Chr.3: 136945584-136945584 on Build GRCh38
rs747529728 Chr.3: 136945707-136945707 on Build GRCh38
rs147481903 Chr.3: 136945740-136945740 on Build GRCh38
rs568946678 Chr.3: 136945701-136945701 on Build GRCh38
rs747727219 Chr.3: 136945618-136945618 on Build GRCh38
rs981278927 Chr.3: 136945680-136945680 on Build GRCh38
rs777928916 Chr.3: 136945572-136945572 on Build GRCh38
rs909975297 Chr.3: 136945414-136945414 on Build GRCh38
rs368340310 Chr.3: 136945691-136945691 on Build GRCh38
rs767504687 Chr.3: 136945658-136945658 on Build GRCh38
rs756148728 Chr.3: 136945369-136945369 on Build GRCh38
rs796360177 Chr.3: 136945508-136945508 on Build GRCh38
rs903641259 Chr.3: 136945512-136945512 on Build GRCh38
rs756123065 Chr.3: 136945664-136945664 on Build GRCh38
rs571846015 Chr.3: 136945518-136945518 on Build GRCh38
rs774239593 Chr.3: 136945643-136945643 on Build GRCh38
rs769135774 Chr.3: 136945708-136945708 on Build GRCh38
rs773344757 Chr.3: 136945663-136945663 on Build GRCh38
rs1046119678 Chr.3: 136945535-136945535 on Build GRCh38
rs141442402 Chr.3: 136945702-136945702 on Build GRCh38
rs772680921 Chr.3: 136945630-136945630 on Build GRCh38
rs766700438 Chr.3: 136945635-136945637 on Build GRCh38
rs905013955 Chr.3: 136945440-136945440 on Build GRCh38
rs777371332 Chr.3: 136945722-136945722 on Build GRCh38
rs757532603 Chr.3: 136945684-136945684 on Build GRCh38
rs752774458 Chr.3: 136945660-136945660 on Build GRCh38
rs1007702856 Chr.3: 136945527-136945527 on Build GRCh38
rs746338229 Chr.3: 136945594-136945594 on Build GRCh38
rs191952779 Chr.3: 136945431-136945431 on Build GRCh38
rs746377943 Chr.3: 136945603-136945610 on Build GRCh38
rs548833073 Chr.3: 136945686-136945686 on Build GRCh38
rs915799054 Chr.3: 136945528-136945528 on Build GRCh38
rs762533477 Chr.3: 136945789-136945789 on Build GRCh38
rs137920757 Chr.3: 136945561-136945561 on Build GRCh38
rs544364534 Chr.3: 136945449-136945449 on Build GRCh38
rs1029293238 Chr.3: 136945679-136945679 on Build GRCh38
rs1056414752 Chr.3: 136945481-136945481 on Build GRCh38
rs767549636 Chr.3: 136945540-136945540 on Build GRCh38
rs759443661 Chr.3: 136945648-136945648 on Build GRCh38
rs990123760 Chr.3: 136945493-136945493 on Build GRCh38
rs748857974 Chr.3: 136945732-136945732 on Build GRCh38
rs140020695 Chr.3: 136945762-136945762 on Build GRCh38
rs545821475 Chr.3: 136945498-136945498 on Build GRCh38
rs531554309 Chr.3: 136945733-136945733 on Build GRCh38
rs374736118 Chr.3: 136945532-136945532 on Build GRCh38
rs1044931569 Chr.3: 136945433-136945433 on Build GRCh38
rs150286331 Chr.3: 136945416-136945416 on Build GRCh38
rs564448025 Chr.3: 136945482-136945482 on Build GRCh38
rs1018473275 Chr.3: 136945530-136945530 on Build GRCh38
rs183326283 Chr.3: 136945611-136945611 on Build GRCh38
rs762687588 Chr.3: 136945633-136945633 on Build GRCh38
rs113253727 Chr.3: 136945599-136945599 on Build GRCh38
rs35310696 Chr.3: 136945660-136945661 on Build GRCh38
rs533411887 Chr.3: 136945507-136945507 on Build GRCh38
rs1009245824 Chr.3: 136945500-136945500 on Build GRCh38
rs771863858 Chr.3: 136945744-136945744 on Build GRCh38
rs887984944 Chr.3: 136945570-136945570 on Build GRCh38
rs769432770 Chr.3: 136945628-136945628 on Build GRCh38
rs764042177 Chr.3: 136945758-136945758 on Build GRCh38
rs1056277981 Chr.3: 136945687-136945687 on Build GRCh38
rs775351154 Chr.3: 136945747-136945747 on Build GRCh38
rs760631298 Chr.3: 136945756-136945756 on Build GRCh38
rs982705367 Chr.3: 136945374-136945374 on Build GRCh38
rs752819563 Chr.3: 136945560-136945560 on Build GRCh38
rs1007197278 Chr.3: 136945793-136945793 on Build GRCh38

View Full Product Details