Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148697759-148698264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTR1
Entrez Gene ID
185
Gene Name
angiotensin II receptor type 1
Gene Aliases
AG2S, AGTR1B, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, HAT1R
UniGene
Hs.477887
Gene Chromosome Location
Chr.3: 148697871-148743003 on Build GRCh38
Transcripts
NM_000685.4 NM_004835.4 NM_009585.3 NM_031850.3

SNP Detail

SNP ID Chromosome Location
rs944058817 Chr.3: 148698212-148698212 on Build GRCh38
rs985559117 Chr.3: 148698189-148698189 on Build GRCh38
rs547351627 Chr.3: 148698020-148698020 on Build GRCh38
rs867925661 Chr.3: 148698119-148698119 on Build GRCh38
rs779753702 Chr.3: 148697835-148697835 on Build GRCh38
rs984504840 Chr.3: 148697908-148697908 on Build GRCh38
rs1018775404 Chr.3: 148697839-148697839 on Build GRCh38
rs886058068 Chr.3: 148697928-148697928 on Build GRCh38
rs916854776 Chr.3: 148697921-148697921 on Build GRCh38
rs893515670 Chr.3: 148698032-148698032 on Build GRCh38
rs922414169 Chr.3: 148698017-148698017 on Build GRCh38
rs1012658089 Chr.3: 148698067-148698067 on Build GRCh38
rs1030662811 Chr.3: 148697787-148697787 on Build GRCh38
rs2307085 Chr.3: 148698075-148698075 on Build GRCh38
rs143307123 Chr.3: 148697793-148697793 on Build GRCh38
rs1022602852 Chr.3: 148697813-148697813 on Build GRCh38
rs903073108 Chr.3: 148697997-148697997 on Build GRCh38
rs868049910 Chr.3: 148697895-148697895 on Build GRCh38
rs570771276 Chr.3: 148698069-148698069 on Build GRCh38
rs912546133 Chr.3: 148697910-148697910 on Build GRCh38
rs1013013013 Chr.3: 148697806-148697806 on Build GRCh38
rs1010589941 Chr.3: 148698050-148698050 on Build GRCh38
rs572286069 Chr.3: 148698190-148698190 on Build GRCh38
rs931305294 Chr.3: 148697900-148697900 on Build GRCh38
rs569309821 Chr.3: 148698083-148698083 on Build GRCh38
rs957077562 Chr.3: 148697800-148697800 on Build GRCh38
rs113745668 Chr.3: 148697810-148697810 on Build GRCh38
rs563682638 Chr.3: 148697837-148697837 on Build GRCh38
rs746560181 Chr.3: 148698001-148698001 on Build GRCh38
rs549707567 Chr.3: 148698080-148698080 on Build GRCh38
rs1046585544 Chr.3: 148697983-148697983 on Build GRCh38
rs971852664 Chr.3: 148698145-148698145 on Build GRCh38
rs561831399 Chr.3: 148698012-148698012 on Build GRCh38
rs961582669 Chr.3: 148698089-148698089 on Build GRCh38
rs948293232 Chr.3: 148697935-148697935 on Build GRCh38
rs893463798 Chr.3: 148697789-148697789 on Build GRCh38
rs527515255 Chr.3: 148698013-148698013 on Build GRCh38
rs148296224 Chr.3: 148697883-148697883 on Build GRCh38
rs182349326 Chr.3: 148698228-148698228 on Build GRCh38
rs1015810633 Chr.3: 148697838-148697838 on Build GRCh38
rs1018825817 Chr.3: 148698093-148698093 on Build GRCh38
rs1003571253 Chr.3: 148698076-148698076 on Build GRCh38
rs1040282649 Chr.3: 148697930-148697930 on Build GRCh38
rs529360494 Chr.3: 148697873-148697873 on Build GRCh38
rs916552911 Chr.3: 148698232-148698232 on Build GRCh38
rs771716932 Chr.3: 148697786-148697786 on Build GRCh38
rs974257630 Chr.3: 148698124-148698124 on Build GRCh38
rs932699376 Chr.3: 148697884-148697884 on Build GRCh38
rs535148653 Chr.3: 148698128-148698128 on Build GRCh38
rs912588062 Chr.3: 148698191-148698191 on Build GRCh38
rs892899199 Chr.3: 148698051-148698051 on Build GRCh38
rs961531182 Chr.3: 148697860-148697860 on Build GRCh38
rs935115393 Chr.3: 148698011-148698011 on Build GRCh38
rs1027141886 Chr.3: 148698151-148698151 on Build GRCh38
rs543841089 Chr.3: 148697802-148697802 on Build GRCh38
rs944658745 Chr.3: 148697928-148697928 on Build GRCh38
rs191683621 Chr.3: 148698161-148698161 on Build GRCh38

View Full Product Details