Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155079555-155080050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MME
Entrez Gene ID
4311
Gene Name
membrane metallo-endopeptidase
Gene Aliases
CALLA, CD10, CMT2T, NEP, SFE
UniGene
Hs.307734
Gene Chromosome Location
Chr.3: 155024124-155183729 on Build GRCh38
Transcripts
NM_000902.3 NM_007287.2 XM_011512855.2

SNP Detail

SNP ID Chromosome Location
rs995681040 Chr.3: 155079903-155079903 on Build GRCh38
rs903048182 Chr.3: 155079810-155079810 on Build GRCh38
rs1018017132 Chr.3: 155079688-155079688 on Build GRCh38
rs1007924730 Chr.3: 155079664-155079664 on Build GRCh38
rs537664389 Chr.3: 155080002-155080002 on Build GRCh38
rs940457469 Chr.3: 155079732-155079732 on Build GRCh38
rs930227832 Chr.3: 155079577-155079577 on Build GRCh38
rs1004468087 Chr.3: 155079926-155079926 on Build GRCh38
rs899550549 Chr.3: 155079902-155079902 on Build GRCh38
rs968813979 Chr.3: 155079999-155079999 on Build GRCh38
rs933018908 Chr.3: 155079750-155079750 on Build GRCh38
rs9809268 Chr.3: 155079639-155079639 on Build GRCh38
rs202082495 Chr.3: 155079639-155079639 on Build GRCh38
rs903741176 Chr.3: 155079636-155079636 on Build GRCh38
rs991627847 Chr.3: 155079964-155079964 on Build GRCh38
rs947370171 Chr.3: 155079634-155079634 on Build GRCh38
rs201751860 Chr.3: 155079636-155079639 on Build GRCh38
rs868044382 Chr.3: 155079772-155079772 on Build GRCh38
rs894480069 Chr.3: 155079633-155079633 on Build GRCh38
rs371601308 Chr.3: 155079961-155079961 on Build GRCh38
rs928782102 Chr.3: 155079771-155079771 on Build GRCh38
rs960237383 Chr.3: 155079776-155079776 on Build GRCh38
rs938736587 Chr.3: 155079613-155079613 on Build GRCh38
rs867678854 Chr.3: 155079630-155079630 on Build GRCh38
rs951156282 Chr.3: 155079910-155079910 on Build GRCh38
rs890955711 Chr.3: 155079851-155079851 on Build GRCh38
rs73874497 Chr.3: 155079597-155079597 on Build GRCh38
rs1052371625 Chr.3: 155079623-155079623 on Build GRCh38
rs533777704 Chr.3: 155079852-155079852 on Build GRCh38
rs566382653 Chr.3: 155079806-155079806 on Build GRCh38
rs999868846 Chr.3: 155079653-155079653 on Build GRCh38
rs935248828 Chr.3: 155079837-155079837 on Build GRCh38
rs951636773 Chr.3: 155079725-155079725 on Build GRCh38
rs998670837 Chr.3: 155079818-155079818 on Build GRCh38
rs868566601 Chr.3: 155079774-155079774 on Build GRCh38
rs1052336458 Chr.3: 155079843-155079843 on Build GRCh38
rs192718956 Chr.3: 155079904-155079904 on Build GRCh38
rs979282794 Chr.3: 155080000-155080000 on Build GRCh38
rs1014011527 Chr.3: 155079855-155079855 on Build GRCh38
rs947472454 Chr.3: 155079798-155079798 on Build GRCh38
rs915963791 Chr.3: 155079794-155079794 on Build GRCh38
rs1027079035 Chr.3: 155079720-155079720 on Build GRCh38
rs992171033 Chr.3: 155079641-155079641 on Build GRCh38
rs962206238 Chr.3: 155079917-155079917 on Build GRCh38
rs982918204 Chr.3: 155079594-155079594 on Build GRCh38
rs9864458 Chr.3: 155079643-155079643 on Build GRCh38
rs567181239 Chr.3: 155079979-155079979 on Build GRCh38
rs999584914 Chr.3: 155079635-155079636 on Build GRCh38
rs991659066 Chr.3: 155079789-155079789 on Build GRCh38
rs770101542 Chr.3: 155079689-155079689 on Build GRCh38
rs896508309 Chr.3: 155079624-155079624 on Build GRCh38
rs551437913 Chr.3: 155079731-155079731 on Build GRCh38
rs969632921 Chr.3: 155079705-155079705 on Build GRCh38
rs539514715 Chr.3: 155079629-155079629 on Build GRCh38
rs1043458509 Chr.3: 155079635-155079635 on Build GRCh38
rs923045714 Chr.3: 155080017-155080017 on Build GRCh38
rs984926678 Chr.3: 155079712-155079712 on Build GRCh38
rs780446799 Chr.3: 155079901-155079901 on Build GRCh38
rs923003794 Chr.3: 155079747-155079747 on Build GRCh38
rs890986715 Chr.3: 155079662-155079662 on Build GRCh38
rs867086527 Chr.3: 155079632-155079632 on Build GRCh38
rs1035906394 Chr.3: 155079944-155079944 on Build GRCh38
rs924629824 Chr.3: 155079830-155079830 on Build GRCh38
rs1031079572 Chr.3: 155079636-155079637 on Build GRCh38
rs774330916 Chr.3: 155079638-155079639 on Build GRCh38
rs970062414 Chr.3: 155079902-155079903 on Build GRCh38
rs973728013 Chr.3: 155079693-155079693 on Build GRCh38
rs1039517977 Chr.3: 155079856-155079856 on Build GRCh38
rs1030772550 Chr.3: 155079661-155079661 on Build GRCh38
rs868320892 Chr.3: 155079631-155079631 on Build GRCh38
rs983062227 Chr.3: 155079756-155079756 on Build GRCh38
rs933047293 Chr.3: 155080027-155080027 on Build GRCh38
rs924626298 Chr.3: 155080028-155080028 on Build GRCh38
rs1036097579 Chr.3: 155079741-155079741 on Build GRCh38

View Full Product Details