Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155080020-155080553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MME
Entrez Gene ID
4311
Gene Name
membrane metallo-endopeptidase
Gene Aliases
CALLA, CD10, CMT2T, NEP, SFE
UniGene
Hs.307734
Gene Chromosome Location
Chr.3: 155024124-155183729 on Build GRCh38
Transcripts
NM_000902.3 NM_007287.2 NM_007288.2 NM_007289.2 XM_006713646.3 XM_006713647.3 XM_011512855.2

SNP Detail

SNP ID Chromosome Location
rs1031849810 Chr.3: 155080512-155080512 on Build GRCh38
rs1056954429 Chr.3: 155080084-155080084 on Build GRCh38
rs911910406 Chr.3: 155080304-155080304 on Build GRCh38
rs905218872 Chr.3: 155080137-155080137 on Build GRCh38
rs921106016 Chr.3: 155080332-155080332 on Build GRCh38
rs200485392 Chr.3: 155080470-155080470 on Build GRCh38
rs555911647 Chr.3: 155080071-155080071 on Build GRCh38
rs763305151 Chr.3: 155080288-155080288 on Build GRCh38
rs201044903 Chr.3: 155080403-155080403 on Build GRCh38
rs939770549 Chr.3: 155080387-155080387 on Build GRCh38
rs1037631651 Chr.3: 155080361-155080361 on Build GRCh38
rs544054425 Chr.3: 155080526-155080526 on Build GRCh38
rs943968474 Chr.3: 155080107-155080107 on Build GRCh38
rs201350033 Chr.3: 155080447-155080447 on Build GRCh38
rs1039514479 Chr.3: 155080123-155080123 on Build GRCh38
rs201537443 Chr.3: 155080491-155080491 on Build GRCh38
rs1006210989 Chr.3: 155080360-155080360 on Build GRCh38
rs375270621 Chr.3: 155080126-155080126 on Build GRCh38
rs566767711 Chr.3: 155080439-155080439 on Build GRCh38
rs1016510097 Chr.3: 155080158-155080158 on Build GRCh38
rs954629972 Chr.3: 155080203-155080203 on Build GRCh38
rs35599824 Chr.3: 155080089-155080090 on Build GRCh38
rs1013198356 Chr.3: 155080187-155080187 on Build GRCh38
rs898101940 Chr.3: 155080369-155080369 on Build GRCh38
rs759675701 Chr.3: 155080437-155080437 on Build GRCh38
rs978865925 Chr.3: 155080231-155080231 on Build GRCh38
rs542962863 Chr.3: 155080376-155080376 on Build GRCh38
rs1023119658 Chr.3: 155080222-155080222 on Build GRCh38
rs116563482 Chr.3: 155080492-155080492 on Build GRCh38
rs899685645 Chr.3: 155080128-155080128 on Build GRCh38
rs368302087 Chr.3: 155080283-155080283 on Build GRCh38
rs1048660359 Chr.3: 155080145-155080145 on Build GRCh38
rs202012044 Chr.3: 155080451-155080451 on Build GRCh38
rs956098123 Chr.3: 155080517-155080517 on Build GRCh38
rs139590167 Chr.3: 155080391-155080391 on Build GRCh38
rs1045178329 Chr.3: 155080346-155080346 on Build GRCh38
rs1037680785 Chr.3: 155080521-155080521 on Build GRCh38
rs545132235 Chr.3: 155080269-155080269 on Build GRCh38
rs969325037 Chr.3: 155080223-155080223 on Build GRCh38
rs371903281 Chr.3: 155080447-155080447 on Build GRCh38
rs772570538 Chr.3: 155080396-155080396 on Build GRCh38
rs995261387 Chr.3: 155080136-155080136 on Build GRCh38
rs978427844 Chr.3: 155080404-155080404 on Build GRCh38
rs1032133345 Chr.3: 155080244-155080244 on Build GRCh38
rs975771767 Chr.3: 155080326-155080326 on Build GRCh38
rs776543707 Chr.3: 155080257-155080257 on Build GRCh38
rs1024069423 Chr.3: 155080133-155080133 on Build GRCh38
rs985928046 Chr.3: 155080216-155080216 on Build GRCh38
rs1057377566 Chr.3: 155080313-155080313 on Build GRCh38
rs895513410 Chr.3: 155080398-155080398 on Build GRCh38
rs1003849727 Chr.3: 155080159-155080159 on Build GRCh38
rs76555086 Chr.3: 155080348-155080348 on Build GRCh38
rs200236371 Chr.3: 155080439-155080439 on Build GRCh38
rs956130643 Chr.3: 155080254-155080254 on Build GRCh38
rs943937425 Chr.3: 155080308-155080308 on Build GRCh38
rs147767864 Chr.3: 155080294-155080294 on Build GRCh38
rs948544321 Chr.3: 155080341-155080341 on Build GRCh38
rs947024898 Chr.3: 155080063-155080063 on Build GRCh38
rs987401697 Chr.3: 155080292-155080292 on Build GRCh38
rs774925082 Chr.3: 155080164-155080164 on Build GRCh38
rs934516007 Chr.3: 155080079-155080079 on Build GRCh38
rs550210207 Chr.3: 155080438-155080439 on Build GRCh38
rs199897372 Chr.3: 155080400-155080400 on Build GRCh38
rs896564817 Chr.3: 155080120-155080120 on Build GRCh38
rs955951655 Chr.3: 155080438-155080438 on Build GRCh38
rs780952697 Chr.3: 155080507-155080510 on Build GRCh38
rs201211722 Chr.3: 155080363-155080363 on Build GRCh38
rs1051717445 Chr.3: 155080064-155080064 on Build GRCh38
rs886829105 Chr.3: 155080380-155080380 on Build GRCh38
rs766737887 Chr.3: 155080291-155080291 on Build GRCh38
rs577393872 Chr.3: 155080161-155080161 on Build GRCh38
rs924248486 Chr.3: 155080408-155080408 on Build GRCh38
rs772688175 Chr.3: 155080394-155080395 on Build GRCh38
rs202176433 Chr.3: 155080415-155080415 on Build GRCh38
rs886868132 Chr.3: 155080146-155080146 on Build GRCh38
rs993683387 Chr.3: 155080370-155080370 on Build GRCh38
rs1048235378 Chr.3: 155080352-155080352 on Build GRCh38
rs768550623 Chr.3: 155080183-155080183 on Build GRCh38

View Full Product Details