Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159262611-159263102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCJ
Entrez Gene ID
654502
Gene Name
IQ motif containing J
Gene Aliases
-
UniGene
Hs.570685
Gene Chromosome Location
Chr.3: 159069252-159266307 on Build GRCh38
Transcripts
NM_001042705.2 NM_001042706.2 NM_001197100.1 NM_001197113.1 NM_001197114.1

Gene Symbol
IQCJ-SCHIP1
Entrez Gene ID
100505385
Gene Name
IQCJ-SCHIP1 readthrough
Gene Aliases
-
UniGene
Hs.134665
Gene Chromosome Location
Chr.3: 159069252-159897366 on Build GRCh38
Transcripts
NM_001042705.2 NM_001042706.2 NM_001197100.1 NM_001197113.1 NM_001197114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4114889 c.220C>A p.P74T Chr.3: 159262612-159262612 on Build GRCh38
3005908 c.277G>A p.D93N Chr.3: 159262669-159262669 on Build GRCh38
368161 c.291C>A p.P97P Chr.3: 159262683-159262683 on Build GRCh38
729448 c.236C>G p.S79* Chr.3: 159262628-159262628 on Build GRCh38
3589409 c.234C>T p.S78S Chr.3: 159262626-159262626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558330490 Chr.3: 159262914-159262914 on Build GRCh38
rs778714834 Chr.3: 159262693-159262693 on Build GRCh38
rs1004025625 Chr.3: 159263080-159263080 on Build GRCh38
rs200957112 Chr.3: 159262669-159262669 on Build GRCh38
rs1003361927 Chr.3: 159262941-159262941 on Build GRCh38
rs565719628 Chr.3: 159262656-159262656 on Build GRCh38
rs747475965 Chr.3: 159262918-159262918 on Build GRCh38
rs960627338 Chr.3: 159262801-159262801 on Build GRCh38
rs894427227 Chr.3: 159262960-159262960 on Build GRCh38
rs765699106 Chr.3: 159262662-159262662 on Build GRCh38
rs1012843812 Chr.3: 159262974-159262974 on Build GRCh38
rs781259338 Chr.3: 159262677-159262677 on Build GRCh38
rs556682846 Chr.3: 159262907-159262907 on Build GRCh38
rs916380024 Chr.3: 159262896-159262896 on Build GRCh38
rs750756440 Chr.3: 159262665-159262665 on Build GRCh38
rs911896533 Chr.3: 159263077-159263077 on Build GRCh38
rs1026831017 Chr.3: 159263072-159263072 on Build GRCh38
rs994510916 Chr.3: 159262690-159262690 on Build GRCh38
rs976476045 Chr.3: 159262697-159262697 on Build GRCh38
rs748470596 Chr.3: 159262682-159262682 on Build GRCh38
rs950430023 Chr.3: 159262737-159262737 on Build GRCh38
rs760296981 Chr.3: 159262649-159262649 on Build GRCh38
rs947512288 Chr.3: 159262826-159262826 on Build GRCh38
rs965483400 Chr.3: 159262684-159262684 on Build GRCh38
rs776140887 Chr.3: 159262745-159262745 on Build GRCh38
rs202040350 Chr.3: 159262642-159262642 on Build GRCh38
rs763804161 Chr.3: 159262653-159262653 on Build GRCh38
rs934642466 Chr.3: 159263037-159263037 on Build GRCh38
rs762067284 Chr.3: 159262660-159262660 on Build GRCh38
rs372523426 Chr.3: 159262739-159262739 on Build GRCh38
rs746491447 Chr.3: 159262719-159262719 on Build GRCh38
rs2621309 Chr.3: 159262956-159262956 on Build GRCh38
rs376824973 Chr.3: 159262746-159262746 on Build GRCh38
rs576185290 Chr.3: 159262799-159262799 on Build GRCh38
rs370356054 Chr.3: 159262940-159262940 on Build GRCh38
rs766893360 Chr.3: 159262668-159262668 on Build GRCh38
rs758785677 Chr.3: 159262708-159262708 on Build GRCh38
rs758844693 Chr.3: 159262667-159262667 on Build GRCh38
rs553548768 Chr.3: 159263034-159263034 on Build GRCh38
rs745684810 Chr.3: 159262704-159262704 on Build GRCh38
rs773576881 Chr.3: 159262773-159262773 on Build GRCh38
rs772098597 Chr.3: 159262705-159262705 on Build GRCh38
rs538463372 Chr.3: 159262899-159262899 on Build GRCh38
rs766850514 Chr.3: 159262776-159262776 on Build GRCh38
rs554739583 Chr.3: 159262707-159262707 on Build GRCh38
rs990752428 Chr.3: 159262860-159262860 on Build GRCh38
rs967882490 Chr.3: 159262897-159262897 on Build GRCh38
rs914755529 Chr.3: 159262805-159262805 on Build GRCh38
rs578193932 Chr.3: 159262938-159262938 on Build GRCh38
rs775875030 Chr.3: 159262778-159262790 on Build GRCh38
rs775096379 Chr.3: 159262647-159262647 on Build GRCh38
rs1053156370 Chr.3: 159263073-159263073 on Build GRCh38
rs752118638 Chr.3: 159262780-159262780 on Build GRCh38
rs763338475 Chr.3: 159262774-159262774 on Build GRCh38

View Full Product Details