Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170419160-170419649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN11
Entrez Gene ID
5010
Gene Name
claudin 11
Gene Aliases
OSP, OTM
UniGene
Hs.31595
Gene Chromosome Location
Chr.3: 170418865-170434691 on Build GRCh38
Transcripts
NM_005602.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6424965 c.100T>C p.C34R Chr.3: 170419166-170419166 on Build GRCh38
1133007 c.118A>C p.T40P Chr.3: 170419184-170419184 on Build GRCh38
4650624 c.125G>A p.R42H Chr.3: 170419191-170419191 on Build GRCh38
1693799 c.146C>T p.S49F Chr.3: 170419212-170419212 on Build GRCh38
3392203 c.144C>T p.G48G Chr.3: 170419210-170419210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530895125 Chr.3: 170419342-170419342 on Build GRCh38
rs868677412 Chr.3: 170419286-170419286 on Build GRCh38
rs529226545 Chr.3: 170419418-170419418 on Build GRCh38
rs939217250 Chr.3: 170419360-170419360 on Build GRCh38
rs116599302 Chr.3: 170419333-170419333 on Build GRCh38
rs747280498 Chr.3: 170419309-170419309 on Build GRCh38
rs574187823 Chr.3: 170419271-170419271 on Build GRCh38
rs578074369 Chr.3: 170419303-170419303 on Build GRCh38
rs927302886 Chr.3: 170419450-170419450 on Build GRCh38
rs930261116 Chr.3: 170419558-170419558 on Build GRCh38
rs990846512 Chr.3: 170419466-170419466 on Build GRCh38
rs980153919 Chr.3: 170419362-170419362 on Build GRCh38
rs868176081 Chr.3: 170419347-170419347 on Build GRCh38
rs754977070 Chr.3: 170419330-170419330 on Build GRCh38
rs757839264 Chr.3: 170419245-170419245 on Build GRCh38
rs565889914 Chr.3: 170419559-170419559 on Build GRCh38
rs774933596 Chr.3: 170419196-170419196 on Build GRCh38
rs551587194 Chr.3: 170419591-170419591 on Build GRCh38
rs765462217 Chr.3: 170419258-170419258 on Build GRCh38
rs767191862 Chr.3: 170419185-170419185 on Build GRCh38
rs148546063 Chr.3: 170419572-170419572 on Build GRCh38
rs368299053 Chr.3: 170419307-170419307 on Build GRCh38
rs370405833 Chr.3: 170419282-170419282 on Build GRCh38
rs866940906 Chr.3: 170419268-170419268 on Build GRCh38
rs754297363 Chr.3: 170419229-170419229 on Build GRCh38
rs964354928 Chr.3: 170419215-170419215 on Build GRCh38
rs1038379791 Chr.3: 170419531-170419531 on Build GRCh38
rs973333670 Chr.3: 170419326-170419326 on Build GRCh38
rs763700655 Chr.3: 170419219-170419219 on Build GRCh38
rs117204526 Chr.3: 170419356-170419356 on Build GRCh38
rs891284187 Chr.3: 170419562-170419562 on Build GRCh38
rs550927500 Chr.3: 170419566-170419567 on Build GRCh38
rs372126929 Chr.3: 170419598-170419598 on Build GRCh38
rs765259314 Chr.3: 170419473-170419473 on Build GRCh38
rs941356886 Chr.3: 170419477-170419477 on Build GRCh38
rs1005809286 Chr.3: 170419305-170419305 on Build GRCh38
rs537142512 Chr.3: 170419630-170419630 on Build GRCh38
rs945298208 Chr.3: 170419579-170419579 on Build GRCh38
rs548993188 Chr.3: 170419350-170419350 on Build GRCh38
rs376285070 Chr.3: 170419195-170419195 on Build GRCh38
rs147186361 Chr.3: 170419533-170419533 on Build GRCh38
rs201720518 Chr.3: 170419184-170419184 on Build GRCh38
rs760157251 Chr.3: 170419200-170419200 on Build GRCh38
rs866352454 Chr.3: 170419250-170419250 on Build GRCh38
rs966656759 Chr.3: 170419335-170419335 on Build GRCh38
rs376171338 Chr.3: 170419204-170419204 on Build GRCh38

View Full Product Details