Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2304586-2305075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE28
Entrez Gene ID
57732
Gene Name
zinc finger FYVE-type containing 28
Gene Aliases
LST2, LYST2
UniGene
Hs.292056
Gene Chromosome Location
Chr.4: 2269585-2418698 on Build GRCh38
Transcripts
NM_001172656.1 NM_001172659.1 NM_020972.2 XM_006713900.3 XM_006713902.3 XM_011513540.2 XM_011513541.2 XM_017008502.1 XM_017008503.1 XM_017008504.1 XM_017008505.1 XM_017008506.1 XM_017008507.1 XM_017008508.1 XM_017008509.1 XM_017008510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6545187 c.1479G>A p.V493V Chr.4: 2304861-2304861 on Build GRCh38
6902541 c.1357G>A p.E453K Chr.4: 2304983-2304983 on Build GRCh38
4585014 c.1332G>A p.A444A Chr.4: 2305008-2305008 on Build GRCh38
3428437 c.1711G>A p.G571R Chr.4: 2304629-2304629 on Build GRCh38
5657887 c.1513G>T p.A505S Chr.4: 2304827-2304827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149193016 Chr.4: 2304919-2304919 on Build GRCh38
rs747743202 Chr.4: 2304650-2304650 on Build GRCh38
rs1032797957 Chr.4: 2304816-2304816 on Build GRCh38
rs199748984 Chr.4: 2304727-2304727 on Build GRCh38
rs771123470 Chr.4: 2304722-2304722 on Build GRCh38
rs1054642659 Chr.4: 2305043-2305043 on Build GRCh38
rs967368316 Chr.4: 2304871-2304871 on Build GRCh38
rs764032902 Chr.4: 2305020-2305020 on Build GRCh38
rs780107929 Chr.4: 2304708-2304708 on Build GRCh38
rs971734513 Chr.4: 2304834-2304834 on Build GRCh38
rs767858245 Chr.4: 2304779-2304779 on Build GRCh38
rs757141657 Chr.4: 2305032-2305032 on Build GRCh38
rs140730787 Chr.4: 2304856-2304856 on Build GRCh38
rs746450436 Chr.4: 2305054-2305054 on Build GRCh38
rs761421621 Chr.4: 2304769-2304769 on Build GRCh38
rs543648457 Chr.4: 2304827-2304827 on Build GRCh38
rs776209104 Chr.4: 2304766-2304766 on Build GRCh38
rs753399338 Chr.4: 2304730-2304730 on Build GRCh38
rs924912754 Chr.4: 2305035-2305035 on Build GRCh38
rs766652682 Chr.4: 2305008-2305008 on Build GRCh38
rs763932763 Chr.4: 2304761-2304761 on Build GRCh38
rs772507744 Chr.4: 2304750-2304750 on Build GRCh38
rs972162250 Chr.4: 2304748-2304748 on Build GRCh38
rs753238074 Chr.4: 2304781-2304781 on Build GRCh38
rs1044186391 Chr.4: 2305047-2305047 on Build GRCh38
rs745502239 Chr.4: 2304987-2304987 on Build GRCh38
rs759517543 Chr.4: 2304723-2304723 on Build GRCh38
rs574021846 Chr.4: 2304848-2304848 on Build GRCh38
rs777916676 Chr.4: 2304746-2304746 on Build GRCh38
rs974943231 Chr.4: 2304998-2304998 on Build GRCh38
rs778540821 Chr.4: 2305037-2305037 on Build GRCh38
rs745876877 Chr.4: 2304812-2304812 on Build GRCh38
rs150674026 Chr.4: 2304638-2304638 on Build GRCh38
rs771862468 Chr.4: 2304754-2304754 on Build GRCh38
rs768877205 Chr.4: 2304668-2304668 on Build GRCh38
rs746230136 Chr.4: 2304747-2304747 on Build GRCh38
rs553390216 Chr.4: 2305012-2305012 on Build GRCh38
rs779163239 Chr.4: 2304811-2304811 on Build GRCh38
rs995315824 Chr.4: 2304992-2304992 on Build GRCh38
rs149721712 Chr.4: 2305013-2305013 on Build GRCh38
rs201929618 Chr.4: 2304887-2304887 on Build GRCh38
rs368978945 Chr.4: 2304792-2304792 on Build GRCh38
rs746958223 Chr.4: 2304818-2304818 on Build GRCh38
rs373919471 Chr.4: 2304958-2304958 on Build GRCh38
rs758910028 Chr.4: 2304705-2304705 on Build GRCh38
rs762432812 Chr.4: 2304836-2304836 on Build GRCh38
rs763072682 Chr.4: 2304721-2304721 on Build GRCh38
rs764390010 Chr.4: 2304844-2304844 on Build GRCh38
rs747303556 Chr.4: 2304667-2304667 on Build GRCh38
rs756623156 Chr.4: 2304842-2304842 on Build GRCh38
rs76467215 Chr.4: 2304751-2304751 on Build GRCh38
rs776892210 Chr.4: 2304669-2304669 on Build GRCh38
rs532995191 Chr.4: 2304714-2304714 on Build GRCh38
rs139543749 Chr.4: 2305009-2305009 on Build GRCh38
rs767557756 Chr.4: 2304960-2304960 on Build GRCh38
rs757997294 Chr.4: 2304758-2304758 on Build GRCh38
rs201625946 Chr.4: 2304783-2304783 on Build GRCh38
rs535273084 Chr.4: 2304656-2304656 on Build GRCh38
rs1036327580 Chr.4: 2304674-2304674 on Build GRCh38
rs149353807 Chr.4: 2304686-2304686 on Build GRCh38
rs199832545 Chr.4: 2304612-2304612 on Build GRCh38
rs553099206 Chr.4: 2304840-2304840 on Build GRCh38
rs755432311 Chr.4: 2304735-2304735 on Build GRCh38
rs376846150 Chr.4: 2304786-2304786 on Build GRCh38
rs368035767 Chr.4: 2304690-2304690 on Build GRCh38
rs371841906 Chr.4: 2304719-2304719 on Build GRCh38
rs372218217 Chr.4: 2304670-2304670 on Build GRCh38
rs146596546 Chr.4: 2304849-2304849 on Build GRCh38
rs541977517 Chr.4: 2304915-2304915 on Build GRCh38
rs754686050 Chr.4: 2304647-2304647 on Build GRCh38
rs141908652 Chr.4: 2304610-2304610 on Build GRCh38
rs151169783 Chr.4: 2304772-2304772 on Build GRCh38
rs753704957 Chr.4: 2305031-2305031 on Build GRCh38
rs751593287 Chr.4: 2304918-2304918 on Build GRCh38
rs200326536 Chr.4: 2304696-2304696 on Build GRCh38
rs749573323 Chr.4: 2304821-2304821 on Build GRCh38
rs201070288 Chr.4: 2304611-2304611 on Build GRCh38
rs138443865 Chr.4: 2304888-2304888 on Build GRCh38
rs773465828 Chr.4: 2305005-2305005 on Build GRCh38
rs267600129 Chr.4: 2304652-2304652 on Build GRCh38
rs762325186 Chr.4: 2304903-2304903 on Build GRCh38
rs773120296 Chr.4: 2304676-2304676 on Build GRCh38
rs751415597 Chr.4: 2304687-2304687 on Build GRCh38
rs745560403 Chr.4: 2305040-2305040 on Build GRCh38
rs755698779 Chr.4: 2304698-2304698 on Build GRCh38
rs527410907 Chr.4: 2304657-2304657 on Build GRCh38
rs746809534 Chr.4: 2304955-2304955 on Build GRCh38
rs988422580 Chr.4: 2304763-2304763 on Build GRCh38
rs534718019 Chr.4: 2304953-2304953 on Build GRCh38
rs1017601144 Chr.4: 2304608-2304608 on Build GRCh38
rs906905954 Chr.4: 2304966-2304966 on Build GRCh38
rs889267840 Chr.4: 2304717-2304717 on Build GRCh38
rs757504353 Chr.4: 2304793-2304793 on Build GRCh38
rs927271051 Chr.4: 2305027-2305027 on Build GRCh38
rs186217426 Chr.4: 2304648-2304648 on Build GRCh38
rs150614771 Chr.4: 2304629-2304629 on Build GRCh38
rs141400000 Chr.4: 2304944-2304944 on Build GRCh38
rs984629958 Chr.4: 2304627-2304627 on Build GRCh38
rs925435122 Chr.4: 2304798-2304798 on Build GRCh38
rs748917099 Chr.4: 2304925-2304925 on Build GRCh38
rs758470767 Chr.4: 2304855-2304855 on Build GRCh38
rs146868997 Chr.4: 2304854-2304854 on Build GRCh38
rs768587282 Chr.4: 2304863-2304863 on Build GRCh38
rs755483258 Chr.4: 2304777-2304777 on Build GRCh38
rs756375798 Chr.4: 2304744-2304744 on Build GRCh38
rs753184462 Chr.4: 2304841-2304841 on Build GRCh38
rs780016186 Chr.4: 2304916-2304916 on Build GRCh38
rs748684512 Chr.4: 2304991-2304991 on Build GRCh38
rs375202183 Chr.4: 2304720-2304720 on Build GRCh38
rs374864545 Chr.4: 2304617-2304617 on Build GRCh38
rs564089384 Chr.4: 2305046-2305046 on Build GRCh38
rs752680562 Chr.4: 2304963-2304963 on Build GRCh38
rs369893273 Chr.4: 2304742-2304742 on Build GRCh38
rs377155128 Chr.4: 2304971-2304971 on Build GRCh38
rs781645956 Chr.4: 2304743-2304743 on Build GRCh38
rs754267408 Chr.4: 2304896-2304896 on Build GRCh38
rs765741813 Chr.4: 2304949-2304949 on Build GRCh38
rs138678197 Chr.4: 2304630-2304630 on Build GRCh38
rs769071523 Chr.4: 2304709-2304709 on Build GRCh38
rs906850002 Chr.4: 2305049-2305049 on Build GRCh38
rs892794558 Chr.4: 2304820-2304820 on Build GRCh38
rs200144782 Chr.4: 2304990-2304990 on Build GRCh38
rs777365361 Chr.4: 2304699-2304699 on Build GRCh38
rs756872061 Chr.4: 2304984-2304984 on Build GRCh38
rs749046922 Chr.4: 2304973-2304973 on Build GRCh38
rs377628449 Chr.4: 2304985-2304985 on Build GRCh38
rs369279005 Chr.4: 2304950-2304950 on Build GRCh38
rs372232663 Chr.4: 2304756-2304756 on Build GRCh38
rs116535710 Chr.4: 2304771-2304771 on Build GRCh38
rs758071176 Chr.4: 2305044-2305044 on Build GRCh38
rs772918892 Chr.4: 2304886-2304886 on Build GRCh38
rs776479164 Chr.4: 2305004-2305004 on Build GRCh38
rs61732926 Chr.4: 2305006-2305006 on Build GRCh38
rs557128139 Chr.4: 2304828-2304828 on Build GRCh38
rs557164903 Chr.4: 2305001-2305001 on Build GRCh38
rs765213994 Chr.4: 2304626-2304626 on Build GRCh38
rs140095568 Chr.4: 2304873-2304873 on Build GRCh38
rs779218553 Chr.4: 2304850-2304850 on Build GRCh38
rs762982143 Chr.4: 2304677-2304677 on Build GRCh38
rs780425997 Chr.4: 2304659-2304659 on Build GRCh38
rs772727363 Chr.4: 2304830-2304830 on Build GRCh38
rs765688641 Chr.4: 2304910-2304910 on Build GRCh38
rs771744807 Chr.4: 2304989-2304989 on Build GRCh38
rs934792203 Chr.4: 2305039-2305039 on Build GRCh38
rs563458821 Chr.4: 2304920-2304920 on Build GRCh38
rs147422794 Chr.4: 2304713-2304713 on Build GRCh38
rs770585256 Chr.4: 2304729-2304729 on Build GRCh38
rs761892791 Chr.4: 2304624-2304624 on Build GRCh38
rs770579351 Chr.4: 2304930-2304930 on Build GRCh38
rs777343128 Chr.4: 2304947-2304947 on Build GRCh38
rs547198907 Chr.4: 2304688-2304688 on Build GRCh38
rs570859638 Chr.4: 2304643-2304643 on Build GRCh38
rs772059250 Chr.4: 2304813-2304813 on Build GRCh38
rs747823964 Chr.4: 2304642-2304642 on Build GRCh38
rs144477285 Chr.4: 2305014-2305014 on Build GRCh38
rs771695667 Chr.4: 2304945-2304945 on Build GRCh38
rs61910680 Chr.4: 2304951-2304951 on Build GRCh38
rs200468587 Chr.4: 2304815-2304815 on Build GRCh38
rs114467899 Chr.4: 2304969-2304969 on Build GRCh38
rs145212126 Chr.4: 2304731-2304731 on Build GRCh38
rs774644261 Chr.4: 2304733-2304733 on Build GRCh38
rs113256779 Chr.4: 2304935-2304935 on Build GRCh38
rs764626382 Chr.4: 2304891-2304891 on Build GRCh38
rs755716669 Chr.4: 2304654-2304654 on Build GRCh38
rs143269293 Chr.4: 2304921-2304921 on Build GRCh38
rs377484299 Chr.4: 2304889-2304889 on Build GRCh38
rs147201935 Chr.4: 2304732-2304732 on Build GRCh38
rs780478868 Chr.4: 2304707-2304707 on Build GRCh38
rs373104966 Chr.4: 2304872-2304872 on Build GRCh38
rs139789544 Chr.4: 2304726-2304726 on Build GRCh38

View Full Product Details