Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56584357-56584756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THEGL
Entrez Gene ID
100506564
Gene Name
theg spermatid protein-like
Gene Aliases
-
UniGene
Hs.573073
Gene Chromosome Location
Chr.4: 56530575-56626214 on Build GRCh38
Transcripts
NM_001256475.1 XM_011534357.1 XM_011534361.2 XM_011534363.2 XM_011534364.1 XM_011534365.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3392919 c.916C>T p.R306W Chr.4: 56584552-56584552 on Build GRCh38
6502204 c.932C>T p.S311L Chr.4: 56584568-56584568 on Build GRCh38
5509353 c.892C>G p.L298V Chr.4: 56584528-56584528 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773293368 Chr.4: 56584533-56584533 on Build GRCh38
rs923030931 Chr.4: 56584438-56584438 on Build GRCh38
rs779772675 Chr.4: 56584505-56584505 on Build GRCh38
rs952877800 Chr.4: 56584634-56584634 on Build GRCh38
rs188780027 Chr.4: 56584579-56584579 on Build GRCh38
rs975826736 Chr.4: 56584431-56584431 on Build GRCh38
rs549227551 Chr.4: 56584531-56584531 on Build GRCh38
rs4527533 Chr.4: 56584454-56584454 on Build GRCh38
rs994314810 Chr.4: 56584608-56584608 on Build GRCh38
rs1044314232 Chr.4: 56584523-56584523 on Build GRCh38
rs1003299996 Chr.4: 56584532-56584532 on Build GRCh38
rs545577817 Chr.4: 56584401-56584401 on Build GRCh38
rs564524924 Chr.4: 56584408-56584408 on Build GRCh38
rs764531241 Chr.4: 56584497-56584497 on Build GRCh38
rs533472675 Chr.4: 56584409-56584409 on Build GRCh38
rs934470731 Chr.4: 56584452-56584452 on Build GRCh38
rs754329736 Chr.4: 56584555-56584555 on Build GRCh38
rs1054309388 Chr.4: 56584547-56584547 on Build GRCh38
rs550336393 Chr.4: 56584426-56584426 on Build GRCh38
rs964384529 Chr.4: 56584699-56584699 on Build GRCh38
rs115989686 Chr.4: 56584647-56584647 on Build GRCh38
rs56255039 Chr.4: 56584716-56584716 on Build GRCh38
rs749180467 Chr.4: 56584526-56584526 on Build GRCh38
rs183368270 Chr.4: 56584485-56584485 on Build GRCh38
rs760720974 Chr.4: 56584548-56584548 on Build GRCh38
rs765524605 Chr.4: 56584599-56584599 on Build GRCh38
rs147762862 Chr.4: 56584623-56584623 on Build GRCh38
rs923010894 Chr.4: 56584717-56584717 on Build GRCh38
rs771036024 Chr.4: 56584552-56584552 on Build GRCh38
rs4345238 Chr.4: 56584682-56584682 on Build GRCh38
rs149702721 Chr.4: 56584553-56584553 on Build GRCh38
rs944391353 Chr.4: 56584480-56584480 on Build GRCh38
rs1040062019 Chr.4: 56584482-56584482 on Build GRCh38
rs991449460 Chr.4: 56584645-56584645 on Build GRCh38
rs756443299 Chr.4: 56584459-56584459 on Build GRCh38
rs543044401 Chr.4: 56584707-56584707 on Build GRCh38

View Full Product Details