Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70247658-70248191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN3
Entrez Gene ID
1448
Gene Name
casein kappa
Gene Aliases
CNS10, CSN10, CSNK, KCA
UniGene
Hs.54415
Gene Chromosome Location
Chr.4: 70238370-70251432 on Build GRCh38
Transcripts
NM_005212.2 XM_011531620.2 XM_017007761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753814 c.81A>T p.Q27H Chr.4: 70247844-70247844 on Build GRCh38
4931498 c.55G>T p.A19S Chr.4: 70247818-70247818 on Build GRCh38
3357826 c.85G>T p.A29S Chr.4: 70247848-70247848 on Build GRCh38
340708 c.55-1G>T p.? Chr.4: 70247817-70247817 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1037748588 Chr.4: 70248036-70248036 on Build GRCh38
rs200728765 Chr.4: 70247856-70247856 on Build GRCh38
rs758086400 Chr.4: 70247839-70247839 on Build GRCh38
rs1024648019 Chr.4: 70247914-70247914 on Build GRCh38
rs200725753 Chr.4: 70247771-70247772 on Build GRCh38
rs897377501 Chr.4: 70247735-70247735 on Build GRCh38
rs60315013 Chr.4: 70247763-70247763 on Build GRCh38
rs959186621 Chr.4: 70247801-70247801 on Build GRCh38
rs747396950 Chr.4: 70247896-70247896 on Build GRCh38
rs756999457 Chr.4: 70247851-70247851 on Build GRCh38
rs557508160 Chr.4: 70247860-70247860 on Build GRCh38
rs748396997 Chr.4: 70247779-70247779 on Build GRCh38
rs767022563 Chr.4: 70247800-70247800 on Build GRCh38
rs540968069 Chr.4: 70248066-70248066 on Build GRCh38
rs777314287 Chr.4: 70247898-70247898 on Build GRCh38
rs773263771 Chr.4: 70247888-70247888 on Build GRCh38
rs552991081 Chr.4: 70247818-70247818 on Build GRCh38
rs62308385 Chr.4: 70248031-70248031 on Build GRCh38
rs985497672 Chr.4: 70248025-70248025 on Build GRCh38
rs1032589744 Chr.4: 70248022-70248022 on Build GRCh38
rs763589871 Chr.4: 70247845-70247845 on Build GRCh38
rs374847559 Chr.4: 70247900-70247900 on Build GRCh38
rs1041038608 Chr.4: 70247715-70247715 on Build GRCh38
rs761051865 Chr.4: 70247791-70247791 on Build GRCh38
rs910534177 Chr.4: 70247977-70247977 on Build GRCh38
rs779036056 Chr.4: 70247874-70247874 on Build GRCh38
rs1026541443 Chr.4: 70247760-70247760 on Build GRCh38
rs888655505 Chr.4: 70247773-70247773 on Build GRCh38
rs373296060 Chr.4: 70247714-70247714 on Build GRCh38
rs944071315 Chr.4: 70247711-70247711 on Build GRCh38
rs543120754 Chr.4: 70247929-70247929 on Build GRCh38
rs976681622 Chr.4: 70248090-70248090 on Build GRCh38
rs930244947 Chr.4: 70247755-70247755 on Build GRCh38
rs772341345 Chr.4: 70247882-70247882 on Build GRCh38
rs1047909216 Chr.4: 70248146-70248146 on Build GRCh38
rs957921667 Chr.4: 70248023-70248023 on Build GRCh38
rs1049005921 Chr.4: 70247766-70247766 on Build GRCh38
rs773517165 Chr.4: 70248038-70248038 on Build GRCh38
rs565392292 Chr.4: 70248068-70248068 on Build GRCh38
rs899347557 Chr.4: 70248045-70248045 on Build GRCh38
rs10518071 Chr.4: 70247916-70247916 on Build GRCh38
rs112136060 Chr.4: 70247782-70247782 on Build GRCh38
rs796849455 Chr.4: 70247771-70247771 on Build GRCh38
rs764768811 Chr.4: 70247827-70247827 on Build GRCh38
rs143568877 Chr.4: 70247947-70247947 on Build GRCh38
rs906824617 Chr.4: 70248165-70248165 on Build GRCh38
rs918602501 Chr.4: 70248099-70248099 on Build GRCh38
rs745775209 Chr.4: 70247857-70247857 on Build GRCh38
rs368533318 Chr.4: 70247846-70247846 on Build GRCh38
rs112798924 Chr.4: 70247952-70247952 on Build GRCh38
rs772368304 Chr.4: 70247891-70247892 on Build GRCh38
rs759162954 Chr.4: 70247821-70247821 on Build GRCh38
rs773638196 Chr.4: 70247787-70247787 on Build GRCh38
rs141141270 Chr.4: 70247836-70247836 on Build GRCh38
rs1033450822 Chr.4: 70247784-70247784 on Build GRCh38
rs930211853 Chr.4: 70248145-70248145 on Build GRCh38
rs371460325 Chr.4: 70247879-70247879 on Build GRCh38
rs987526636 Chr.4: 70247964-70247964 on Build GRCh38

View Full Product Details