Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70249240-70249769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN3
Entrez Gene ID
1448
Gene Name
casein kappa
Gene Aliases
CNS10, CSN10, CSNK, KCA
UniGene
Hs.54415
Gene Chromosome Location
Chr.4: 70238370-70251432 on Build GRCh38
Transcripts
NM_005212.2 XM_011531620.2 XM_017007761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1694482 c.379G>A p.D127N Chr.4: 70249289-70249289 on Build GRCh38
6894954 c.449C>T p.P150L Chr.4: 70249359-70249359 on Build GRCh38
4125733 c.543G>A p.T181T Chr.4: 70249453-70249453 on Build GRCh38
7215524 c.426T>A p.P142P Chr.4: 70249336-70249336 on Build GRCh38
4675129 c.360delC p.P122fs*8 Chr.4: 70249270-70249270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116485404 Chr.4: 70249727-70249727 on Build GRCh38
rs982484008 Chr.4: 70249691-70249691 on Build GRCh38
rs79660442 Chr.4: 70249475-70249475 on Build GRCh38
rs114625660 Chr.4: 70249613-70249613 on Build GRCh38
rs749896746 Chr.4: 70249483-70249483 on Build GRCh38
rs753256991 Chr.4: 70249361-70249361 on Build GRCh38
rs760767613 Chr.4: 70249344-70249344 on Build GRCh38
rs780751662 Chr.4: 70249402-70249402 on Build GRCh38
rs769771562 Chr.4: 70249407-70249407 on Build GRCh38
rs3775737 Chr.4: 70249526-70249526 on Build GRCh38
rs754185055 Chr.4: 70249286-70249286 on Build GRCh38
rs745801637 Chr.4: 70249366-70249366 on Build GRCh38
rs556456544 Chr.4: 70249525-70249525 on Build GRCh38
rs199690256 Chr.4: 70249287-70249287 on Build GRCh38
rs779814487 Chr.4: 70249490-70249490 on Build GRCh38
rs751057733 Chr.4: 70249383-70249383 on Build GRCh38
rs775548973 Chr.4: 70249270-70249270 on Build GRCh38
rs933922789 Chr.4: 70249403-70249403 on Build GRCh38
rs908434413 Chr.4: 70249692-70249692 on Build GRCh38
rs1046912064 Chr.4: 70249365-70249365 on Build GRCh38
rs375519684 Chr.4: 70249566-70249566 on Build GRCh38
rs765579689 Chr.4: 70249360-70249360 on Build GRCh38
rs752077228 Chr.4: 70249462-70249462 on Build GRCh38
rs1012899041 Chr.4: 70249280-70249280 on Build GRCh38
rs778306360 Chr.4: 70249363-70249363 on Build GRCh38
rs770525492 Chr.4: 70249322-70249322 on Build GRCh38
rs754067840 Chr.4: 70249357-70249357 on Build GRCh38
rs115386969 Chr.4: 70249328-70249328 on Build GRCh38
rs759931928 Chr.4: 70249359-70249359 on Build GRCh38
rs79364398 Chr.4: 70249474-70249474 on Build GRCh38
rs3775738 Chr.4: 70249435-70249435 on Build GRCh38
rs201406795 Chr.4: 70249411-70249411 on Build GRCh38
rs765381118 Chr.4: 70249452-70249452 on Build GRCh38
rs776302059 Chr.4: 70249323-70249323 on Build GRCh38
rs867368852 Chr.4: 70249437-70249437 on Build GRCh38
rs749116926 Chr.4: 70249497-70249497 on Build GRCh38
rs766555323 Chr.4: 70249285-70249285 on Build GRCh38
rs371221054 Chr.4: 70249479-70249479 on Build GRCh38
rs942469297 Chr.4: 70249370-70249370 on Build GRCh38
rs999097567 Chr.4: 70249506-70249506 on Build GRCh38
rs75883824 Chr.4: 70249731-70249731 on Build GRCh38
rs767703318 Chr.4: 70249274-70249274 on Build GRCh38
rs777129939 Chr.4: 70249316-70249316 on Build GRCh38
rs748620098 Chr.4: 70249295-70249295 on Build GRCh38
rs150059997 Chr.4: 70249508-70249508 on Build GRCh38
rs760715639 Chr.4: 70249429-70249429 on Build GRCh38
rs769684427 Chr.4: 70249333-70249333 on Build GRCh38
rs763259161 Chr.4: 70249454-70249454 on Build GRCh38
rs748828100 Chr.4: 70249741-70249741 on Build GRCh38
rs768656211 Chr.4: 70249339-70249339 on Build GRCh38
rs866976977 Chr.4: 70249364-70249364 on Build GRCh38
rs746465509 Chr.4: 70249318-70249318 on Build GRCh38
rs200499713 Chr.4: 70249291-70249291 on Build GRCh38
rs922267381 Chr.4: 70249401-70249401 on Build GRCh38
rs146477730 Chr.4: 70249398-70249398 on Build GRCh38
rs76463522 Chr.4: 70249642-70249642 on Build GRCh38
rs1012277298 Chr.4: 70249657-70249657 on Build GRCh38
rs765290571 Chr.4: 70249269-70249269 on Build GRCh38
rs3822164 Chr.4: 70249615-70249615 on Build GRCh38
rs774296649 Chr.4: 70249418-70249418 on Build GRCh38
rs941260870 Chr.4: 70249706-70249706 on Build GRCh38
rs6811366 Chr.4: 70249726-70249726 on Build GRCh38
rs751197924 Chr.4: 70249464-70249464 on Build GRCh38
rs774924280 Chr.4: 70249737-70249737 on Build GRCh38
rs3775740 Chr.4: 70249336-70249336 on Build GRCh38
rs1046964673 Chr.4: 70249564-70249564 on Build GRCh38
rs764562051 Chr.4: 70249457-70249457 on Build GRCh38
rs745513111 Chr.4: 70249405-70249405 on Build GRCh38
rs762617305 Chr.4: 70249270-70249270 on Build GRCh38
rs762026523 Chr.4: 70249272-70249272 on Build GRCh38
rs6811318 Chr.4: 70249659-70249659 on Build GRCh38
rs756304614 Chr.4: 70249279-70249279 on Build GRCh38
rs955246816 Chr.4: 70249675-70249675 on Build GRCh38
rs766530286 Chr.4: 70249345-70249345 on Build GRCh38
rs776633396 Chr.4: 70249438-70249438 on Build GRCh38
rs267600233 Chr.4: 70249408-70249408 on Build GRCh38
rs908341876 Chr.4: 70249572-70249572 on Build GRCh38
rs759340303 Chr.4: 70249266-70249266 on Build GRCh38
rs903982544 Chr.4: 70249694-70249694 on Build GRCh38
rs187096108 Chr.4: 70249489-70249489 on Build GRCh38
rs765976654 Chr.4: 70249424-70249425 on Build GRCh38
rs758916271 Chr.4: 70249362-70249362 on Build GRCh38
rs144706700 Chr.4: 70249442-70249442 on Build GRCh38
rs778755160 Chr.4: 70249509-70249509 on Build GRCh38
rs753060878 Chr.4: 70249453-70249453 on Build GRCh38
rs3775739 Chr.4: 70249343-70249343 on Build GRCh38
rs757968238 Chr.4: 70249311-70249311 on Build GRCh38
rs368478632 Chr.4: 70249459-70249459 on Build GRCh38
rs191548817 Chr.4: 70249632-70249632 on Build GRCh38

View Full Product Details