Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99336351-99336892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1C
Entrez Gene ID
126
Gene Name
alcohol dehydrogenase 1C (class I), gamma polypeptide
Gene Aliases
ADH3
UniGene
Hs.654537
Gene Chromosome Location
Chr.4: 99336492-99353045 on Build GRCh38
Transcripts
NM_000669.4 NR_133005.1 XM_011531588.2 XM_011531589.1

SNP Detail

SNP ID Chromosome Location
rs749054164 Chr.4: 99336787-99336787 on Build GRCh38
rs760170468 Chr.4: 99336765-99336765 on Build GRCh38
rs1057000383 Chr.4: 99336401-99336401 on Build GRCh38
rs552125685 Chr.4: 99336672-99336672 on Build GRCh38
rs1056946056 Chr.4: 99336712-99336712 on Build GRCh38
rs1015743100 Chr.4: 99336497-99336497 on Build GRCh38
rs986124965 Chr.4: 99336605-99336605 on Build GRCh38
rs926909829 Chr.4: 99336648-99336648 on Build GRCh38
rs978473243 Chr.4: 99336833-99336833 on Build GRCh38
rs549453609 Chr.4: 99336739-99336739 on Build GRCh38
rs984529564 Chr.4: 99336572-99336572 on Build GRCh38
rs941522428 Chr.4: 99336621-99336621 on Build GRCh38
rs777871146 Chr.4: 99336769-99336769 on Build GRCh38
rs761509405 Chr.4: 99336813-99336813 on Build GRCh38
rs1007912351 Chr.4: 99336539-99336539 on Build GRCh38
rs1022594445 Chr.4: 99336463-99336463 on Build GRCh38
rs757669841 Chr.4: 99336425-99336425 on Build GRCh38
rs760256176 Chr.4: 99336820-99336820 on Build GRCh38
rs745763146 Chr.4: 99336741-99336741 on Build GRCh38
rs372063372 Chr.4: 99336696-99336696 on Build GRCh38
rs370818269 Chr.4: 99336766-99336766 on Build GRCh38
rs770912952 Chr.4: 99336861-99336861 on Build GRCh38
rs372991001 Chr.4: 99336725-99336725 on Build GRCh38
rs557637110 Chr.4: 99336832-99336832 on Build GRCh38
rs888391463 Chr.4: 99336745-99336745 on Build GRCh38
rs954204814 Chr.4: 99336783-99336783 on Build GRCh38
rs780128692 Chr.4: 99336728-99336728 on Build GRCh38
rs923992424 Chr.4: 99336658-99336658 on Build GRCh38
rs55808188 Chr.4: 99336816-99336816 on Build GRCh38
rs67420531 Chr.4: 99336772-99336772 on Build GRCh38
rs961142989 Chr.4: 99336543-99336543 on Build GRCh38
rs746455020 Chr.4: 99336492-99336492 on Build GRCh38
rs2298753 Chr.4: 99336750-99336750 on Build GRCh38
rs966326251 Chr.4: 99336549-99336549 on Build GRCh38
rs980973286 Chr.4: 99336864-99336864 on Build GRCh38
rs918884961 Chr.4: 99336565-99336565 on Build GRCh38
rs77761914 Chr.4: 99336764-99336764 on Build GRCh38
rs915530114 Chr.4: 99336614-99336614 on Build GRCh38
rs577743501 Chr.4: 99336441-99336442 on Build GRCh38
rs368254860 Chr.4: 99336664-99336664 on Build GRCh38
rs757354662 Chr.4: 99336771-99336771 on Build GRCh38
rs998675670 Chr.4: 99336391-99336391 on Build GRCh38
rs1042419301 Chr.4: 99336634-99336634 on Build GRCh38
rs1009568699 Chr.4: 99336472-99336472 on Build GRCh38
rs774484394 Chr.4: 99336767-99336767 on Build GRCh38
rs1012840182 Chr.4: 99336440-99336440 on Build GRCh38
rs922793294 Chr.4: 99336564-99336564 on Build GRCh38
rs756205530 Chr.4: 99336652-99336652 on Build GRCh38
rs566227161 Chr.4: 99336568-99336568 on Build GRCh38
rs768434307 Chr.4: 99336798-99336798 on Build GRCh38
rs758570528 Chr.4: 99336743-99336744 on Build GRCh38
rs761235124 Chr.4: 99336761-99336761 on Build GRCh38
rs537651923 Chr.4: 99336715-99336715 on Build GRCh38
rs375726509 Chr.4: 99336786-99336786 on Build GRCh38
rs939565863 Chr.4: 99336654-99336654 on Build GRCh38
rs376262520 Chr.4: 99336738-99336738 on Build GRCh38
rs950347877 Chr.4: 99336849-99336849 on Build GRCh38
rs767010428 Chr.4: 99336705-99336705 on Build GRCh38
rs778052585 Chr.4: 99336729-99336729 on Build GRCh38
rs768255034 Chr.4: 99336758-99336758 on Build GRCh38
rs189718786 Chr.4: 99336486-99336486 on Build GRCh38
rs572227882 Chr.4: 99336759-99336759 on Build GRCh38
rs533708228 Chr.4: 99336632-99336632 on Build GRCh38
rs1612735 Chr.4: 99336850-99336850 on Build GRCh38
rs185034778 Chr.4: 99336800-99336800 on Build GRCh38
rs976861329 Chr.4: 99336561-99336561 on Build GRCh38
rs905217984 Chr.4: 99336387-99336387 on Build GRCh38
rs751734120 Chr.4: 99336770-99336770 on Build GRCh38
rs79502992 Chr.4: 99336744-99336744 on Build GRCh38
rs772890172 Chr.4: 99336819-99336819 on Build GRCh38
rs375850790 Chr.4: 99336708-99336708 on Build GRCh38
rs182066673 Chr.4: 99336562-99336562 on Build GRCh38
rs369280217 Chr.4: 99336720-99336720 on Build GRCh38
rs77988166 Chr.4: 99336747-99336747 on Build GRCh38

View Full Product Details