Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99344721-99345245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1C
Entrez Gene ID
126
Gene Name
alcohol dehydrogenase 1C (class I), gamma polypeptide
Gene Aliases
ADH3
UniGene
Hs.654537
Gene Chromosome Location
Chr.4: 99336492-99353045 on Build GRCh38
Transcripts
NM_000669.4 NR_133005.1 XM_011531588.2 XM_011531589.1

SNP Detail

SNP ID Chromosome Location
rs537229023 Chr.4: 99344843-99344843 on Build GRCh38
rs551254289 Chr.4: 99344792-99344792 on Build GRCh38
rs757356943 Chr.4: 99344932-99344932 on Build GRCh38
rs756812159 Chr.4: 99345163-99345163 on Build GRCh38
rs755985172 Chr.4: 99344866-99344866 on Build GRCh38
rs753685476 Chr.4: 99344954-99344954 on Build GRCh38
rs768392939 Chr.4: 99344814-99344814 on Build GRCh38
rs752593323 Chr.4: 99345126-99345126 on Build GRCh38
rs376551492 Chr.4: 99345005-99345005 on Build GRCh38
rs2241894 Chr.4: 99344976-99344976 on Build GRCh38
rs780641849 Chr.4: 99345076-99345076 on Build GRCh38
rs926904082 Chr.4: 99344753-99344753 on Build GRCh38
rs753947779 Chr.4: 99345115-99345115 on Build GRCh38
rs766776134 Chr.4: 99344983-99344983 on Build GRCh38
rs375271816 Chr.4: 99345003-99345003 on Build GRCh38
rs773255566 Chr.4: 99345047-99345073 on Build GRCh38
rs779842455 Chr.4: 99344953-99344953 on Build GRCh38
rs769558836 Chr.4: 99344999-99345001 on Build GRCh38
rs770280294 Chr.4: 99345068-99345068 on Build GRCh38
rs532369179 Chr.4: 99345027-99345027 on Build GRCh38
rs79813519 Chr.4: 99345099-99345099 on Build GRCh38
rs778742207 Chr.4: 99344965-99344965 on Build GRCh38
rs79372027 Chr.4: 99345000-99345000 on Build GRCh38
rs36050634 Chr.4: 99344751-99344751 on Build GRCh38
rs571261376 Chr.4: 99344760-99344760 on Build GRCh38
rs78241133 Chr.4: 99344999-99344999 on Build GRCh38
rs762790389 Chr.4: 99345180-99345180 on Build GRCh38
rs745776463 Chr.4: 99344865-99344865 on Build GRCh38
rs759583397 Chr.4: 99345122-99345122 on Build GRCh38
rs55717907 Chr.4: 99344907-99344907 on Build GRCh38
rs759712864 Chr.4: 99344769-99344769 on Build GRCh38
rs764292389 Chr.4: 99344851-99344851 on Build GRCh38
rs1693426 Chr.4: 99345173-99345173 on Build GRCh38
rs774576021 Chr.4: 99345010-99345010 on Build GRCh38
rs541924884 Chr.4: 99344938-99344938 on Build GRCh38
rs764313136 Chr.4: 99344917-99344917 on Build GRCh38
rs534676738 Chr.4: 99344885-99344885 on Build GRCh38
rs761399654 Chr.4: 99344827-99344827 on Build GRCh38
rs35733876 Chr.4: 99344773-99344773 on Build GRCh38
rs907795945 Chr.4: 99344799-99344799 on Build GRCh38
rs283411 Chr.4: 99344800-99344800 on Build GRCh38
rs773244842 Chr.4: 99345100-99345100 on Build GRCh38
rs1693425 Chr.4: 99344955-99344955 on Build GRCh38
rs750441233 Chr.4: 99344947-99344947 on Build GRCh38
rs548939369 Chr.4: 99344844-99344844 on Build GRCh38
rs750574929 Chr.4: 99345015-99345015 on Build GRCh38
rs567301265 Chr.4: 99345207-99345207 on Build GRCh38
rs765298517 Chr.4: 99344747-99344747 on Build GRCh38
rs371660994 Chr.4: 99345014-99345014 on Build GRCh38
rs370819059 Chr.4: 99344833-99344833 on Build GRCh38
rs774092660 Chr.4: 99344896-99344896 on Build GRCh38
rs937114232 Chr.4: 99344818-99344818 on Build GRCh38
rs763210161 Chr.4: 99344912-99344912 on Build GRCh38
rs748517793 Chr.4: 99345203-99345203 on Build GRCh38
rs775034716 Chr.4: 99344812-99344812 on Build GRCh38
rs771826482 Chr.4: 99344995-99344995 on Build GRCh38
rs752790819 Chr.4: 99344846-99344846 on Build GRCh38
rs77285688 Chr.4: 99344957-99344957 on Build GRCh38
rs774766061 Chr.4: 99345078-99345078 on Build GRCh38
rs778903541 Chr.4: 99345043-99345043 on Build GRCh38
rs766323345 Chr.4: 99345111-99345111 on Build GRCh38
rs1789915 Chr.4: 99345214-99345214 on Build GRCh38
rs75861024 Chr.4: 99344901-99344901 on Build GRCh38
rs75797048 Chr.4: 99345160-99345160 on Build GRCh38
rs1015170364 Chr.4: 99345004-99345004 on Build GRCh38
rs560665811 Chr.4: 99345154-99345154 on Build GRCh38
rs758187825 Chr.4: 99345046-99345046 on Build GRCh38
rs763812033 Chr.4: 99345151-99345151 on Build GRCh38
rs373664235 Chr.4: 99344845-99344845 on Build GRCh38
rs553105935 Chr.4: 99344895-99344895 on Build GRCh38
rs777274828 Chr.4: 99345055-99345055 on Build GRCh38
rs762064113 Chr.4: 99345012-99345012 on Build GRCh38
rs765628134 Chr.4: 99344910-99344910 on Build GRCh38
rs752231216 Chr.4: 99345167-99345167 on Build GRCh38
rs765257888 Chr.4: 99345125-99345125 on Build GRCh38
rs759852027 Chr.4: 99344835-99344835 on Build GRCh38
rs760614352 Chr.4: 99345026-99345026 on Build GRCh38
rs201424907 Chr.4: 99344858-99344858 on Build GRCh38
rs751713187 Chr.4: 99344923-99344923 on Build GRCh38
rs745379524 Chr.4: 99345165-99345165 on Build GRCh38
rs1001108935 Chr.4: 99344873-99344873 on Build GRCh38
rs76624281 Chr.4: 99345040-99345040 on Build GRCh38
rs758067364 Chr.4: 99345135-99345135 on Build GRCh38
rs373660291 Chr.4: 99344826-99344826 on Build GRCh38
rs757269298 Chr.4: 99344860-99344860 on Build GRCh38
rs756144588 Chr.4: 99344950-99344950 on Build GRCh38
rs778298993 Chr.4: 99345070-99345070 on Build GRCh38
rs111652942 Chr.4: 99345158-99345158 on Build GRCh38
rs577668832 Chr.4: 99344898-99344898 on Build GRCh38
rs929337039 Chr.4: 99345080-99345080 on Build GRCh38
rs78904199 Chr.4: 99345066-99345066 on Build GRCh38
rs749020288 Chr.4: 99344813-99344813 on Build GRCh38
rs781110621 Chr.4: 99344864-99344864 on Build GRCh38
rs752507314 Chr.4: 99345045-99345045 on Build GRCh38
rs113554896 Chr.4: 99344911-99344911 on Build GRCh38
rs546707394 Chr.4: 99345219-99345219 on Build GRCh38
rs563584021 Chr.4: 99345107-99345107 on Build GRCh38
rs35725608 Chr.4: 99345006-99345006 on Build GRCh38
rs980989198 Chr.4: 99344752-99344752 on Build GRCh38
rs75760332 Chr.4: 99345086-99345086 on Build GRCh38
rs746538706 Chr.4: 99345057-99345057 on Build GRCh38
rs762152705 Chr.4: 99345094-99345094 on Build GRCh38
rs780614769 Chr.4: 99345164-99345164 on Build GRCh38
rs777029507 Chr.4: 99344904-99344904 on Build GRCh38
rs746492884 Chr.4: 99345002-99345002 on Build GRCh38
rs181982105 Chr.4: 99345077-99345077 on Build GRCh38
rs34195308 Chr.4: 99344933-99344933 on Build GRCh38

View Full Product Details