Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99345062-99345580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1C
Entrez Gene ID
126
Gene Name
alcohol dehydrogenase 1C (class I), gamma polypeptide
Gene Aliases
ADH3
UniGene
Hs.654537
Gene Chromosome Location
Chr.4: 99336492-99353045 on Build GRCh38
Transcripts
NM_000669.4 NR_133005.1 XM_011531588.2 XM_011531589.1

SNP Detail

SNP ID Chromosome Location
rs567301265 Chr.4: 99345207-99345207 on Build GRCh38
rs890586015 Chr.4: 99345309-99345309 on Build GRCh38
rs950733394 Chr.4: 99345363-99345363 on Build GRCh38
rs775497806 Chr.4: 99345251-99345251 on Build GRCh38
rs921092177 Chr.4: 99345435-99345435 on Build GRCh38
rs748517793 Chr.4: 99345203-99345203 on Build GRCh38
rs575220230 Chr.4: 99345336-99345336 on Build GRCh38
rs749950828 Chr.4: 99345283-99345283 on Build GRCh38
rs756812159 Chr.4: 99345163-99345163 on Build GRCh38
rs992545420 Chr.4: 99345382-99345382 on Build GRCh38
rs757127207 Chr.4: 99345277-99345277 on Build GRCh38
rs963546611 Chr.4: 99345333-99345333 on Build GRCh38
rs70956000 Chr.4: 99345322-99345322 on Build GRCh38
rs368717043 Chr.4: 99345345-99345345 on Build GRCh38
rs796462709 Chr.4: 99345548-99345548 on Build GRCh38
rs752593323 Chr.4: 99345126-99345126 on Build GRCh38
rs369178099 Chr.4: 99345250-99345250 on Build GRCh38
rs6834363 Chr.4: 99345351-99345351 on Build GRCh38
rs766323345 Chr.4: 99345111-99345111 on Build GRCh38
rs987890698 Chr.4: 99345394-99345394 on Build GRCh38
rs1789915 Chr.4: 99345214-99345214 on Build GRCh38
rs751345244 Chr.4: 99345264-99345264 on Build GRCh38
rs200651908 Chr.4: 99345326-99345326 on Build GRCh38
rs55830692 Chr.4: 99345497-99345497 on Build GRCh38
rs892129703 Chr.4: 99345262-99345262 on Build GRCh38
rs571410967 Chr.4: 99345266-99345266 on Build GRCh38
rs75797048 Chr.4: 99345160-99345160 on Build GRCh38
rs945036079 Chr.4: 99345495-99345495 on Build GRCh38
rs560665811 Chr.4: 99345154-99345154 on Build GRCh38
rs748609882 Chr.4: 99345303-99345303 on Build GRCh38
rs753947779 Chr.4: 99345115-99345115 on Build GRCh38
rs922184525 Chr.4: 99345482-99345482 on Build GRCh38
rs936529986 Chr.4: 99345446-99345446 on Build GRCh38
rs763812033 Chr.4: 99345151-99345151 on Build GRCh38
rs71600259 Chr.4: 99345325-99345326 on Build GRCh38
rs977557506 Chr.4: 99345427-99345427 on Build GRCh38
rs1662053 Chr.4: 99345426-99345426 on Build GRCh38
rs368647228 Chr.4: 99345400-99345400 on Build GRCh38
rs766270905 Chr.4: 99345259-99345260 on Build GRCh38
rs79813519 Chr.4: 99345099-99345099 on Build GRCh38
rs202000818 Chr.4: 99345242-99345242 on Build GRCh38
rs778042117 Chr.4: 99345304-99345304 on Build GRCh38
rs752231216 Chr.4: 99345167-99345167 on Build GRCh38
rs769699958 Chr.4: 99345484-99345484 on Build GRCh38
rs765257888 Chr.4: 99345125-99345125 on Build GRCh38
rs113872839 Chr.4: 99345408-99345408 on Build GRCh38
rs759673010 Chr.4: 99345247-99345247 on Build GRCh38
rs372619007 Chr.4: 99345294-99345294 on Build GRCh38
rs913839720 Chr.4: 99345483-99345483 on Build GRCh38
rs745379524 Chr.4: 99345165-99345165 on Build GRCh38
rs1054058556 Chr.4: 99345261-99345261 on Build GRCh38
rs779431442 Chr.4: 99345302-99345302 on Build GRCh38
rs771155881 Chr.4: 99345236-99345236 on Build GRCh38
rs762790389 Chr.4: 99345180-99345180 on Build GRCh38
rs758067364 Chr.4: 99345135-99345135 on Build GRCh38
rs759583397 Chr.4: 99345122-99345122 on Build GRCh38
rs111652942 Chr.4: 99345158-99345158 on Build GRCh38
rs899019824 Chr.4: 99345328-99345328 on Build GRCh38
rs1789916 Chr.4: 99345539-99345539 on Build GRCh38
rs556714464 Chr.4: 99345332-99345332 on Build GRCh38
rs747393514 Chr.4: 99345316-99345316 on Build GRCh38
rs911956625 Chr.4: 99345399-99345399 on Build GRCh38
rs1693426 Chr.4: 99345173-99345173 on Build GRCh38
rs546707394 Chr.4: 99345219-99345219 on Build GRCh38
rs763747241 Chr.4: 99345253-99345253 on Build GRCh38
rs563584021 Chr.4: 99345107-99345107 on Build GRCh38
rs75760332 Chr.4: 99345086-99345086 on Build GRCh38
rs1662052 Chr.4: 99345478-99345478 on Build GRCh38
rs762152705 Chr.4: 99345094-99345094 on Build GRCh38
rs890440996 Chr.4: 99345538-99345538 on Build GRCh38
rs936836976 Chr.4: 99345453-99345453 on Build GRCh38
rs780614769 Chr.4: 99345164-99345164 on Build GRCh38
rs1051807729 Chr.4: 99345503-99345503 on Build GRCh38
rs767144899 Chr.4: 99345276-99345276 on Build GRCh38
rs1027005617 Chr.4: 99345359-99345359 on Build GRCh38
rs1046488012 Chr.4: 99345528-99345528 on Build GRCh38
rs372954582 Chr.4: 99345270-99345270 on Build GRCh38
rs773244842 Chr.4: 99345100-99345100 on Build GRCh38
rs540472967 Chr.4: 99345508-99345508 on Build GRCh38
rs866883071 Chr.4: 99345267-99345267 on Build GRCh38

View Full Product Details